Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103629794-103630305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEAL1
Entrez Gene ID
9338
Gene Name
transcription elongation factor A like 1
Gene Aliases
SIIR, WEX9, p21, pp21
UniGene
Hs.95243
Gene Chromosome Location
Chr.X: 103628720-103630953 on Build GRCh38
Transcripts
NM_001006639.1 NM_001006640.1 NM_004780.2

SNP Detail

SNP ID Chromosome Location
rs767267315 Chr.X: 103630192-103630192 on Build GRCh38
rs749064107 Chr.X: 103630108-103630108 on Build GRCh38
rs1052146 Chr.X: 103630158-103630158 on Build GRCh38
rs374247678 Chr.X: 103629950-103629950 on Build GRCh38
rs1030471650 Chr.X: 103630166-103630166 on Build GRCh38
rs777155549 Chr.X: 103630277-103630277 on Build GRCh38
rs184713983 Chr.X: 103629922-103629922 on Build GRCh38
rs751181266 Chr.X: 103630014-103630014 on Build GRCh38
rs113790212 Chr.X: 103629931-103629931 on Build GRCh38
rs766835211 Chr.X: 103629973-103629973 on Build GRCh38
rs113587097 Chr.X: 103629881-103629881 on Build GRCh38
rs200374529 Chr.X: 103630031-103630031 on Build GRCh38
rs749042527 Chr.X: 103629866-103629866 on Build GRCh38
rs1052147 Chr.X: 103630159-103630159 on Build GRCh38
rs775237143 Chr.X: 103630245-103630245 on Build GRCh38
rs768460936 Chr.X: 103630117-103630117 on Build GRCh38
rs779980584 Chr.X: 103630097-103630097 on Build GRCh38
rs750265883 Chr.X: 103630231-103630231 on Build GRCh38
rs772337828 Chr.X: 103630202-103630202 on Build GRCh38
rs187602806 Chr.X: 103630051-103630051 on Build GRCh38
rs34904221 Chr.X: 103630015-103630015 on Build GRCh38
rs770191600 Chr.X: 103629941-103629943 on Build GRCh38
rs34421776 Chr.X: 103629929-103629929 on Build GRCh38
rs773422658 Chr.X: 103630234-103630234 on Build GRCh38
rs763685742 Chr.X: 103629998-103629998 on Build GRCh38
rs780665016 Chr.X: 103630018-103630018 on Build GRCh38
rs1033618962 Chr.X: 103630032-103630032 on Build GRCh38
rs761252179 Chr.X: 103629966-103629966 on Build GRCh38
rs201420287 Chr.X: 103630033-103630033 on Build GRCh38
rs1052148 Chr.X: 103630204-103630204 on Build GRCh38
rs759474630 Chr.X: 103630019-103630019 on Build GRCh38
rs774757358 Chr.X: 103630131-103630131 on Build GRCh38
rs1052145 Chr.X: 103630116-103630116 on Build GRCh38
rs748361525 Chr.X: 103630133-103630133 on Build GRCh38
rs866226033 Chr.X: 103629986-103629986 on Build GRCh38
rs760687124 Chr.X: 103630275-103630275 on Build GRCh38
rs755313044 Chr.X: 103629988-103629988 on Build GRCh38
rs147481649 Chr.X: 103629913-103629913 on Build GRCh38
rs757044352 Chr.X: 103630019-103630033 on Build GRCh38
rs747562930 Chr.X: 103629843-103629843 on Build GRCh38
rs1052149 Chr.X: 103630218-103630218 on Build GRCh38
rs199724057 Chr.X: 103630030-103630030 on Build GRCh38
rs1051299896 Chr.X: 103629894-103629894 on Build GRCh38
rs760771251 Chr.X: 103630251-103630251 on Build GRCh38
rs762018669 Chr.X: 103629923-103629923 on Build GRCh38
rs953537154 Chr.X: 103630189-103630189 on Build GRCh38

View Full Product Details