Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105830994-105831543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRK
Entrez Gene ID
203447
Gene Name
Nik related kinase
Gene Aliases
NESK
UniGene
Hs.209527
Gene Chromosome Location
Chr.X: 105821944-105958610 on Build GRCh38
Transcripts
NM_198465.3 XM_006724632.3 XM_006724633.3 XM_006724634.3 XM_011530887.2

SNP Detail

SNP ID Chromosome Location
rs780829493 Chr.X: 105831078-105831078 on Build GRCh38
rs369465536 Chr.X: 105831153-105831153 on Build GRCh38
rs752967007 Chr.X: 105831048-105831048 on Build GRCh38
rs745612822 Chr.X: 105831269-105831269 on Build GRCh38
rs561918633 Chr.X: 105831409-105831409 on Build GRCh38
rs1027157916 Chr.X: 105831421-105831421 on Build GRCh38
rs762249160 Chr.X: 105831242-105831245 on Build GRCh38
rs755758256 Chr.X: 105831279-105831279 on Build GRCh38
rs978599252 Chr.X: 105831367-105831367 on Build GRCh38
rs982876802 Chr.X: 105831506-105831506 on Build GRCh38
rs773375649 Chr.X: 105831404-105831404 on Build GRCh38
rs12011926 Chr.X: 105831303-105831303 on Build GRCh38
rs769967912 Chr.X: 105831391-105831391 on Build GRCh38
rs1039562099 Chr.X: 105831516-105831516 on Build GRCh38
rs759705929 Chr.X: 105831035-105831035 on Build GRCh38
rs942613555 Chr.X: 105831504-105831504 on Build GRCh38
rs1005021680 Chr.X: 105831185-105831185 on Build GRCh38
rs778458581 Chr.X: 105831165-105831165 on Build GRCh38
rs780044195 Chr.X: 105831137-105831137 on Build GRCh38
rs1013478790 Chr.X: 105831281-105831281 on Build GRCh38
rs771780630 Chr.X: 105831288-105831288 on Build GRCh38
rs750254865 Chr.X: 105831101-105831101 on Build GRCh38
rs1016869758 Chr.X: 105831212-105831212 on Build GRCh38
rs755966572 Chr.X: 105831104-105831104 on Build GRCh38
rs993256844 Chr.X: 105831307-105831307 on Build GRCh38
rs913288760 Chr.X: 105831317-105831317 on Build GRCh38
rs760567404 Chr.X: 105831357-105831358 on Build GRCh38
rs779863707 Chr.X: 105831283-105831283 on Build GRCh38
rs765473059 Chr.X: 105831037-105831037 on Build GRCh38
rs749030184 Chr.X: 105831033-105831037 on Build GRCh38
rs952456828 Chr.X: 105831436-105831436 on Build GRCh38
rs1001279523 Chr.X: 105831107-105831107 on Build GRCh38
rs1001823413 Chr.X: 105831272-105831272 on Build GRCh38
rs778572720 Chr.X: 105831238-105831238 on Build GRCh38
rs374868220 Chr.X: 105831030-105831030 on Build GRCh38
rs747765338 Chr.X: 105831166-105831166 on Build GRCh38
rs748738496 Chr.X: 105831144-105831144 on Build GRCh38
rs967896753 Chr.X: 105831334-105831334 on Build GRCh38
rs758734972 Chr.X: 105831075-105831075 on Build GRCh38
rs969208202 Chr.X: 105831339-105831339 on Build GRCh38

View Full Product Details