Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185118126-185118642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT1L
Entrez Gene ID
81627
Gene Name
tRNA methyltransferase 1 like
Gene Aliases
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3
UniGene
Hs.107149
Gene Chromosome Location
Chr.1: 185118085-185157098 on Build GRCh38
Transcripts
NM_001202423.1 NM_030934.4 XM_011510028.2

SNP Detail

SNP ID Chromosome Location
rs543464557 Chr.1: 185118457-185118457 on Build GRCh38
rs748146873 Chr.1: 185118520-185118520 on Build GRCh38
rs1006930770 Chr.1: 185118336-185118336 on Build GRCh38
rs577882469 Chr.1: 185118194-185118194 on Build GRCh38
rs942954249 Chr.1: 185118587-185118587 on Build GRCh38
rs12030554 Chr.1: 185118502-185118502 on Build GRCh38
rs933550066 Chr.1: 185118375-185118375 on Build GRCh38
rs988380011 Chr.1: 185118350-185118350 on Build GRCh38
rs966558524 Chr.1: 185118464-185118464 on Build GRCh38
rs575141263 Chr.1: 185118283-185118283 on Build GRCh38
rs917583834 Chr.1: 185118538-185118538 on Build GRCh38
rs972477058 Chr.1: 185118548-185118548 on Build GRCh38
rs780084506 Chr.1: 185118167-185118167 on Build GRCh38
rs1020776279 Chr.1: 185118460-185118460 on Build GRCh38
rs1037129515 Chr.1: 185118193-185118193 on Build GRCh38
rs911437335 Chr.1: 185118585-185118585 on Build GRCh38
rs754203905 Chr.1: 185118229-185118230 on Build GRCh38
rs895723868 Chr.1: 185118541-185118541 on Build GRCh38
rs1032295526 Chr.1: 185118468-185118468 on Build GRCh38
rs545315956 Chr.1: 185118214-185118214 on Build GRCh38
rs766749667 Chr.1: 185118394-185118394 on Build GRCh38
rs931545427 Chr.1: 185118276-185118276 on Build GRCh38
rs553200597 Chr.1: 185118166-185118166 on Build GRCh38
rs148629957 Chr.1: 185118368-185118371 on Build GRCh38
rs1048624832 Chr.1: 185118584-185118584 on Build GRCh38
rs772131091 Chr.1: 185118562-185118562 on Build GRCh38
rs901222691 Chr.1: 185118376-185118376 on Build GRCh38
rs889774234 Chr.1: 185118289-185118289 on Build GRCh38
rs1056942715 Chr.1: 185118540-185118541 on Build GRCh38
rs1000772306 Chr.1: 185118467-185118467 on Build GRCh38
rs918741190 Chr.1: 185118560-185118560 on Build GRCh38
rs917717020 Chr.1: 185118177-185118177 on Build GRCh38
rs753958857 Chr.1: 185118284-185118284 on Build GRCh38
rs956850180 Chr.1: 185118347-185118347 on Build GRCh38
rs897692044 Chr.1: 185118471-185118471 on Build GRCh38
rs949162573 Chr.1: 185118183-185118183 on Build GRCh38
rs1013014592 Chr.1: 185118267-185118267 on Build GRCh38
rs754974958 Chr.1: 185118461-185118461 on Build GRCh38
rs774072188 Chr.1: 185118292-185118293 on Build GRCh38
rs867505256 Chr.1: 185118280-185118280 on Build GRCh38
rs1010749788 Chr.1: 185118434-185118434 on Build GRCh38
rs970478733 Chr.1: 185118541-185118541 on Build GRCh38
rs1020036182 Chr.1: 185118358-185118358 on Build GRCh38
rs563127739 Chr.1: 185118222-185118222 on Build GRCh38

View Full Product Details