Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108733530-108734042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS4
Entrez Gene ID
8471
Gene Name
insulin receptor substrate 4
Gene Aliases
IRS-4, PY160
UniGene
Hs.407141
Gene Chromosome Location
Chr.X: 108719949-108736584 on Build GRCh38
Transcripts
NM_003604.2 XM_005262220.2 XM_006724713.3 XM_011531061.1

SNP Detail

SNP ID Chromosome Location
rs752041983 Chr.X: 108733783-108733783 on Build GRCh38
rs772009977 Chr.X: 108733669-108733669 on Build GRCh38
rs201542593 Chr.X: 108733917-108733917 on Build GRCh38
rs775332530 Chr.X: 108733633-108733633 on Build GRCh38
rs766690582 Chr.X: 108733993-108733993 on Build GRCh38
rs948680986 Chr.X: 108733638-108733638 on Build GRCh38
rs774164887 Chr.X: 108733846-108733846 on Build GRCh38
rs745412094 Chr.X: 108733898-108733898 on Build GRCh38
rs763248496 Chr.X: 108733736-108733736 on Build GRCh38
rs764037038 Chr.X: 108733822-108733822 on Build GRCh38
rs142497632 Chr.X: 108733852-108733852 on Build GRCh38
rs780457158 Chr.X: 108733829-108733829 on Build GRCh38
rs922587529 Chr.X: 108733801-108733801 on Build GRCh38
rs761539060 Chr.X: 108733847-108733847 on Build GRCh38
rs902955518 Chr.X: 108733597-108733597 on Build GRCh38
rs1013575034 Chr.X: 108733649-108733649 on Build GRCh38
rs756340378 Chr.X: 108733827-108733827 on Build GRCh38
rs750542288 Chr.X: 108733815-108733815 on Build GRCh38
rs371393976 Chr.X: 108733873-108733873 on Build GRCh38
rs778790945 Chr.X: 108733631-108733631 on Build GRCh38
rs746155707 Chr.X: 108733614-108733614 on Build GRCh38
rs780624311 Chr.X: 108733890-108733890 on Build GRCh38
rs765657601 Chr.X: 108733574-108733574 on Build GRCh38
rs768964193 Chr.X: 108733941-108733941 on Build GRCh38
rs976747524 Chr.X: 108733791-108733791 on Build GRCh38
rs111868172 Chr.X: 108733651-108733651 on Build GRCh38
rs777734959 Chr.X: 108733858-108733858 on Build GRCh38
rs932708268 Chr.X: 108733899-108733899 on Build GRCh38
rs886181821 Chr.X: 108733835-108733835 on Build GRCh38
rs760481348 Chr.X: 108733605-108733605 on Build GRCh38
rs1801165 Chr.X: 108733697-108733697 on Build GRCh38
rs181517892 Chr.X: 108733706-108733706 on Build GRCh38
rs375073273 Chr.X: 108733648-108733648 on Build GRCh38
rs753191496 Chr.X: 108733582-108733582 on Build GRCh38
rs908598176 Chr.X: 108733756-108733756 on Build GRCh38
rs750493468 Chr.X: 108733969-108733969 on Build GRCh38
rs112671783 Chr.X: 108733712-108733712 on Build GRCh38
rs746987284 Chr.X: 108733869-108733869 on Build GRCh38
rs947195350 Chr.X: 108733995-108733995 on Build GRCh38
rs376318453 Chr.X: 108733954-108733954 on Build GRCh38
rs370554029 Chr.X: 108733647-108733647 on Build GRCh38
rs1801164 Chr.X: 108733710-108733710 on Build GRCh38
rs767548987 Chr.X: 108733807-108733807 on Build GRCh38
rs746510289 Chr.X: 108733698-108733698 on Build GRCh38
rs186496726 Chr.X: 108733945-108733945 on Build GRCh38
rs769439231 Chr.X: 108733715-108733715 on Build GRCh38
rs762165470 Chr.X: 108733951-108733951 on Build GRCh38
rs900143792 Chr.X: 108733953-108733953 on Build GRCh38
rs754804637 Chr.X: 108733615-108733615 on Build GRCh38
rs144557303 Chr.X: 108733585-108733585 on Build GRCh38
rs1030555460 Chr.X: 108734008-108734008 on Build GRCh38
rs145401626 Chr.X: 108733612-108733612 on Build GRCh38
rs758209012 Chr.X: 108734015-108734015 on Build GRCh38
rs773048763 Chr.X: 108733673-108733673 on Build GRCh38
rs775170903 Chr.X: 108733935-108733935 on Build GRCh38
rs1015912814 Chr.X: 108733906-108733906 on Build GRCh38
rs761944161 Chr.X: 108733800-108733800 on Build GRCh38
rs376750039 Chr.X: 108733841-108733841 on Build GRCh38
rs775790597 Chr.X: 108733719-108733719 on Build GRCh38
rs751607647 Chr.X: 108733592-108733592 on Build GRCh38
rs760863193 Chr.X: 108733985-108733985 on Build GRCh38
rs764640800 Chr.X: 108733773-108733773 on Build GRCh38
rs367686563 Chr.X: 108733997-108733997 on Build GRCh38

View Full Product Details