Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109535759-109536301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXT2
Entrez Gene ID
55916
Gene Name
nuclear transport factor 2 like export factor 2
Gene Aliases
P15-2
UniGene
Hs.25010
Gene Chromosome Location
Chr.X: 109535781-109544698 on Build GRCh38
Transcripts
NM_018698.4

SNP Detail

SNP ID Chromosome Location
rs765256389 Chr.X: 109535873-109535873 on Build GRCh38
rs752816354 Chr.X: 109536176-109536176 on Build GRCh38
rs73639132 Chr.X: 109535819-109535819 on Build GRCh38
rs761973139 Chr.X: 109535997-109535997 on Build GRCh38
rs755870972 Chr.X: 109535903-109535903 on Build GRCh38
rs761340733 Chr.X: 109536198-109536199 on Build GRCh38
rs1015540626 Chr.X: 109535961-109535961 on Build GRCh38
rs764432699 Chr.X: 109535880-109535880 on Build GRCh38
rs1022717878 Chr.X: 109535834-109535834 on Build GRCh38
rs34620663 Chr.X: 109536027-109536027 on Build GRCh38
rs887153212 Chr.X: 109536072-109536072 on Build GRCh38
rs752811443 Chr.X: 109535876-109535876 on Build GRCh38
rs903338221 Chr.X: 109536233-109536233 on Build GRCh38
rs959945720 Chr.X: 109535816-109535816 on Build GRCh38
rs1043188787 Chr.X: 109536185-109536185 on Build GRCh38
rs751746335 Chr.X: 109535994-109535994 on Build GRCh38
rs182432266 Chr.X: 109536056-109536056 on Build GRCh38
rs921541246 Chr.X: 109535823-109535823 on Build GRCh38
rs142540133 Chr.X: 109535925-109535925 on Build GRCh38
rs769216040 Chr.X: 109535933-109535935 on Build GRCh38
rs929613442 Chr.X: 109535867-109535867 on Build GRCh38
rs779725846 Chr.X: 109535914-109535914 on Build GRCh38
rs1057496468 Chr.X: 109536271-109536271 on Build GRCh38
rs764577927 Chr.X: 109535973-109535973 on Build GRCh38
rs187892518 Chr.X: 109536079-109536079 on Build GRCh38
rs963264042 Chr.X: 109535777-109535777 on Build GRCh38
rs934942342 Chr.X: 109536256-109536256 on Build GRCh38
rs778545985 Chr.X: 109535933-109535933 on Build GRCh38
rs201184200 Chr.X: 109535964-109535964 on Build GRCh38
rs376238335 Chr.X: 109536028-109536028 on Build GRCh38
rs750177910 Chr.X: 109535896-109535896 on Build GRCh38
rs986273363 Chr.X: 109536127-109536127 on Build GRCh38
rs998951032 Chr.X: 109535795-109535795 on Build GRCh38
rs753074814 Chr.X: 109535839-109535839 on Build GRCh38
rs763086059 Chr.X: 109535991-109535991 on Build GRCh38
rs775531097 Chr.X: 109535981-109535981 on Build GRCh38
rs201588928 Chr.X: 109536198-109536200 on Build GRCh38
rs781354582 Chr.X: 109536128-109536128 on Build GRCh38
rs955548871 Chr.X: 109535894-109535894 on Build GRCh38
rs1035380820 Chr.X: 109535803-109535803 on Build GRCh38
rs760054773 Chr.X: 109535851-109535851 on Build GRCh38
rs372800448 Chr.X: 109536009-109536009 on Build GRCh38
rs947587339 Chr.X: 109536173-109536173 on Build GRCh38
rs113731871 Chr.X: 109536181-109536181 on Build GRCh38
rs903433191 Chr.X: 109535794-109535794 on Build GRCh38
rs755006162 Chr.X: 109535932-109535932 on Build GRCh38
rs746658531 Chr.X: 109535957-109535957 on Build GRCh38
rs1017318886 Chr.X: 109536081-109536081 on Build GRCh38
rs777141261 Chr.X: 109535842-109535842 on Build GRCh38
rs771849323 Chr.X: 109535941-109535941 on Build GRCh38
rs763291868 Chr.X: 109535934-109535934 on Build GRCh38
rs749680098 Chr.X: 109535864-109535865 on Build GRCh38
rs36012663 Chr.X: 109535976-109535976 on Build GRCh38
rs760001620 Chr.X: 109535967-109535967 on Build GRCh38
rs771282304 Chr.X: 109535962-109535962 on Build GRCh38
rs1054009020 Chr.X: 109535966-109535966 on Build GRCh38
rs764083224 Chr.X: 109535993-109535993 on Build GRCh38
rs764838013 Chr.X: 109536252-109536252 on Build GRCh38
rs369334711 Chr.X: 109536029-109536029 on Build GRCh38
rs747719439 Chr.X: 109535935-109535935 on Build GRCh38
rs775330593 Chr.X: 109536054-109536054 on Build GRCh38
rs777419792 Chr.X: 109535947-109535947 on Build GRCh38
rs763135817 Chr.X: 109535877-109535877 on Build GRCh38
rs200616803 Chr.X: 109536195-109536196 on Build GRCh38
rs757327321 Chr.X: 109536040-109536040 on Build GRCh38

View Full Product Details