Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 116442591-116443094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A14
Entrez Gene ID
11254
Gene Name
solute carrier family 6 member 14
Gene Aliases
BMIQ11
UniGene
Hs.522109
Gene Chromosome Location
Chr.X: 116436579-116461458 on Build GRCh38
Transcripts
NM_007231.4

SNP Detail

SNP ID Chromosome Location
rs782082858 Chr.X: 116442674-116442674 on Build GRCh38
rs782600646 Chr.X: 116442769-116442769 on Build GRCh38
rs782611656 Chr.X: 116442898-116442898 on Build GRCh38
rs146435019 Chr.X: 116442844-116442844 on Build GRCh38
rs201550225 Chr.X: 116442814-116442814 on Build GRCh38
rs782574852 Chr.X: 116442810-116442810 on Build GRCh38
rs143894279 Chr.X: 116442796-116442796 on Build GRCh38
rs782204139 Chr.X: 116442813-116442813 on Build GRCh38
rs12720079 Chr.X: 116442644-116442644 on Build GRCh38
rs781804835 Chr.X: 116442674-116442675 on Build GRCh38
rs146252872 Chr.X: 116442995-116442995 on Build GRCh38
rs1053247587 Chr.X: 116442978-116442978 on Build GRCh38
rs782506887 Chr.X: 116442786-116442786 on Build GRCh38
rs782739692 Chr.X: 116442703-116442703 on Build GRCh38
rs781860708 Chr.X: 116442888-116442889 on Build GRCh38
rs782632580 Chr.X: 116443043-116443043 on Build GRCh38
rs782694671 Chr.X: 116442884-116442884 on Build GRCh38
rs781858467 Chr.X: 116442712-116442712 on Build GRCh38
rs984679978 Chr.X: 116442743-116442743 on Build GRCh38
rs782153288 Chr.X: 116442675-116442675 on Build GRCh38
rs782230849 Chr.X: 116442900-116442900 on Build GRCh38
rs782018592 Chr.X: 116442856-116442856 on Build GRCh38
rs370750794 Chr.X: 116442749-116442749 on Build GRCh38
rs782458807 Chr.X: 116442889-116442889 on Build GRCh38
rs782524050 Chr.X: 116442879-116442879 on Build GRCh38
rs782813130 Chr.X: 116442887-116442887 on Build GRCh38
rs373327658 Chr.X: 116442646-116442646 on Build GRCh38
rs375653551 Chr.X: 116442775-116442775 on Build GRCh38
rs781866682 Chr.X: 116442873-116442873 on Build GRCh38
rs782131785 Chr.X: 116442714-116442714 on Build GRCh38
rs782711760 Chr.X: 116442862-116442862 on Build GRCh38
rs782290997 Chr.X: 116443051-116443051 on Build GRCh38
rs782432779 Chr.X: 116442793-116442793 on Build GRCh38
rs1043275944 Chr.X: 116443060-116443060 on Build GRCh38
rs1011802643 Chr.X: 116443039-116443039 on Build GRCh38
rs943348262 Chr.X: 116442792-116442792 on Build GRCh38
rs782395808 Chr.X: 116442855-116442855 on Build GRCh38
rs781822786 Chr.X: 116442863-116442863 on Build GRCh38
rs782810098 Chr.X: 116442727-116442727 on Build GRCh38
rs377535410 Chr.X: 116442671-116442671 on Build GRCh38
rs782022318 Chr.X: 116442639-116442639 on Build GRCh38
rs782047970 Chr.X: 116442857-116442857 on Build GRCh38
rs781788758 Chr.X: 116442737-116442737 on Build GRCh38
rs782092865 Chr.X: 116442869-116442869 on Build GRCh38
rs782635897 Chr.X: 116442790-116442790 on Build GRCh38
rs782747833 Chr.X: 116442870-116442870 on Build GRCh38

View Full Product Details