Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 118567943-118568468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
UniGene
Hs.368203
Gene Chromosome Location
Chr.X: 118495664-118686162 on Build GRCh38
Transcripts
NM_144658.3 XM_005262368.3 XM_005262369.4 XM_011531276.2

SNP Detail

SNP ID Chromosome Location
rs934522862 Chr.X: 118568345-118568345 on Build GRCh38
rs770341895 Chr.X: 118568382-118568382 on Build GRCh38
rs1039252800 Chr.X: 118568394-118568418 on Build GRCh38
rs924319485 Chr.X: 118568308-118568308 on Build GRCh38
rs773969388 Chr.X: 118568386-118568410 on Build GRCh38
rs149921668 Chr.X: 118568132-118568132 on Build GRCh38
rs769162843 Chr.X: 118568269-118568269 on Build GRCh38
rs762858840 Chr.X: 118568149-118568149 on Build GRCh38
rs780723340 Chr.X: 118568188-118568188 on Build GRCh38
rs373058895 Chr.X: 118568200-118568200 on Build GRCh38
rs780156146 Chr.X: 118568176-118568176 on Build GRCh38
rs752692900 Chr.X: 118568147-118568147 on Build GRCh38
rs965696985 Chr.X: 118568225-118568225 on Build GRCh38
rs569113770 Chr.X: 118568400-118568425 on Build GRCh38
rs905191526 Chr.X: 118567977-118567977 on Build GRCh38
rs376684103 Chr.X: 118568067-118568067 on Build GRCh38
rs139686157 Chr.X: 118568109-118568109 on Build GRCh38
rs936474310 Chr.X: 118568009-118568009 on Build GRCh38
rs41300205 Chr.X: 118567999-118567999 on Build GRCh38
rs147372245 Chr.X: 118568099-118568099 on Build GRCh38
rs962239854 Chr.X: 118568096-118568096 on Build GRCh38
rs777251111 Chr.X: 118568271-118568271 on Build GRCh38
rs868514976 Chr.X: 118568390-118568390 on Build GRCh38
rs748855406 Chr.X: 118568033-118568034 on Build GRCh38
rs765008919 Chr.X: 118568144-118568144 on Build GRCh38
rs112643752 Chr.X: 118568330-118568330 on Build GRCh38
rs375849535 Chr.X: 118568388-118568388 on Build GRCh38
rs989585052 Chr.X: 118568068-118568068 on Build GRCh38
rs77784406 Chr.X: 118568063-118568063 on Build GRCh38
rs762445955 Chr.X: 118568295-118568295 on Build GRCh38
rs926479471 Chr.X: 118568002-118568002 on Build GRCh38
rs867109604 Chr.X: 118568431-118568431 on Build GRCh38
rs761911366 Chr.X: 118568080-118568080 on Build GRCh38
rs202060296 Chr.X: 118568180-118568180 on Build GRCh38
rs756744345 Chr.X: 118568181-118568181 on Build GRCh38
rs943673712 Chr.X: 118568316-118568319 on Build GRCh38
rs397839351 Chr.X: 118568374-118568377 on Build GRCh38
rs111993233 Chr.X: 118568186-118568186 on Build GRCh38

View Full Product Details