Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119871263-119871769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF113A
Entrez Gene ID
7737
Gene Name
ring finger protein 113A
Gene Aliases
Cwc24, RNF113, TTD5, ZNF183
UniGene
Hs.458365
Gene Chromosome Location
Chr.X: 119870532-119871828 on Build GRCh38
Transcripts
NM_006978.2

SNP Detail

SNP ID Chromosome Location
rs963921798 Chr.X: 119871691-119871691 on Build GRCh38
rs908412913 Chr.X: 119871447-119871447 on Build GRCh38
rs772741803 Chr.X: 119871735-119871735 on Build GRCh38
rs11542909 Chr.X: 119871400-119871400 on Build GRCh38
rs781301280 Chr.X: 119871298-119871298 on Build GRCh38
rs774848716 Chr.X: 119871331-119871331 on Build GRCh38
rs369106726 Chr.X: 119871538-119871538 on Build GRCh38
rs1050659898 Chr.X: 119871720-119871720 on Build GRCh38
rs768195723 Chr.X: 119871334-119871334 on Build GRCh38
rs708463 Chr.X: 119871742-119871742 on Build GRCh38
rs769849012 Chr.X: 119871525-119871525 on Build GRCh38
rs34063391 Chr.X: 119871299-119871299 on Build GRCh38
rs954810120 Chr.X: 119871496-119871496 on Build GRCh38
rs768589089 Chr.X: 119871727-119871727 on Build GRCh38
rs773994808 Chr.X: 119871335-119871335 on Build GRCh38
rs889361201 Chr.X: 119871723-119871723 on Build GRCh38
rs779018953 Chr.X: 119871417-119871417 on Build GRCh38
rs375042930 Chr.X: 119871285-119871285 on Build GRCh38
rs991389900 Chr.X: 119871322-119871322 on Build GRCh38
rs762732386 Chr.X: 119871333-119871333 on Build GRCh38
rs777139315 Chr.X: 119871648-119871648 on Build GRCh38
rs768160360 Chr.X: 119871586-119871586 on Build GRCh38
rs757151061 Chr.X: 119871466-119871466 on Build GRCh38
rs773903348 Chr.X: 119871606-119871606 on Build GRCh38
rs762103846 Chr.X: 119871337-119871337 on Build GRCh38
rs766879054 Chr.X: 119871642-119871642 on Build GRCh38
rs776401851 Chr.X: 119871370-119871372 on Build GRCh38
rs878909171 Chr.X: 119871698-119871698 on Build GRCh38
rs745858628 Chr.X: 119871515-119871515 on Build GRCh38
rs752917059 Chr.X: 119871459-119871459 on Build GRCh38
rs779972737 Chr.X: 119871531-119871531 on Build GRCh38
rs1800823 Chr.X: 119871624-119871624 on Build GRCh38
rs147930134 Chr.X: 119871404-119871404 on Build GRCh38
rs747763370 Chr.X: 119871614-119871614 on Build GRCh38
rs759557740 Chr.X: 119871651-119871651 on Build GRCh38
rs750964180 Chr.X: 119871377-119871377 on Build GRCh38
rs939756752 Chr.X: 119871473-119871473 on Build GRCh38
rs199657887 Chr.X: 119871350-119871350 on Build GRCh38
rs981234204 Chr.X: 119871418-119871418 on Build GRCh38
rs932316608 Chr.X: 119871644-119871644 on Build GRCh38
rs755021073 Chr.X: 119871410-119871410 on Build GRCh38
rs142377248 Chr.X: 119871329-119871329 on Build GRCh38
rs781103087 Chr.X: 119871494-119871494 on Build GRCh38
rs386352357 Chr.X: 119871367-119871367 on Build GRCh38
rs778095662 Chr.X: 119871378-119871378 on Build GRCh38
rs761240377 Chr.X: 119871641-119871641 on Build GRCh38
rs983047256 Chr.X: 119871736-119871736 on Build GRCh38
rs374130836 Chr.X: 119871291-119871291 on Build GRCh38
rs957615955 Chr.X: 119871319-119871319 on Build GRCh38
rs773556907 Chr.X: 119871639-119871639 on Build GRCh38
rs757385436 Chr.X: 119871730-119871730 on Build GRCh38
rs370353872 Chr.X: 119871389-119871389 on Build GRCh38
rs748759529 Chr.X: 119871575-119871575 on Build GRCh38
rs746787077 Chr.X: 119871289-119871289 on Build GRCh38

View Full Product Details