Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120252590-120253090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255A
Entrez Gene ID
55026
Gene Name
transmembrane protein 255A
Gene Aliases
FAM70A
UniGene
Hs.437563
Gene Chromosome Location
Chr.X: 120250984-120311556 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

Gene Symbol
ZBTB33
Entrez Gene ID
10009
Gene Name
zinc finger and BTB domain containing 33
Gene Aliases
ZNF-kaiso, ZNF348
UniGene
Hs.143604
Gene Chromosome Location
Chr.X: 120250752-120258396 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

SNP Detail

SNP ID Chromosome Location
rs921115197 Chr.X: 120252981-120252981 on Build GRCh38
rs782277866 Chr.X: 120252826-120252826 on Build GRCh38
rs782738106 Chr.X: 120252841-120252841 on Build GRCh38
rs781950319 Chr.X: 120252851-120252851 on Build GRCh38
rs1000415724 Chr.X: 120252665-120252665 on Build GRCh38
rs975346584 Chr.X: 120252959-120252959 on Build GRCh38
rs1017357957 Chr.X: 120252908-120252908 on Build GRCh38
rs945114086 Chr.X: 120253055-120253055 on Build GRCh38
rs913609532 Chr.X: 120253038-120253038 on Build GRCh38
rs782696939 Chr.X: 120252773-120252773 on Build GRCh38
rs782399661 Chr.X: 120252871-120252871 on Build GRCh38
rs955161011 Chr.X: 120252983-120252983 on Build GRCh38
rs782062265 Chr.X: 120252857-120252857 on Build GRCh38
rs986960931 Chr.X: 120252995-120252995 on Build GRCh38
rs904737738 Chr.X: 120252659-120252659 on Build GRCh38
rs962592220 Chr.X: 120252935-120252935 on Build GRCh38
rs1055864206 Chr.X: 120252761-120252761 on Build GRCh38

View Full Product Details