Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129794665-129795169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SASH3
Entrez Gene ID
54440
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
753P9, CXorf9, HACS2, SH3D6C, SLY
UniGene
Hs.61469
Gene Chromosome Location
Chr.X: 129779916-129795201 on Build GRCh38
Transcripts
NM_018990.3 XM_006724763.1

SNP Detail

SNP ID Chromosome Location
rs900906407 Chr.X: 129794803-129794803 on Build GRCh38
rs917280614 Chr.X: 129794925-129794925 on Build GRCh38
rs906061089 Chr.X: 129794721-129794721 on Build GRCh38
rs1042016032 Chr.X: 129795000-129795000 on Build GRCh38
rs1020589338 Chr.X: 129794800-129794800 on Build GRCh38
rs775615936 Chr.X: 129794954-129794954 on Build GRCh38
rs781690385 Chr.X: 129795055-129795055 on Build GRCh38
rs752341990 Chr.X: 129794878-129794878 on Build GRCh38
rs927441299 Chr.X: 129794978-129794978 on Build GRCh38
rs984364036 Chr.X: 129794877-129794877 on Build GRCh38
rs751633728 Chr.X: 129794692-129794692 on Build GRCh38
rs900875409 Chr.X: 129795038-129795038 on Build GRCh38
rs761834344 Chr.X: 129794699-129794699 on Build GRCh38
rs972676507 Chr.X: 129795140-129795140 on Build GRCh38
rs16320 Chr.X: 129794691-129794691 on Build GRCh38
rs568868768 Chr.X: 129794700-129794700 on Build GRCh38
rs920829081 Chr.X: 129795134-129795134 on Build GRCh38
rs980218848 Chr.X: 129794962-129794962 on Build GRCh38
rs746394944 Chr.X: 129795056-129795056 on Build GRCh38
rs1044568288 Chr.X: 129794708-129794708 on Build GRCh38
rs137871504 Chr.X: 129794707-129794707 on Build GRCh38
rs972737651 Chr.X: 129794884-129794884 on Build GRCh38
rs1048194851 Chr.X: 129794887-129794887 on Build GRCh38
rs889541302 Chr.X: 129795076-129795076 on Build GRCh38
rs181999253 Chr.X: 129795102-129795102 on Build GRCh38
rs940829028 Chr.X: 129795093-129795093 on Build GRCh38
rs761920716 Chr.X: 129795111-129795111 on Build GRCh38
rs912147602 Chr.X: 129794849-129794849 on Build GRCh38
rs961604925 Chr.X: 129794879-129794879 on Build GRCh38
rs190763855 Chr.X: 129794981-129794981 on Build GRCh38
rs989561470 Chr.X: 129794712-129794712 on Build GRCh38
rs961301070 Chr.X: 129795137-129795137 on Build GRCh38
rs953461323 Chr.X: 129795048-129795048 on Build GRCh38
rs1028099353 Chr.X: 129794837-129794837 on Build GRCh38
rs770398745 Chr.X: 129795007-129795007 on Build GRCh38
rs940882689 Chr.X: 129794854-129794854 on Build GRCh38
rs1051926143 Chr.X: 129794762-129794762 on Build GRCh38
rs923904642 Chr.X: 129794693-129794693 on Build GRCh38
rs774773932 Chr.X: 129794839-129794839 on Build GRCh38
rs771459111 Chr.X: 129794825-129794825 on Build GRCh38
rs764362730 Chr.X: 129794982-129794982 on Build GRCh38
rs946309309 Chr.X: 129794992-129794992 on Build GRCh38
rs772468125 Chr.X: 129794885-129794885 on Build GRCh38
rs1027989820 Chr.X: 129795070-129795070 on Build GRCh38
rs1054751126 Chr.X: 129794782-129794782 on Build GRCh38
rs1024786713 Chr.X: 129794944-129794944 on Build GRCh38
rs759829972 Chr.X: 129794868-129794868 on Build GRCh38
rs761519240 Chr.X: 129794776-129794777 on Build GRCh38
rs892044158 Chr.X: 129794783-129794783 on Build GRCh38
rs371826558 Chr.X: 129795077-129795077 on Build GRCh38
rs894881622 Chr.X: 129794931-129794931 on Build GRCh38
rs922039471 Chr.X: 129794731-129794731 on Build GRCh38
rs892017393 Chr.X: 129794986-129794986 on Build GRCh38
rs1012506009 Chr.X: 129794934-129794938 on Build GRCh38
rs548322002 Chr.X: 129794743-129794743 on Build GRCh38
rs1000845886 Chr.X: 129794737-129794737 on Build GRCh38
rs184303879 Chr.X: 129794795-129794795 on Build GRCh38
rs41300283 Chr.X: 129795069-129795069 on Build GRCh38
rs932166400 Chr.X: 129794748-129794748 on Build GRCh38
rs912029019 Chr.X: 129795084-129795084 on Build GRCh38
rs778858418 Chr.X: 129794876-129794877 on Build GRCh38
rs950102212 Chr.X: 129794945-129794945 on Build GRCh38
rs766434367 Chr.X: 129795141-129795141 on Build GRCh38

View Full Product Details