Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132626277-132626807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST2
Entrez Gene ID
90161
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Gene Aliases
-
UniGene
Hs.385956
Gene Chromosome Location
Chr.X: 132626010-132961395 on Build GRCh38
Transcripts
NM_001077188.1 NM_147175.3 XM_005262490.3 XM_005262491.2 XM_011531406.1 XM_017029944.1 XM_017029945.1 XM_017029946.1

SNP Detail

SNP ID Chromosome Location
rs748518573 Chr.X: 132626767-132626767 on Build GRCh38
rs948915863 Chr.X: 132626338-132626338 on Build GRCh38
rs998455717 Chr.X: 132626413-132626413 on Build GRCh38
rs961000995 Chr.X: 132626640-132626640 on Build GRCh38
rs761625184 Chr.X: 132626444-132626444 on Build GRCh38
rs992218189 Chr.X: 132626674-132626674 on Build GRCh38
rs776973660 Chr.X: 132626587-132626587 on Build GRCh38
rs557836111 Chr.X: 132626500-132626500 on Build GRCh38
rs768613903 Chr.X: 132626328-132626328 on Build GRCh38
rs774035565 Chr.X: 132626367-132626370 on Build GRCh38
rs749275871 Chr.X: 132626306-132626306 on Build GRCh38
rs964882701 Chr.X: 132626702-132626702 on Build GRCh38
rs1009219448 Chr.X: 132626662-132626662 on Build GRCh38
rs1042777139 Chr.X: 132626378-132626378 on Build GRCh38
rs765344165 Chr.X: 132626781-132626781 on Build GRCh38
rs894633568 Chr.X: 132626308-132626308 on Build GRCh38
rs1033723035 Chr.X: 132626616-132626616 on Build GRCh38
rs755187448 Chr.X: 132626521-132626521 on Build GRCh38
rs886234639 Chr.X: 132626388-132626388 on Build GRCh38
rs243462 Chr.X: 132626751-132626751 on Build GRCh38
rs904295819 Chr.X: 132626401-132626401 on Build GRCh38
rs1017558851 Chr.X: 132626696-132626696 on Build GRCh38
rs771059119 Chr.X: 132626469-132626469 on Build GRCh38
rs751609400 Chr.X: 132626301-132626301 on Build GRCh38

View Full Product Details