Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186312964-186313465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Gene Symbol
TPR
Entrez Gene ID
7175
Gene Name
translocated promoter region, nuclear basket protein
Gene Aliases
-
UniGene
Hs.279640
Gene Chromosome Location
Chr.1: 186311652-186375325 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

SNP Detail

SNP ID Chromosome Location
rs544807583 Chr.1: 186313074-186313074 on Build GRCh38
rs922257149 Chr.1: 186313119-186313119 on Build GRCh38
rs538996915 Chr.1: 186313013-186313013 on Build GRCh38
rs551031864 Chr.1: 186313209-186313209 on Build GRCh38
rs1050813633 Chr.1: 186313101-186313101 on Build GRCh38
rs1055625236 Chr.1: 186313337-186313337 on Build GRCh38
rs1006496497 Chr.1: 186313117-186313117 on Build GRCh38
rs1005683005 Chr.1: 186313025-186313025 on Build GRCh38
rs762320192 Chr.1: 186313190-186313190 on Build GRCh38
rs966986658 Chr.1: 186313174-186313174 on Build GRCh38
rs374971550 Chr.1: 186313290-186313290 on Build GRCh38
rs977597377 Chr.1: 186313000-186313000 on Build GRCh38
rs539138917 Chr.1: 186313133-186313133 on Build GRCh38
rs925854372 Chr.1: 186313314-186313314 on Build GRCh38
rs542610053 Chr.1: 186313090-186313090 on Build GRCh38
rs373050687 Chr.1: 186313166-186313166 on Build GRCh38
rs557224189 Chr.1: 186313019-186313019 on Build GRCh38
rs958728335 Chr.1: 186313273-186313273 on Build GRCh38
rs994410258 Chr.1: 186312999-186312999 on Build GRCh38
rs901526136 Chr.1: 186313048-186313048 on Build GRCh38
rs976493865 Chr.1: 186313110-186313110 on Build GRCh38
rs968982094 Chr.1: 186313389-186313389 on Build GRCh38
rs185519458 Chr.1: 186313419-186313419 on Build GRCh38
rs757628385 Chr.1: 186313201-186313201 on Build GRCh38
rs1025870826 Chr.1: 186313004-186313004 on Build GRCh38
rs572325418 Chr.1: 186313370-186313370 on Build GRCh38
rs1040074963 Chr.1: 186313033-186313033 on Build GRCh38
rs935924738 Chr.1: 186313332-186313332 on Build GRCh38
rs554305144 Chr.1: 186313109-186313109 on Build GRCh38
rs1022316640 Chr.1: 186313372-186313372 on Build GRCh38
rs753601576 Chr.1: 186313129-186313129 on Build GRCh38
rs1002962398 Chr.1: 186313137-186313137 on Build GRCh38

View Full Product Details