Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196235751-196236255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
UniGene
Hs.657046
Gene Chromosome Location
Chr.1: 196225779-196608905 on Build GRCh38
Transcripts
NM_001287819.1 NM_001287820.1 NM_198503.3 XM_006711294.2 XM_006711295.2 XM_011509482.2 XM_017001181.1 XM_017001182.1 XM_017001183.1 XM_017001184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373259 c.3226C>A p.H1076N Chr.1: 196236056-196236056 on Build GRCh38
6433733 c.3224A>G p.D1075G Chr.1: 196236058-196236058 on Build GRCh38
7373260 c.3028C>A p.H1010N Chr.1: 196236056-196236056 on Build GRCh38
4026447 c.3296+2T>C p.? Chr.1: 196235984-196235984 on Build GRCh38
6527965 c.3040C>G p.L1014V Chr.1: 196236044-196236044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758203062 Chr.1: 196236088-196236088 on Build GRCh38
rs578226686 Chr.1: 196236090-196236090 on Build GRCh38
rs916591816 Chr.1: 196236182-196236182 on Build GRCh38
rs966656496 Chr.1: 196235835-196235835 on Build GRCh38
rs1028621200 Chr.1: 196236040-196236040 on Build GRCh38
rs776072435 Chr.1: 196236117-196236117 on Build GRCh38
rs112896769 Chr.1: 196235864-196235864 on Build GRCh38
rs573616600 Chr.1: 196236010-196236010 on Build GRCh38
rs1029139025 Chr.1: 196236197-196236197 on Build GRCh38
rs74133635 Chr.1: 196235782-196235782 on Build GRCh38
rs376760816 Chr.1: 196236186-196236186 on Build GRCh38
rs751133808 Chr.1: 196236080-196236081 on Build GRCh38
rs770443462 Chr.1: 196236116-196236116 on Build GRCh38
rs777642338 Chr.1: 196236103-196236103 on Build GRCh38
rs866361922 Chr.1: 196236077-196236077 on Build GRCh38
rs764566142 Chr.1: 196236038-196236038 on Build GRCh38
rs960693295 Chr.1: 196236170-196236170 on Build GRCh38
rs543009008 Chr.1: 196236191-196236191 on Build GRCh38
rs368828604 Chr.1: 196235852-196235852 on Build GRCh38
rs750693818 Chr.1: 196236074-196236074 on Build GRCh38
rs948136681 Chr.1: 196235889-196235890 on Build GRCh38
rs771948142 Chr.1: 196235938-196235938 on Build GRCh38
rs371861098 Chr.1: 196235956-196235956 on Build GRCh38
rs756360956 Chr.1: 196236078-196236078 on Build GRCh38
rs767642359 Chr.1: 196236056-196236056 on Build GRCh38
rs765412124 Chr.1: 196235817-196235817 on Build GRCh38
rs781029843 Chr.1: 196236120-196236121 on Build GRCh38
rs192873552 Chr.1: 196235814-196235814 on Build GRCh38
rs928114065 Chr.1: 196235925-196235925 on Build GRCh38
rs775756740 Chr.1: 196236019-196236019 on Build GRCh38
rs972962900 Chr.1: 196235923-196235923 on Build GRCh38
rs967641551 Chr.1: 196236137-196236137 on Build GRCh38
rs1018891293 Chr.1: 196236156-196236156 on Build GRCh38
rs1011756223 Chr.1: 196236104-196236104 on Build GRCh38
rs895271619 Chr.1: 196235886-196235886 on Build GRCh38
rs939362677 Chr.1: 196235879-196235879 on Build GRCh38
rs375199969 Chr.1: 196235961-196235961 on Build GRCh38
rs754487175 Chr.1: 196236082-196236083 on Build GRCh38
rs1018755238 Chr.1: 196235856-196235856 on Build GRCh38
rs75327918 Chr.1: 196236155-196236155 on Build GRCh38
rs910129429 Chr.1: 196235914-196235914 on Build GRCh38
rs779810555 Chr.1: 196235962-196235966 on Build GRCh38
rs1056553538 Chr.1: 196235882-196235882 on Build GRCh38
rs951867189 Chr.1: 196236199-196236199 on Build GRCh38
rs748112594 Chr.1: 196236121-196236122 on Build GRCh38
rs539347760 Chr.1: 196235846-196235846 on Build GRCh38
rs901646028 Chr.1: 196236165-196236165 on Build GRCh38
rs1021436090 Chr.1: 196236134-196236134 on Build GRCh38
rs558124452 Chr.1: 196235912-196235912 on Build GRCh38
rs185046172 Chr.1: 196236211-196236211 on Build GRCh38
rs367996950 Chr.1: 196236045-196236045 on Build GRCh38
rs201788126 Chr.1: 196235955-196235955 on Build GRCh38
rs1043766441 Chr.1: 196236106-196236106 on Build GRCh38
rs1049629329 Chr.1: 196236218-196236218 on Build GRCh38
rs770424892 Chr.1: 196235968-196235968 on Build GRCh38
rs747981032 Chr.1: 196235867-196235867 on Build GRCh38
rs545701461 Chr.1: 196236133-196236133 on Build GRCh38
rs867616993 Chr.1: 196236068-196236068 on Build GRCh38
rs752101550 Chr.1: 196236039-196236039 on Build GRCh38
rs984495611 Chr.1: 196235910-196235910 on Build GRCh38
rs746882785 Chr.1: 196236105-196236105 on Build GRCh38
rs776270024 Chr.1: 196235976-196235976 on Build GRCh38
rs111707109 Chr.1: 196235939-196235939 on Build GRCh38
rs887954552 Chr.1: 196235918-196235918 on Build GRCh38
rs951786417 Chr.1: 196235894-196235894 on Build GRCh38
rs538439492 Chr.1: 196236032-196236032 on Build GRCh38
rs921713183 Chr.1: 196235878-196235878 on Build GRCh38
rs572494408 Chr.1: 196236160-196236160 on Build GRCh38
rs556479756 Chr.1: 196236079-196236079 on Build GRCh38
rs953100449 Chr.1: 196236049-196236049 on Build GRCh38
rs992120677 Chr.1: 196236176-196236176 on Build GRCh38
rs914400411 Chr.1: 196235803-196235803 on Build GRCh38
rs759132983 Chr.1: 196235987-196235987 on Build GRCh38
rs940244888 Chr.1: 196235916-196235916 on Build GRCh38

View Full Product Details