Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196826915-196827446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR1
Entrez Gene ID
3078
Gene Name
complement factor H related 1
Gene Aliases
CFHL, CFHL1, CFHL1P, CFHR1P, FHR1, H36-1, H36-2, HFL1, HFL2
UniGene
Hs.575869
Gene Chromosome Location
Chr.1: 196819730-196832189 on Build GRCh38
Transcripts
NM_002113.2 XM_011509457.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4544164 c.350G>A p.G117E Chr.1: 196826925-196826925 on Build GRCh38
159959 c.396G>A p.R132R Chr.1: 196826971-196826971 on Build GRCh38
1337497 c.408C>T p.T136T Chr.1: 196826983-196826983 on Build GRCh38
5785655 c.428C>G p.T143S Chr.1: 196827003-196827003 on Build GRCh38
1689488 c.401G>A p.W134* Chr.1: 196826976-196826976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912283838 Chr.1: 196827202-196827202 on Build GRCh38
rs568183209 Chr.1: 196826985-196826985 on Build GRCh38
rs775738898 Chr.1: 196827024-196827024 on Build GRCh38
rs766156594 Chr.1: 196827042-196827042 on Build GRCh38
rs573448941 Chr.1: 196827136-196827136 on Build GRCh38
rs532758928 Chr.1: 196827301-196827301 on Build GRCh38
rs112189960 Chr.1: 196827107-196827107 on Build GRCh38
rs189410604 Chr.1: 196827030-196827030 on Build GRCh38
rs556680440 Chr.1: 196827038-196827038 on Build GRCh38
rs982575306 Chr.1: 196827193-196827193 on Build GRCh38
rs432234 Chr.1: 196827231-196827231 on Build GRCh38
rs762901236 Chr.1: 196827028-196827028 on Build GRCh38
rs540591991 Chr.1: 196827163-196827163 on Build GRCh38
rs566765367 Chr.1: 196827374-196827374 on Build GRCh38
rs965129372 Chr.1: 196827230-196827230 on Build GRCh38
rs553095951 Chr.1: 196826982-196826982 on Build GRCh38
rs438781 Chr.1: 196827110-196827110 on Build GRCh38
rs865997503 Chr.1: 196827267-196827267 on Build GRCh38
rs417843 Chr.1: 196827396-196827396 on Build GRCh38
rs562287224 Chr.1: 196827209-196827209 on Build GRCh38
rs181826269 Chr.1: 196827265-196827265 on Build GRCh38
rs372102455 Chr.1: 196826950-196826950 on Build GRCh38
rs1027188097 Chr.1: 196827128-196827128 on Build GRCh38
rs776991220 Chr.1: 196827189-196827189 on Build GRCh38
rs769183299 Chr.1: 196827099-196827099 on Build GRCh38
rs747401894 Chr.1: 196826991-196826991 on Build GRCh38
rs759874591 Chr.1: 196827005-196827005 on Build GRCh38
rs544708004 Chr.1: 196827242-196827242 on Build GRCh38
rs368205158 Chr.1: 196827354-196827354 on Build GRCh38
rs747847034 Chr.1: 196826969-196826969 on Build GRCh38
rs760578619 Chr.1: 196827040-196827040 on Build GRCh38
rs145410290 Chr.1: 196827333-196827333 on Build GRCh38
rs778362473 Chr.1: 196826966-196826966 on Build GRCh38
rs202193350 Chr.1: 196826962-196826962 on Build GRCh38
rs372835745 Chr.1: 196827005-196827005 on Build GRCh38
rs760092564 Chr.1: 196827009-196827009 on Build GRCh38
rs377142732 Chr.1: 196826994-196826994 on Build GRCh38
rs771140893 Chr.1: 196826992-196826992 on Build GRCh38
rs770052368 Chr.1: 196827015-196827015 on Build GRCh38
rs368078240 Chr.1: 196826945-196826945 on Build GRCh38
rs868759520 Chr.1: 196827241-196827241 on Build GRCh38
rs748377588 Chr.1: 196827199-196827199 on Build GRCh38
rs753812846 Chr.1: 196827049-196827049 on Build GRCh38
rs399507 Chr.1: 196827054-196827054 on Build GRCh38

View Full Product Details