Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196906884-196907380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1 XM_017000114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6024852 c.614A>G p.N205S Chr.1: 196907316-196907316 on Build GRCh38
3997122 c.626A>G p.N209S Chr.1: 196907328-196907328 on Build GRCh38
2124960 c.593C>A p.S198Y Chr.1: 196907017-196907017 on Build GRCh38
3480880 c.620C>T p.S207L Chr.1: 196907322-196907322 on Build GRCh38
5992751 c.475A>C p.K159Q Chr.1: 196906899-196906899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014463019 Chr.1: 196907290-196907290 on Build GRCh38
rs757628759 Chr.1: 196907334-196907334 on Build GRCh38
rs556777105 Chr.1: 196907305-196907305 on Build GRCh38
rs7417769 Chr.1: 196907328-196907328 on Build GRCh38
rs918428437 Chr.1: 196907052-196907052 on Build GRCh38
rs182228217 Chr.1: 196907123-196907123 on Build GRCh38
rs757709741 Chr.1: 196907053-196907053 on Build GRCh38
rs773186422 Chr.1: 196906987-196906987 on Build GRCh38
rs185135139 Chr.1: 196906935-196906935 on Build GRCh38
rs188089623 Chr.1: 196907084-196907084 on Build GRCh38
rs115298512 Chr.1: 196907316-196907316 on Build GRCh38
rs758872040 Chr.1: 196907035-196907035 on Build GRCh38
rs747986585 Chr.1: 196907339-196907339 on Build GRCh38
rs758820612 Chr.1: 196906923-196906923 on Build GRCh38
rs907844747 Chr.1: 196907233-196907233 on Build GRCh38
rs768821818 Chr.1: 196907273-196907273 on Build GRCh38
rs146529225 Chr.1: 196907127-196907127 on Build GRCh38
rs762716997 Chr.1: 196907317-196907317 on Build GRCh38
rs773026933 Chr.1: 196907289-196907289 on Build GRCh38
rs983427680 Chr.1: 196907150-196907150 on Build GRCh38
rs746259829 Chr.1: 196907130-196907130 on Build GRCh38
rs199731883 Chr.1: 196907017-196907017 on Build GRCh38
rs201837888 Chr.1: 196907180-196907180 on Build GRCh38
rs578099508 Chr.1: 196906952-196906952 on Build GRCh38
rs765930678 Chr.1: 196907060-196907061 on Build GRCh38
rs964900638 Chr.1: 196907119-196907119 on Build GRCh38
rs780815269 Chr.1: 196906976-196906976 on Build GRCh38
rs749493363 Chr.1: 196907271-196907271 on Build GRCh38
rs776595898 Chr.1: 196906960-196906960 on Build GRCh38
rs1032179541 Chr.1: 196907113-196907113 on Build GRCh38
rs754976122 Chr.1: 196907305-196907305 on Build GRCh38
rs771183005 Chr.1: 196906954-196906954 on Build GRCh38
rs527775900 Chr.1: 196907049-196907049 on Build GRCh38
rs1031803778 Chr.1: 196907187-196907187 on Build GRCh38
rs769915071 Chr.1: 196907083-196907083 on Build GRCh38
rs765787778 Chr.1: 196906911-196906911 on Build GRCh38
rs549670885 Chr.1: 196907296-196907296 on Build GRCh38
rs548732013 Chr.1: 196907252-196907252 on Build GRCh38
rs764771054 Chr.1: 196907307-196907307 on Build GRCh38
rs745894813 Chr.1: 196906970-196906970 on Build GRCh38
rs551159807 Chr.1: 196907161-196907161 on Build GRCh38
rs761094915 Chr.1: 196906969-196906970 on Build GRCh38
rs757908026 Chr.1: 196907037-196907040 on Build GRCh38
rs748471569 Chr.1: 196906917-196906917 on Build GRCh38
rs200259127 Chr.1: 196907294-196907294 on Build GRCh38
rs778557552 Chr.1: 196906915-196906915 on Build GRCh38
rs1030194805 Chr.1: 196907129-196907129 on Build GRCh38
rs767271778 Chr.1: 196906986-196906986 on Build GRCh38
rs1000990616 Chr.1: 196907095-196907095 on Build GRCh38
rs750100833 Chr.1: 196906995-196906996 on Build GRCh38
rs34175798 Chr.1: 196907304-196907304 on Build GRCh38
rs769923643 Chr.1: 196906977-196906977 on Build GRCh38
rs758645335 Chr.1: 196907139-196907139 on Build GRCh38
rs765018542 Chr.1: 196906970-196906970 on Build GRCh38
rs114567637 Chr.1: 196907061-196907061 on Build GRCh38
rs776614847 Chr.1: 196907298-196907298 on Build GRCh38
rs937646695 Chr.1: 196907006-196907006 on Build GRCh38
rs191740006 Chr.1: 196907240-196907240 on Build GRCh38
rs781682806 Chr.1: 196907055-196907055 on Build GRCh38
rs529507599 Chr.1: 196907154-196907154 on Build GRCh38
rs766295702 Chr.1: 196907011-196907011 on Build GRCh38
rs1016964376 Chr.1: 196907312-196907312 on Build GRCh38
rs560613052 Chr.1: 196907034-196907034 on Build GRCh38
rs970459942 Chr.1: 196906966-196906966 on Build GRCh38
rs764853537 Chr.1: 196907022-196907022 on Build GRCh38
rs778259654 Chr.1: 196907046-196907046 on Build GRCh38
rs566272368 Chr.1: 196907180-196907180 on Build GRCh38
rs900364110 Chr.1: 196907352-196907352 on Build GRCh38
rs754614145 Chr.1: 196906914-196906914 on Build GRCh38
rs1000661123 Chr.1: 196907342-196907342 on Build GRCh38
rs879619527 Chr.1: 196907071-196907071 on Build GRCh38
rs554389553 Chr.1: 196907329-196907329 on Build GRCh38
rs902277059 Chr.1: 196907089-196907089 on Build GRCh38
rs775770425 Chr.1: 196906982-196906982 on Build GRCh38
rs973892306 Chr.1: 196907261-196907261 on Build GRCh38
rs780943969 Chr.1: 196907304-196907305 on Build GRCh38
rs760633052 Chr.1: 196906993-196906993 on Build GRCh38
rs1053939573 Chr.1: 196907350-196907350 on Build GRCh38
rs761665547 Chr.1: 196906984-196906984 on Build GRCh38
rs187365627 Chr.1: 196907148-196907148 on Build GRCh38
rs757517178 Chr.1: 196906969-196906969 on Build GRCh38
rs527879165 Chr.1: 196907110-196907111 on Build GRCh38
rs201225095 Chr.1: 196906912-196906912 on Build GRCh38
rs566735089 Chr.1: 196907110-196907110 on Build GRCh38
rs562549283 Chr.1: 196907137-196907137 on Build GRCh38
rs750969942 Chr.1: 196907304-196907304 on Build GRCh38

View Full Product Details