Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 198296996-198297530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
-
UniGene
Hs.24119
Gene Chromosome Location
Chr.1: 198156978-198322420 on Build GRCh38
Transcripts
NM_133494.2 XM_011509209.1 XM_017000344.1 XM_017000345.1 XM_017000346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3481158 c.765C>T p.Y255Y Chr.1: 198297207-198297207 on Build GRCh38
6576316 c.716A>G p.D239G Chr.1: 198297158-198297158 on Build GRCh38
3481159 c.776C>T p.P259L Chr.1: 198297218-198297218 on Build GRCh38
1659263 c.699A>G p.Q233Q Chr.1: 198297141-198297141 on Build GRCh38
5181953 c.717C>T p.D239D Chr.1: 198297159-198297159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369279714 Chr.1: 198297271-198297271 on Build GRCh38
rs892907807 Chr.1: 198297361-198297374 on Build GRCh38
rs754231613 Chr.1: 198297259-198297259 on Build GRCh38
rs912501761 Chr.1: 198297163-198297170 on Build GRCh38
rs148698699 Chr.1: 198297203-198297203 on Build GRCh38
rs772191165 Chr.1: 198297115-198297115 on Build GRCh38
rs775369930 Chr.1: 198297177-198297177 on Build GRCh38
rs781081703 Chr.1: 198297133-198297133 on Build GRCh38
rs140258543 Chr.1: 198297426-198297426 on Build GRCh38
rs372604967 Chr.1: 198297052-198297052 on Build GRCh38
rs759785600 Chr.1: 198297050-198297050 on Build GRCh38
rs116170469 Chr.1: 198297316-198297316 on Build GRCh38
rs1013606592 Chr.1: 198297353-198297353 on Build GRCh38
rs892431875 Chr.1: 198297347-198297347 on Build GRCh38
rs780628698 Chr.1: 198297088-198297088 on Build GRCh38
rs748827203 Chr.1: 198297180-198297180 on Build GRCh38
rs756516013 Chr.1: 198297084-198297084 on Build GRCh38
rs973760375 Chr.1: 198297469-198297469 on Build GRCh38
rs924162697 Chr.1: 198297396-198297396 on Build GRCh38
rs372109327 Chr.1: 198297103-198297103 on Build GRCh38
rs978306011 Chr.1: 198297376-198297376 on Build GRCh38
rs973706200 Chr.1: 198297053-198297053 on Build GRCh38
rs546439239 Chr.1: 198297450-198297450 on Build GRCh38
rs766788903 Chr.1: 198297256-198297256 on Build GRCh38
rs371217816 Chr.1: 198297332-198297332 on Build GRCh38
rs745875793 Chr.1: 198297152-198297152 on Build GRCh38
rs753989788 Chr.1: 198297092-198297092 on Build GRCh38
rs751015886 Chr.1: 198297082-198297082 on Build GRCh38
rs769564996 Chr.1: 198297173-198297173 on Build GRCh38
rs535272186 Chr.1: 198297073-198297073 on Build GRCh38
rs1025629340 Chr.1: 198297356-198297356 on Build GRCh38
rs574534692 Chr.1: 198297449-198297449 on Build GRCh38
rs768082605 Chr.1: 198297369-198297370 on Build GRCh38
rs545784396 Chr.1: 198297391-198297391 on Build GRCh38
rs1025408007 Chr.1: 198297501-198297501 on Build GRCh38
rs559148232 Chr.1: 198297051-198297051 on Build GRCh38
rs958326349 Chr.1: 198297409-198297409 on Build GRCh38
rs918214655 Chr.1: 198297076-198297076 on Build GRCh38
rs761319466 Chr.1: 198297255-198297255 on Build GRCh38
rs764640301 Chr.1: 198297467-198297467 on Build GRCh38
rs575469094 Chr.1: 198297362-198297362 on Build GRCh38
rs758473597 Chr.1: 198297284-198297284 on Build GRCh38
rs773847675 Chr.1: 198297247-198297247 on Build GRCh38
rs1052528782 Chr.1: 198297343-198297343 on Build GRCh38
rs963517480 Chr.1: 198297456-198297456 on Build GRCh38
rs186933869 Chr.1: 198297333-198297333 on Build GRCh38
rs1051559770 Chr.1: 198297059-198297059 on Build GRCh38
rs899107090 Chr.1: 198297098-198297098 on Build GRCh38
rs769954765 Chr.1: 198297283-198297283 on Build GRCh38
rs541794600 Chr.1: 198297326-198297326 on Build GRCh38
rs777929623 Chr.1: 198297116-198297118 on Build GRCh38
rs773210797 Chr.1: 198297503-198297503 on Build GRCh38
rs767176141 Chr.1: 198297240-198297240 on Build GRCh38
rs775957627 Chr.1: 198297430-198297430 on Build GRCh38
rs959409069 Chr.1: 198297116-198297116 on Build GRCh38
rs116732910 Chr.1: 198297109-198297109 on Build GRCh38
rs748284573 Chr.1: 198297110-198297110 on Build GRCh38
rs1002318207 Chr.1: 198297403-198297403 on Build GRCh38
rs768387407 Chr.1: 198297182-198297182 on Build GRCh38
rs765478190 Chr.1: 198297274-198297274 on Build GRCh38
rs761363477 Chr.1: 198297228-198297228 on Build GRCh38

View Full Product Details