Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200650233-200650732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX59
Entrez Gene ID
83479
Gene Name
DEAD-box helicase 59
Gene Aliases
OFD5, ZNHIT5
UniGene
Hs.497332
Gene Chromosome Location
Chr.1: 200640807-200669998 on Build GRCh38
Transcripts
NM_001031725.5 NM_001320181.1 NM_001320182.1 XM_005245519.1 XM_005245521.2 XM_006711562.1 XM_011510035.2 XM_017002431.1 XM_017002432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
70425 c.1142T>C p.I381T Chr.1: 200650597-200650597 on Build GRCh38
1337700 c.1294A>G p.K432E Chr.1: 200650445-200650445 on Build GRCh38
1337701 c.1211G>T p.R404I Chr.1: 200650528-200650528 on Build GRCh38
4026789 c.1230G>T p.K410N Chr.1: 200650509-200650509 on Build GRCh38
5435583 c.1063-14_1063-3delCAATTTTATTTT p.? Chr.1: 200650679-200650690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554889172 Chr.1: 200650555-200650555 on Build GRCh38
rs779909611 Chr.1: 200650443-200650443 on Build GRCh38
rs766384817 Chr.1: 200650504-200650504 on Build GRCh38
rs998128614 Chr.1: 200650262-200650262 on Build GRCh38
rs893953076 Chr.1: 200650344-200650344 on Build GRCh38
rs143031313 Chr.1: 200650539-200650539 on Build GRCh38
rs762738543 Chr.1: 200650322-200650322 on Build GRCh38
rs950639462 Chr.1: 200650590-200650590 on Build GRCh38
rs984321387 Chr.1: 200650396-200650396 on Build GRCh38
rs770302480 Chr.1: 200650605-200650605 on Build GRCh38
rs763283015 Chr.1: 200650401-200650402 on Build GRCh38
rs764228907 Chr.1: 200650488-200650488 on Build GRCh38
rs147185513 Chr.1: 200650644-200650644 on Build GRCh38
rs927997802 Chr.1: 200650487-200650487 on Build GRCh38
rs759864493 Chr.1: 200650475-200650475 on Build GRCh38
rs765331144 Chr.1: 200650660-200650660 on Build GRCh38
rs746570294 Chr.1: 200650459-200650459 on Build GRCh38
rs147441430 Chr.1: 200650636-200650636 on Build GRCh38
rs757327015 Chr.1: 200650499-200650499 on Build GRCh38
rs1025559665 Chr.1: 200650324-200650324 on Build GRCh38
rs751642340 Chr.1: 200650492-200650492 on Build GRCh38
rs777350083 Chr.1: 200650540-200650540 on Build GRCh38
rs587777067 Chr.1: 200650639-200650639 on Build GRCh38
rs777450958 Chr.1: 200650682-200650682 on Build GRCh38
rs763297418 Chr.1: 200650625-200650625 on Build GRCh38
rs753783027 Chr.1: 200650505-200650505 on Build GRCh38
rs746329013 Chr.1: 200650599-200650599 on Build GRCh38
rs969855320 Chr.1: 200650432-200650432 on Build GRCh38
rs747753860 Chr.1: 200650412-200650412 on Build GRCh38
rs932863694 Chr.1: 200650313-200650313 on Build GRCh38
rs377060361 Chr.1: 200650654-200650654 on Build GRCh38
rs145102433 Chr.1: 200650526-200650526 on Build GRCh38
rs569964371 Chr.1: 200650281-200650281 on Build GRCh38
rs749066586 Chr.1: 200650381-200650381 on Build GRCh38
rs766495360 Chr.1: 200650621-200650621 on Build GRCh38
rs1054553140 Chr.1: 200650658-200650658 on Build GRCh38
rs372512251 Chr.1: 200650613-200650613 on Build GRCh38
rs35667326 Chr.1: 200650283-200650284 on Build GRCh38
rs879109616 Chr.1: 200650513-200650513 on Build GRCh38
rs772852928 Chr.1: 200650400-200650401 on Build GRCh38
rs758177893 Chr.1: 200650673-200650673 on Build GRCh38
rs747898610 Chr.1: 200650530-200650530 on Build GRCh38
rs2809365 Chr.1: 200650402-200650402 on Build GRCh38
rs369141532 Chr.1: 200650424-200650424 on Build GRCh38
rs776150281 Chr.1: 200650580-200650580 on Build GRCh38
rs552386742 Chr.1: 200650323-200650323 on Build GRCh38
rs141148503 Chr.1: 200650562-200650562 on Build GRCh38
rs530769328 Chr.1: 200650319-200650319 on Build GRCh38
rs377485552 Chr.1: 200650455-200650455 on Build GRCh38
rs778882910 Chr.1: 200650409-200650409 on Build GRCh38
rs762123107 Chr.1: 200650641-200650641 on Build GRCh38
rs2808230 Chr.1: 200650350-200650350 on Build GRCh38
rs1132058 Chr.1: 200650449-200650449 on Build GRCh38
rs774638984 Chr.1: 200650641-200650643 on Build GRCh38
rs750542771 Chr.1: 200650647-200650647 on Build GRCh38
rs146518838 Chr.1: 200650507-200650507 on Build GRCh38
rs771458131 Chr.1: 200650465-200650465 on Build GRCh38
rs78882239 Chr.1: 200650486-200650486 on Build GRCh38
rs370730702 Chr.1: 200650479-200650479 on Build GRCh38
rs752524933 Chr.1: 200650666-200650666 on Build GRCh38
rs1003585389 Chr.1: 200650451-200650451 on Build GRCh38
rs777172345 Chr.1: 200650471-200650471 on Build GRCh38

View Full Product Details