Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201154304-201154833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM9
Entrez Gene ID
252839
Gene Name
transmembrane protein 9
Gene Aliases
DERM4, TMEM9A
UniGene
Hs.181444
Gene Chromosome Location
Chr.1: 201134771-201171625 on Build GRCh38
Transcripts
NM_001288564.1 NM_001288565.1 NM_001288566.1 NM_001288567.1 NM_001288568.1 NM_001288569.1 NM_001288570.1 NM_001288571.1 NM_016456.4 NR_109920.1 NR_109921.1 XM_011509385.1 XM_011509386.2 XM_011509387.2

SNP Detail

SNP ID Chromosome Location
rs971134542 Chr.1: 201154808-201154808 on Build GRCh38
rs1023671038 Chr.1: 201154768-201154768 on Build GRCh38
rs1012899990 Chr.1: 201154767-201154767 on Build GRCh38
rs182618237 Chr.1: 201154810-201154810 on Build GRCh38
rs747949315 Chr.1: 201154625-201154625 on Build GRCh38
rs920224555 Chr.1: 201154524-201154524 on Build GRCh38
rs746114117 Chr.1: 201154458-201154458 on Build GRCh38
rs1027080215 Chr.1: 201154727-201154727 on Build GRCh38
rs934599342 Chr.1: 201154456-201154456 on Build GRCh38
rs763058292 Chr.1: 201154331-201154331 on Build GRCh38
rs914508012 Chr.1: 201154624-201154624 on Build GRCh38
rs536452186 Chr.1: 201154367-201154367 on Build GRCh38
rs990848537 Chr.1: 201154553-201154553 on Build GRCh38
rs760462902 Chr.1: 201154774-201154774 on Build GRCh38
rs1018804997 Chr.1: 201154395-201154395 on Build GRCh38
rs903018026 Chr.1: 201154639-201154639 on Build GRCh38
rs767743346 Chr.1: 201154534-201154534 on Build GRCh38
rs1045592355 Chr.1: 201154423-201154423 on Build GRCh38
rs893021294 Chr.1: 201154459-201154459 on Build GRCh38
rs961515749 Chr.1: 201154483-201154483 on Build GRCh38
rs889434383 Chr.1: 201154678-201154678 on Build GRCh38
rs971540210 Chr.1: 201154477-201154477 on Build GRCh38
rs928492652 Chr.1: 201154600-201154600 on Build GRCh38
rs940308022 Chr.1: 201154497-201154497 on Build GRCh38
rs202192014 Chr.1: 201154391-201154392 on Build GRCh38
rs1040767293 Chr.1: 201154700-201154700 on Build GRCh38
rs560818267 Chr.1: 201154567-201154567 on Build GRCh38
rs942709 Chr.1: 201154701-201154701 on Build GRCh38
rs1106729 Chr.1: 201154671-201154671 on Build GRCh38
rs991187905 Chr.1: 201154386-201154386 on Build GRCh38
rs3753964 Chr.1: 201154653-201154653 on Build GRCh38
rs1049834689 Chr.1: 201154464-201154464 on Build GRCh38
rs1027195618 Chr.1: 201154522-201154522 on Build GRCh38
rs76351351 Chr.1: 201154796-201154796 on Build GRCh38
rs7541591 Chr.1: 201154372-201154372 on Build GRCh38
rs908788890 Chr.1: 201154487-201154487 on Build GRCh38
rs994417195 Chr.1: 201154519-201154519 on Build GRCh38
rs981606864 Chr.1: 201154752-201154752 on Build GRCh38
rs949469919 Chr.1: 201154411-201154411 on Build GRCh38
rs981637974 Chr.1: 201154486-201154486 on Build GRCh38
rs570467609 Chr.1: 201154713-201154713 on Build GRCh38
rs754611963 Chr.1: 201154587-201154587 on Build GRCh38
rs532175041 Chr.1: 201154701-201154701 on Build GRCh38
rs557853480 Chr.1: 201154471-201154471 on Build GRCh38
rs191296989 Chr.1: 201154757-201154757 on Build GRCh38
rs934484962 Chr.1: 201154601-201154601 on Build GRCh38
rs928440115 Chr.1: 201154439-201154439 on Build GRCh38
rs1011250605 Chr.1: 201154463-201154463 on Build GRCh38
rs1019768134 Chr.1: 201154479-201154479 on Build GRCh38
rs544254005 Chr.1: 201154461-201154461 on Build GRCh38
rs1008371739 Chr.1: 201154474-201154474 on Build GRCh38
rs879382766 Chr.1: 201154408-201154408 on Build GRCh38
rs769960753 Chr.1: 201154573-201154573 on Build GRCh38
rs530522686 Chr.1: 201154691-201154691 on Build GRCh38
rs994302767 Chr.1: 201154712-201154712 on Build GRCh38
rs1033388121 Chr.1: 201154417-201154417 on Build GRCh38
rs1010502 Chr.1: 201154617-201154617 on Build GRCh38
rs1050144553 Chr.1: 201154370-201154370 on Build GRCh38
rs1008009563 Chr.1: 201154696-201154696 on Build GRCh38
rs770804414 Chr.1: 201154760-201154760 on Build GRCh38
rs953009686 Chr.1: 201154729-201154729 on Build GRCh38
rs575248965 Chr.1: 201154389-201154389 on Build GRCh38
rs546503636 Chr.1: 201154776-201154776 on Build GRCh38
rs374808665 Chr.1: 201154531-201154531 on Build GRCh38
rs116294744 Chr.1: 201154533-201154533 on Build GRCh38
rs764710263 Chr.1: 201154334-201154334 on Build GRCh38
rs1053022203 Chr.1: 201154457-201154457 on Build GRCh38
rs916677133 Chr.1: 201154399-201154399 on Build GRCh38
rs778590410 Chr.1: 201154593-201154593 on Build GRCh38

View Full Product Details