Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201852786-201853206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO9
Entrez Gene ID
55705
Gene Name
importin 9
Gene Aliases
Imp9
UniGene
Hs.596014
Gene Chromosome Location
Chr.1: 201829160-201884294 on Build GRCh38
Transcripts
NM_018085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502923 c.626C>T p.S209F Chr.1: 201853033-201853033 on Build GRCh38
5477909 c.616C>T p.R206* Chr.1: 201853023-201853023 on Build GRCh38
4692852 c.628C>T p.R210* Chr.1: 201853035-201853035 on Build GRCh38
6686736 c.632C>T p.A211V Chr.1: 201853039-201853039 on Build GRCh38
6686743 c.605T>C p.V202A Chr.1: 201853012-201853012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140251055 Chr.1: 201853049-201853049 on Build GRCh38
rs546202746 Chr.1: 201852853-201852853 on Build GRCh38
rs535374381 Chr.1: 201852918-201852918 on Build GRCh38
rs766926811 Chr.1: 201853046-201853046 on Build GRCh38
rs147704344 Chr.1: 201853080-201853080 on Build GRCh38
rs779959255 Chr.1: 201853113-201853113 on Build GRCh38
rs758610598 Chr.1: 201853081-201853081 on Build GRCh38
rs773526749 Chr.1: 201852997-201852997 on Build GRCh38
rs151321512 Chr.1: 201852970-201852970 on Build GRCh38
rs772300675 Chr.1: 201852912-201852912 on Build GRCh38
rs768361477 Chr.1: 201853013-201853013 on Build GRCh38
rs564287960 Chr.1: 201852910-201852910 on Build GRCh38
rs375505268 Chr.1: 201853078-201853078 on Build GRCh38
rs200050020 Chr.1: 201853108-201853108 on Build GRCh38
rs979294839 Chr.1: 201852953-201852953 on Build GRCh38
rs74947610 Chr.1: 201853002-201853002 on Build GRCh38
rs749012804 Chr.1: 201853008-201853008 on Build GRCh38
rs769031530 Chr.1: 201852982-201852982 on Build GRCh38
rs572690457 Chr.1: 201852863-201852863 on Build GRCh38
rs754742701 Chr.1: 201853118-201853118 on Build GRCh38
rs546528389 Chr.1: 201853033-201853033 on Build GRCh38
rs781235018 Chr.1: 201852987-201852987 on Build GRCh38
rs759967942 Chr.1: 201853066-201853066 on Build GRCh38
rs948217500 Chr.1: 201853159-201853159 on Build GRCh38
rs910052277 Chr.1: 201853152-201853152 on Build GRCh38
rs900764585 Chr.1: 201853154-201853154 on Build GRCh38
rs917655959 Chr.1: 201852998-201852998 on Build GRCh38
rs755257524 Chr.1: 201852976-201852976 on Build GRCh38
rs766873523 Chr.1: 201852968-201852968 on Build GRCh38
rs987029071 Chr.1: 201852881-201852881 on Build GRCh38
rs765547478 Chr.1: 201853073-201853073 on Build GRCh38
rs764107524 Chr.1: 201853096-201853096 on Build GRCh38
rs767482887 Chr.1: 201853082-201853084 on Build GRCh38
rs759529773 Chr.1: 201852989-201852990 on Build GRCh38
rs1030791382 Chr.1: 201852848-201852848 on Build GRCh38
rs954855339 Chr.1: 201852876-201852876 on Build GRCh38
rs748287808 Chr.1: 201852880-201852880 on Build GRCh38
rs771697629 Chr.1: 201853144-201853144 on Build GRCh38
rs1045353915 Chr.1: 201853177-201853177 on Build GRCh38
rs370406011 Chr.1: 201853155-201853155 on Build GRCh38
rs575059014 Chr.1: 201852959-201852959 on Build GRCh38
rs772737508 Chr.1: 201853040-201853040 on Build GRCh38
rs755992542 Chr.1: 201852993-201852993 on Build GRCh38
rs747786220 Chr.1: 201853143-201853143 on Build GRCh38
rs191541517 Chr.1: 201853041-201853041 on Build GRCh38
rs147724148 Chr.1: 201853116-201853116 on Build GRCh38
rs1010584178 Chr.1: 201852852-201852852 on Build GRCh38
rs555179454 Chr.1: 201852955-201852955 on Build GRCh38
rs762060179 Chr.1: 201853019-201853019 on Build GRCh38
rs745827128 Chr.1: 201852991-201852991 on Build GRCh38
rs1020812889 Chr.1: 201852906-201852907 on Build GRCh38
rs773527392 Chr.1: 201852949-201852950 on Build GRCh38
rs778666009 Chr.1: 201853126-201853126 on Build GRCh38
rs1022136493 Chr.1: 201852890-201852890 on Build GRCh38
rs747742098 Chr.1: 201853021-201853021 on Build GRCh38
rs758388798 Chr.1: 201852984-201852984 on Build GRCh38

View Full Product Details