Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203870494-203870990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPE
Entrez Gene ID
6635
Gene Name
small nuclear ribonucleoprotein polypeptide E
Gene Aliases
HYPT11, SME, Sm-E, snRNP-E
UniGene
Hs.334612
Gene Chromosome Location
Chr.1: 203861585-203871152 on Build GRCh38
Transcripts
NM_001304464.1 NM_003094.3 NR_130746.1

SNP Detail

SNP ID Chromosome Location
rs913775817 Chr.1: 203870719-203870719 on Build GRCh38
rs913551080 Chr.1: 203870938-203870938 on Build GRCh38
rs879685828 Chr.1: 203870725-203870725 on Build GRCh38
rs987573390 Chr.1: 203870610-203870610 on Build GRCh38
rs989523905 Chr.1: 203870927-203870927 on Build GRCh38
rs921057092 Chr.1: 203870675-203870675 on Build GRCh38
rs927905263 Chr.1: 203870959-203870959 on Build GRCh38
rs973456553 Chr.1: 203870842-203870842 on Build GRCh38
rs541911689 Chr.1: 203870820-203870820 on Build GRCh38
rs375044740 Chr.1: 203870753-203870753 on Build GRCh38
rs535550741 Chr.1: 203870521-203870521 on Build GRCh38
rs183934532 Chr.1: 203870598-203870598 on Build GRCh38
rs549005560 Chr.1: 203870632-203870632 on Build GRCh38
rs538627140 Chr.1: 203870689-203870689 on Build GRCh38
rs994884542 Chr.1: 203870569-203870569 on Build GRCh38
rs752357329 Chr.1: 203870527-203870527 on Build GRCh38
rs999904486 Chr.1: 203870732-203870732 on Build GRCh38
rs576977653 Chr.1: 203870766-203870766 on Build GRCh38
rs970875698 Chr.1: 203870778-203870778 on Build GRCh38
rs995211438 Chr.1: 203870800-203870800 on Build GRCh38
rs890311380 Chr.1: 203870677-203870677 on Build GRCh38
rs116257093 Chr.1: 203870817-203870817 on Build GRCh38
rs921825911 Chr.1: 203870869-203870869 on Build GRCh38
rs757856767 Chr.1: 203870528-203870528 on Build GRCh38
rs987806251 Chr.1: 203870893-203870893 on Build GRCh38
rs539631127 Chr.1: 203870720-203870720 on Build GRCh38
rs4598553 Chr.1: 203870909-203870909 on Build GRCh38
rs1026325436 Chr.1: 203870812-203870812 on Build GRCh38
rs11240611 Chr.1: 203870761-203870761 on Build GRCh38
rs937785607 Chr.1: 203870960-203870960 on Build GRCh38
rs1019187431 Chr.1: 203870825-203870825 on Build GRCh38
rs150578163 Chr.1: 203870705-203870705 on Build GRCh38
rs755821493 Chr.1: 203870810-203870813 on Build GRCh38
rs982017666 Chr.1: 203870956-203870956 on Build GRCh38
rs974978696 Chr.1: 203870907-203870907 on Build GRCh38
rs964842648 Chr.1: 203870877-203870877 on Build GRCh38
rs899248265 Chr.1: 203870797-203870797 on Build GRCh38
rs931893229 Chr.1: 203870885-203870885 on Build GRCh38
rs764824747 Chr.1: 203870518-203870518 on Build GRCh38
rs141785435 Chr.1: 203870653-203870653 on Build GRCh38
rs957821511 Chr.1: 203870910-203870910 on Build GRCh38
rs950974752 Chr.1: 203870607-203870607 on Build GRCh38
rs964875047 Chr.1: 203870641-203870641 on Build GRCh38
rs1033000663 Chr.1: 203870751-203870751 on Build GRCh38
rs963698945 Chr.1: 203870519-203870519 on Build GRCh38
rs567181830 Chr.1: 203870787-203870787 on Build GRCh38
rs942037446 Chr.1: 203870536-203870536 on Build GRCh38
rs757798634 Chr.1: 203870703-203870703 on Build GRCh38
rs945051568 Chr.1: 203870955-203870955 on Build GRCh38

View Full Product Details