Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205228114-205228616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCC2
Entrez Gene ID
9911
Gene Name
transmembrane and coiled-coil domain family 2
Gene Aliases
HUCEP11
UniGene
Hs.6360
Gene Chromosome Location
Chr.1: 205227910-205273343 on Build GRCh38
Transcripts
NM_014858.3 XM_005245686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415314 c.26T>A p.L9Q Chr.1: 205228590-205228590 on Build GRCh38
6124033 c.49G>A p.D17N Chr.1: 205228613-205228613 on Build GRCh38
458299 c.3G>A p.M1I Chr.1: 205228567-205228567 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951837895 Chr.1: 205228175-205228175 on Build GRCh38
rs113162818 Chr.1: 205228133-205228133 on Build GRCh38
rs772354233 Chr.1: 205228517-205228517 on Build GRCh38
rs556864194 Chr.1: 205228143-205228143 on Build GRCh38
rs1026211227 Chr.1: 205228163-205228163 on Build GRCh38
rs78605680 Chr.1: 205228151-205228151 on Build GRCh38
rs1035685853 Chr.1: 205228358-205228358 on Build GRCh38
rs905954730 Chr.1: 205228329-205228329 on Build GRCh38
rs550476336 Chr.1: 205228434-205228434 on Build GRCh38
rs1002040843 Chr.1: 205228132-205228132 on Build GRCh38
rs777167743 Chr.1: 205228553-205228553 on Build GRCh38
rs1026690961 Chr.1: 205228412-205228412 on Build GRCh38
rs984472851 Chr.1: 205228181-205228181 on Build GRCh38
rs774927345 Chr.1: 205228576-205228576 on Build GRCh38
rs910297631 Chr.1: 205228189-205228189 on Build GRCh38
rs900826403 Chr.1: 205228524-205228524 on Build GRCh38
rs976235120 Chr.1: 205228226-205228226 on Build GRCh38
rs956240144 Chr.1: 205228502-205228502 on Build GRCh38
rs747366355 Chr.1: 205228545-205228545 on Build GRCh38
rs964449115 Chr.1: 205228459-205228459 on Build GRCh38
rs908752330 Chr.1: 205228487-205228487 on Build GRCh38
rs763719909 Chr.1: 205228585-205228585 on Build GRCh38
rs545559958 Chr.1: 205228251-205228251 on Build GRCh38
rs541214389 Chr.1: 205228353-205228353 on Build GRCh38
rs964397966 Chr.1: 205228219-205228219 on Build GRCh38
rs916110767 Chr.1: 205228387-205228387 on Build GRCh38
rs1032010577 Chr.1: 205228266-205228266 on Build GRCh38
rs559132096 Chr.1: 205228273-205228273 on Build GRCh38
rs34677862 Chr.1: 205228196-205228196 on Build GRCh38
rs1044364169 Chr.1: 205228324-205228324 on Build GRCh38
rs79431750 Chr.1: 205228474-205228474 on Build GRCh38
rs199536910 Chr.1: 205228546-205228546 on Build GRCh38
rs975827988 Chr.1: 205228473-205228473 on Build GRCh38
rs1053810470 Chr.1: 205228254-205228254 on Build GRCh38
rs113745983 Chr.1: 205228365-205228365 on Build GRCh38
rs960624380 Chr.1: 205228154-205228154 on Build GRCh38
rs991964257 Chr.1: 205228377-205228377 on Build GRCh38
rs778349573 Chr.1: 205228522-205228522 on Build GRCh38
rs769372518 Chr.1: 205228561-205228561 on Build GRCh38
rs532862096 Chr.1: 205228506-205228506 on Build GRCh38
rs77230573 Chr.1: 205228229-205228229 on Build GRCh38
rs993316258 Chr.1: 205228403-205228403 on Build GRCh38
rs374736860 Chr.1: 205228554-205228554 on Build GRCh38
rs528231594 Chr.1: 205228361-205228361 on Build GRCh38
rs78966833 Chr.1: 205228311-205228311 on Build GRCh38
rs993388431 Chr.1: 205228161-205228161 on Build GRCh38
rs914820292 Chr.1: 205228266-205228266 on Build GRCh38
rs779827219 Chr.1: 205228581-205228581 on Build GRCh38
rs977386643 Chr.1: 205228337-205228337 on Build GRCh38
rs940268471 Chr.1: 205228508-205228508 on Build GRCh38
rs530075613 Chr.1: 205228398-205228398 on Build GRCh38
rs887663967 Chr.1: 205228413-205228413 on Build GRCh38

View Full Product Details