Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206012769-206012274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSE
Entrez Gene ID
1510
Gene Name
cathepsin E
Gene Aliases
CATE
UniGene
Hs.644082
Gene Chromosome Location
Chr.1: 206009264-206023909 on Build GRCh38
Transcripts
NM_001317331.1 NM_001910.3 NM_148964.2 XM_011509244.1 XM_011509245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4170579 c.981T>A p.I327I Chr.1: 206012353-206012353 on Build GRCh38
3482393 c.919G>A p.D307N Chr.1: 206012516-206012516 on Build GRCh38
5379396 c.833C>T p.A278V Chr.1: 206012602-206012602 on Build GRCh38
2215842 c.936G>T p.V312V Chr.1: 206012398-206012398 on Build GRCh38
902913 c.826T>C p.C276R Chr.1: 206012609-206012609 on Build GRCh38
View More

View Full Product Details