Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 211491685-211492193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RD3
Entrez Gene ID
343035
Gene Name
retinal degeneration 3
Gene Aliases
C1orf36, LCA12
UniGene
Hs.632495
Gene Chromosome Location
Chr.1: 211476522-211492917 on Build GRCh38
Transcripts
NM_001164688.1 NM_183059.2

SNP Detail

SNP ID Chromosome Location
rs1015430813 Chr.1: 211491831-211491831 on Build GRCh38
rs866013658 Chr.1: 211492152-211492152 on Build GRCh38
rs113540264 Chr.1: 211491956-211491956 on Build GRCh38
rs886045905 Chr.1: 211492047-211492048 on Build GRCh38
rs886045912 Chr.1: 211492058-211492058 on Build GRCh38
rs376931725 Chr.1: 211492096-211492096 on Build GRCh38
rs56742170 Chr.1: 211492085-211492092 on Build GRCh38
rs958424826 Chr.1: 211492131-211492131 on Build GRCh38
rs1020847478 Chr.1: 211491976-211491976 on Build GRCh38
rs924145121 Chr.1: 211491724-211491724 on Build GRCh38
rs886045900 Chr.1: 211491758-211491758 on Build GRCh38
rs187218461 Chr.1: 211491889-211491889 on Build GRCh38
rs1048579109 Chr.1: 211492088-211492088 on Build GRCh38
rs944168111 Chr.1: 211491714-211491714 on Build GRCh38
rs965590446 Chr.1: 211491983-211491983 on Build GRCh38
rs771444370 Chr.1: 211491762-211491762 on Build GRCh38
rs573005358 Chr.1: 211491945-211491945 on Build GRCh38
rs754348712 Chr.1: 211492049-211492056 on Build GRCh38
rs1044314717 Chr.1: 211491781-211491781 on Build GRCh38
rs543999042 Chr.1: 211491757-211491757 on Build GRCh38
rs886045917 Chr.1: 211492111-211492111 on Build GRCh38
rs938580182 Chr.1: 211492066-211492066 on Build GRCh38
rs556215060 Chr.1: 211491761-211491761 on Build GRCh38
rs187620033 Chr.1: 211492151-211492151 on Build GRCh38
rs574651452 Chr.1: 211491836-211491836 on Build GRCh38
rs995367990 Chr.1: 211492085-211492085 on Build GRCh38
rs999718090 Chr.1: 211492111-211492111 on Build GRCh38
rs965602916 Chr.1: 211492002-211492002 on Build GRCh38
rs969492717 Chr.1: 211492150-211492150 on Build GRCh38
rs940012498 Chr.1: 211492110-211492110 on Build GRCh38
rs17017574 Chr.1: 211492133-211492133 on Build GRCh38
rs767092998 Chr.1: 211491974-211491974 on Build GRCh38
rs200914005 Chr.1: 211491957-211491957 on Build GRCh38
rs749800342 Chr.1: 211492081-211492081 on Build GRCh38
rs368463447 Chr.1: 211491894-211491894 on Build GRCh38
rs940251158 Chr.1: 211491885-211491885 on Build GRCh38
rs866406243 Chr.1: 211492060-211492060 on Build GRCh38
rs886045916 Chr.1: 211492093-211492096 on Build GRCh38
rs367636756 Chr.1: 211492099-211492099 on Build GRCh38
rs889548597 Chr.1: 211492093-211492093 on Build GRCh38
rs577418198 Chr.1: 211491909-211491909 on Build GRCh38
rs921322313 Chr.1: 211491825-211491825 on Build GRCh38
rs200398344 Chr.1: 211492049-211492058 on Build GRCh38
rs886045911 Chr.1: 211492050-211492060 on Build GRCh38
rs565362913 Chr.1: 211492095-211492095 on Build GRCh38
rs886045904 Chr.1: 211492047-211492048 on Build GRCh38
rs978352861 Chr.1: 211492021-211492021 on Build GRCh38
rs886045908 Chr.1: 211492049-211492052 on Build GRCh38
rs1024737320 Chr.1: 211491919-211491920 on Build GRCh38
rs1048378123 Chr.1: 211491863-211491864 on Build GRCh38
rs897027891 Chr.1: 211491871-211491871 on Build GRCh38
rs913712325 Chr.1: 211491797-211491797 on Build GRCh38
rs927987385 Chr.1: 211492062-211492062 on Build GRCh38
rs202137622 Chr.1: 211492050-211492050 on Build GRCh38
rs886045914 Chr.1: 211492088-211492088 on Build GRCh38
rs532622973 Chr.1: 211492097-211492097 on Build GRCh38
rs886045907 Chr.1: 211492048-211492049 on Build GRCh38
rs879847361 Chr.1: 211492052-211492058 on Build GRCh38
rs34703190 Chr.1: 211491957-211491958 on Build GRCh38
rs559687793 Chr.1: 211492030-211492030 on Build GRCh38
rs1001909984 Chr.1: 211491949-211491949 on Build GRCh38
rs989943339 Chr.1: 211492052-211492052 on Build GRCh38
rs886045915 Chr.1: 211492092-211492093 on Build GRCh38
rs750022984 Chr.1: 211492041-211492041 on Build GRCh38
rs892050352 Chr.1: 211492059-211492059 on Build GRCh38
rs969055926 Chr.1: 211491746-211491746 on Build GRCh38
rs927935031 Chr.1: 211492169-211492169 on Build GRCh38
rs11463656 Chr.1: 211491956-211491957 on Build GRCh38
rs1043262232 Chr.1: 211492079-211492079 on Build GRCh38
rs1053897057 Chr.1: 211491754-211491754 on Build GRCh38
rs563896537 Chr.1: 211491902-211491908 on Build GRCh38
rs916690002 Chr.1: 211492056-211492056 on Build GRCh38
rs190784396 Chr.1: 211492170-211492170 on Build GRCh38
rs886045910 Chr.1: 211492050-211492058 on Build GRCh38
rs978236639 Chr.1: 211491717-211491717 on Build GRCh38
rs895990055 Chr.1: 211491901-211491901 on Build GRCh38
rs886045903 Chr.1: 211491919-211491920 on Build GRCh38
rs936882753 Chr.1: 211492048-211492048 on Build GRCh38
rs111236662 Chr.1: 211492072-211492072 on Build GRCh38
rs980399923 Chr.1: 211491760-211491760 on Build GRCh38
rs886045906 Chr.1: 211492047-211492047 on Build GRCh38
rs924094041 Chr.1: 211492047-211492047 on Build GRCh38
rs2884414 Chr.1: 211492051-211492051 on Build GRCh38
rs879176665 Chr.1: 211492049-211492060 on Build GRCh38
rs886045913 Chr.1: 211492085-211492085 on Build GRCh38
rs530374496 Chr.1: 211492148-211492148 on Build GRCh38
rs896969372 Chr.1: 211492084-211492084 on Build GRCh38
rs976523141 Chr.1: 211492053-211492053 on Build GRCh38
rs112616778 Chr.1: 211492071-211492071 on Build GRCh38
rs886045901 Chr.1: 211491781-211491781 on Build GRCh38
rs907122251 Chr.1: 211491798-211491798 on Build GRCh38
rs370878401 Chr.1: 211491872-211491873 on Build GRCh38
rs538031677 Chr.1: 211491725-211491725 on Build GRCh38
rs948010751 Chr.1: 211492058-211492058 on Build GRCh38
rs958193320 Chr.1: 211491774-211491774 on Build GRCh38
rs866734170 Chr.1: 211492087-211492087 on Build GRCh38
rs34485370 Chr.1: 211492049-211492050 on Build GRCh38
rs1002674153 Chr.1: 211491817-211491817 on Build GRCh38
rs886045902 Chr.1: 211491895-211491895 on Build GRCh38
rs909854415 Chr.1: 211492109-211492109 on Build GRCh38
rs886045909 Chr.1: 211492049-211492054 on Build GRCh38
rs77237846 Chr.1: 211492112-211492112 on Build GRCh38
rs879689866 Chr.1: 211492050-211492050 on Build GRCh38
rs985092962 Chr.1: 211491966-211491966 on Build GRCh38
rs551882177 Chr.1: 211491944-211491944 on Build GRCh38
rs1019788028 Chr.1: 211491972-211491972 on Build GRCh38
rs759999110 Chr.1: 211492157-211492157 on Build GRCh38
rs888737227 Chr.1: 211491864-211491864 on Build GRCh38

View Full Product Details