Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 214357077-214357575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN14
Entrez Gene ID
5784
Gene Name
protein tyrosine phosphatase, non-receptor type 14
Gene Aliases
PEZ, PTP36
UniGene
Hs.193557
Gene Chromosome Location
Chr.1: 214348696-214551681 on Build GRCh38
Transcripts
NM_005401.4 XM_017001941.1

SNP Detail

SNP ID Chromosome Location
rs995090778 Chr.1: 214357275-214357275 on Build GRCh38
rs924378752 Chr.1: 214357348-214357348 on Build GRCh38
rs543453582 Chr.1: 214357359-214357359 on Build GRCh38
rs953799782 Chr.1: 214357415-214357415 on Build GRCh38
rs1004289653 Chr.1: 214357245-214357245 on Build GRCh38
rs568369821 Chr.1: 214357197-214357197 on Build GRCh38
rs557441223 Chr.1: 214357313-214357313 on Build GRCh38
rs1048875259 Chr.1: 214357292-214357292 on Build GRCh38
rs148278834 Chr.1: 214357292-214357292 on Build GRCh38
rs957502246 Chr.1: 214357350-214357350 on Build GRCh38
rs575113987 Chr.1: 214357409-214357409 on Build GRCh38
rs550793756 Chr.1: 214357465-214357465 on Build GRCh38
rs771075893 Chr.1: 214357407-214357407 on Build GRCh38
rs202202833 Chr.1: 214357118-214357118 on Build GRCh38
rs114190994 Chr.1: 214357358-214357358 on Build GRCh38
rs936195458 Chr.1: 214357131-214357131 on Build GRCh38
rs188413998 Chr.1: 214357185-214357185 on Build GRCh38
rs73092358 Chr.1: 214357109-214357109 on Build GRCh38
rs147564603 Chr.1: 214357153-214357153 on Build GRCh38
rs535369684 Chr.1: 214357225-214357225 on Build GRCh38
rs943830183 Chr.1: 214357250-214357250 on Build GRCh38
rs183742544 Chr.1: 214357105-214357105 on Build GRCh38
rs948710992 Chr.1: 214357147-214357147 on Build GRCh38
rs1035710778 Chr.1: 214357298-214357298 on Build GRCh38
rs995525097 Chr.1: 214357285-214357286 on Build GRCh38
rs977121751 Chr.1: 214357316-214357316 on Build GRCh38
rs916157306 Chr.1: 214357146-214357146 on Build GRCh38
rs373881524 Chr.1: 214357450-214357450 on Build GRCh38
rs371730110 Chr.1: 214357226-214357226 on Build GRCh38
rs555042998 Chr.1: 214357367-214357367 on Build GRCh38
rs531172016 Chr.1: 214357177-214357177 on Build GRCh38
rs1054686718 Chr.1: 214357135-214357135 on Build GRCh38
rs916120829 Chr.1: 214357370-214357370 on Build GRCh38
rs777887428 Chr.1: 214357319-214357319 on Build GRCh38
rs1007514359 Chr.1: 214357293-214357293 on Build GRCh38
rs969372970 Chr.1: 214357374-214357374 on Build GRCh38
rs1018839575 Chr.1: 214357549-214357549 on Build GRCh38
rs912381826 Chr.1: 214357190-214357190 on Build GRCh38
rs759008634 Chr.1: 214357428-214357428 on Build GRCh38
rs1007876289 Chr.1: 214357494-214357494 on Build GRCh38
rs897639821 Chr.1: 214357267-214357267 on Build GRCh38
rs907238697 Chr.1: 214357242-214357242 on Build GRCh38
rs1049363972 Chr.1: 214357490-214357490 on Build GRCh38
rs553614012 Chr.1: 214357252-214357252 on Build GRCh38
rs912287877 Chr.1: 214357434-214357434 on Build GRCh38
rs867052544 Chr.1: 214357130-214357130 on Build GRCh38
rs928603704 Chr.1: 214357408-214357408 on Build GRCh38
rs749369686 Chr.1: 214357404-214357404 on Build GRCh38
rs925095848 Chr.1: 214357164-214357164 on Build GRCh38
rs769917859 Chr.1: 214357108-214357108 on Build GRCh38
rs1027537536 Chr.1: 214357286-214357286 on Build GRCh38
rs979154006 Chr.1: 214357385-214357385 on Build GRCh38
rs960209225 Chr.1: 214357344-214357344 on Build GRCh38
rs932381843 Chr.1: 214357180-214357180 on Build GRCh38
rs1019018200 Chr.1: 214357294-214357294 on Build GRCh38
rs930452382 Chr.1: 214357466-214357466 on Build GRCh38
rs965712660 Chr.1: 214357314-214357314 on Build GRCh38
rs979467805 Chr.1: 214357151-214357151 on Build GRCh38
rs888987875 Chr.1: 214357492-214357492 on Build GRCh38
rs943861283 Chr.1: 214357456-214357456 on Build GRCh38

View Full Product Details