Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 218435835-218436338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB2
Entrez Gene ID
7042
Gene Name
transforming growth factor beta 2
Gene Aliases
LDS4, TGF-beta2
UniGene
Hs.133379
Gene Chromosome Location
Chr.1: 218345334-218444619 on Build GRCh38
Transcripts
NM_001135599.2 NM_003238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1267714 c.760G>A p.D254N Chr.1: 218435975-218435975 on Build GRCh38
3977055 c.864C>A p.S288S Chr.1: 218436079-218436079 on Build GRCh38
392415 c.826delG p.K277fs*7 Chr.1: 218436041-218436041 on Build GRCh38
5952067 c.977G>A p.R326Q Chr.1: 218436108-218436108 on Build GRCh38
4613307 c.897_898insA p.N302fs*19 Chr.1: 218436028-218436029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760662920 Chr.1: 218435921-218435921 on Build GRCh38
rs1041531774 Chr.1: 218436261-218436261 on Build GRCh38
rs191509998 Chr.1: 218435997-218435997 on Build GRCh38
rs373352179 Chr.1: 218435988-218435988 on Build GRCh38
rs551456505 Chr.1: 218436224-218436224 on Build GRCh38
rs776212058 Chr.1: 218436083-218436083 on Build GRCh38
rs373646036 Chr.1: 218436079-218436079 on Build GRCh38
rs368738712 Chr.1: 218436169-218436169 on Build GRCh38
rs575861642 Chr.1: 218436102-218436102 on Build GRCh38
rs762561484 Chr.1: 218436108-218436108 on Build GRCh38
rs778961900 Chr.1: 218436025-218436025 on Build GRCh38
rs766726662 Chr.1: 218436156-218436156 on Build GRCh38
rs370630769 Chr.1: 218435959-218435959 on Build GRCh38
rs1022918067 Chr.1: 218436250-218436250 on Build GRCh38
rs139825195 Chr.1: 218436038-218436038 on Build GRCh38
rs747979194 Chr.1: 218436033-218436033 on Build GRCh38
rs758685759 Chr.1: 218436191-218436191 on Build GRCh38
rs546357945 Chr.1: 218435899-218435899 on Build GRCh38
rs779027841 Chr.1: 218436176-218436176 on Build GRCh38
rs748416834 Chr.1: 218436178-218436178 on Build GRCh38
rs756628289 Chr.1: 218436153-218436153 on Build GRCh38
rs368282599 Chr.1: 218436227-218436227 on Build GRCh38
rs863223792 Chr.1: 218436110-218436110 on Build GRCh38
rs187068112 Chr.1: 218435878-218435878 on Build GRCh38
rs370439281 Chr.1: 218436027-218436027 on Build GRCh38
rs767936611 Chr.1: 218436133-218436133 on Build GRCh38
rs979952670 Chr.1: 218435923-218435923 on Build GRCh38
rs754962774 Chr.1: 218436173-218436173 on Build GRCh38
rs947761185 Chr.1: 218436024-218436024 on Build GRCh38
rs955377243 Chr.1: 218435891-218435891 on Build GRCh38
rs781514831 Chr.1: 218435979-218435979 on Build GRCh38
rs11466410 Chr.1: 218435893-218435893 on Build GRCh38
rs945512498 Chr.1: 218436244-218436244 on Build GRCh38
rs376354795 Chr.1: 218436013-218436013 on Build GRCh38
rs745440967 Chr.1: 218436154-218436155 on Build GRCh38
rs559315266 Chr.1: 218436036-218436036 on Build GRCh38
rs764733060 Chr.1: 218435957-218435957 on Build GRCh38
rs1049975705 Chr.1: 218436294-218436294 on Build GRCh38
rs869312903 Chr.1: 218436119-218436119 on Build GRCh38
rs757573768 Chr.1: 218435964-218435964 on Build GRCh38
rs771319501 Chr.1: 218436067-218436067 on Build GRCh38
rs758931687 Chr.1: 218436093-218436093 on Build GRCh38
rs750774144 Chr.1: 218436138-218436138 on Build GRCh38
rs1011074533 Chr.1: 218436233-218436233 on Build GRCh38
rs760758897 Chr.1: 218436047-218436047 on Build GRCh38
rs975429638 Chr.1: 218435877-218435877 on Build GRCh38
rs571211165 Chr.1: 218436259-218436259 on Build GRCh38
rs989462091 Chr.1: 218436132-218436132 on Build GRCh38
rs1052677525 Chr.1: 218436202-218436202 on Build GRCh38
rs759242510 Chr.1: 218435954-218435954 on Build GRCh38
rs749266746 Chr.1: 218436023-218436023 on Build GRCh38
rs970306241 Chr.1: 218435899-218435899 on Build GRCh38
rs550795031 Chr.1: 218436277-218436277 on Build GRCh38
rs888728525 Chr.1: 218436304-218436304 on Build GRCh38
rs563685124 Chr.1: 218435857-218435857 on Build GRCh38
rs10482817 Chr.1: 218436274-218436274 on Build GRCh38
rs781358821 Chr.1: 218435861-218435861 on Build GRCh38
rs372130514 Chr.1: 218435952-218435952 on Build GRCh38
rs777766017 Chr.1: 218436194-218436194 on Build GRCh38
rs764968568 Chr.1: 218436097-218436097 on Build GRCh38
rs775057495 Chr.1: 218436103-218436103 on Build GRCh38
rs766022491 Chr.1: 218435932-218435932 on Build GRCh38
rs778455218 Chr.1: 218435959-218435960 on Build GRCh38
rs886038965 Chr.1: 218436008-218436008 on Build GRCh38
rs863223796 Chr.1: 218436036-218436037 on Build GRCh38
rs587776396 Chr.1: 218436283-218436283 on Build GRCh38

View Full Product Details