Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220102039-220102551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5350049 c.602G>T p.R201L Chr.1: 220102180-220102180 on Build GRCh38
6683456 c.713G>A p.R238Q Chr.1: 220102376-220102376 on Build GRCh38
1338967 c.560T>C p.F187S Chr.1: 220102138-220102138 on Build GRCh38
4953773 c.681T>G p.F227L Chr.1: 220102259-220102259 on Build GRCh38
1210208 c.785C>A p.P262H Chr.1: 220102530-220102530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756393373 Chr.1: 220102395-220102395 on Build GRCh38
rs752799132 Chr.1: 220102142-220102142 on Build GRCh38
rs768101579 Chr.1: 220102179-220102179 on Build GRCh38
rs780849818 Chr.1: 220102153-220102153 on Build GRCh38
rs765198775 Chr.1: 220102108-220102108 on Build GRCh38
rs753047105 Chr.1: 220102108-220102108 on Build GRCh38
rs1042026666 Chr.1: 220102226-220102226 on Build GRCh38
rs201046842 Chr.1: 220102133-220102133 on Build GRCh38
rs762202072 Chr.1: 220102465-220102465 on Build GRCh38
rs753250629 Chr.1: 220102257-220102257 on Build GRCh38
rs757319390 Chr.1: 220102370-220102370 on Build GRCh38
rs771176238 Chr.1: 220102152-220102152 on Build GRCh38
rs751095095 Chr.1: 220102512-220102512 on Build GRCh38
rs754889916 Chr.1: 220102409-220102409 on Build GRCh38
rs904984929 Chr.1: 220102455-220102455 on Build GRCh38
rs571001903 Chr.1: 220102288-220102289 on Build GRCh38
rs1032292776 Chr.1: 220102518-220102518 on Build GRCh38
rs546640845 Chr.1: 220102274-220102274 on Build GRCh38
rs751613884 Chr.1: 220102367-220102367 on Build GRCh38
rs779338044 Chr.1: 220102140-220102140 on Build GRCh38
rs767766570 Chr.1: 220102467-220102467 on Build GRCh38
rs561155210 Chr.1: 220102135-220102135 on Build GRCh38
rs189084759 Chr.1: 220102327-220102327 on Build GRCh38
rs777779272 Chr.1: 220102269-220102269 on Build GRCh38
rs373433981 Chr.1: 220102520-220102520 on Build GRCh38
rs775153600 Chr.1: 220102112-220102112 on Build GRCh38
rs775526438 Chr.1: 220102169-220102169 on Build GRCh38
rs368937943 Chr.1: 220102203-220102203 on Build GRCh38
rs775825237 Chr.1: 220102357-220102357 on Build GRCh38
rs958850547 Chr.1: 220102443-220102443 on Build GRCh38
rs749298044 Chr.1: 220102319-220102319 on Build GRCh38
rs766651661 Chr.1: 220102493-220102493 on Build GRCh38
rs767672750 Chr.1: 220102376-220102376 on Build GRCh38
rs545199797 Chr.1: 220102085-220102085 on Build GRCh38
rs377482718 Chr.1: 220102116-220102116 on Build GRCh38
rs777884187 Chr.1: 220102146-220102146 on Build GRCh38
rs1018589849 Chr.1: 220102092-220102092 on Build GRCh38
rs903078644 Chr.1: 220102236-220102236 on Build GRCh38
rs544502232 Chr.1: 220102121-220102121 on Build GRCh38
rs148149998 Chr.1: 220102148-220102148 on Build GRCh38
rs773738046 Chr.1: 220102332-220102332 on Build GRCh38
rs999995412 Chr.1: 220102079-220102079 on Build GRCh38
rs751212940 Chr.1: 220102114-220102114 on Build GRCh38
rs750528193 Chr.1: 220102478-220102478 on Build GRCh38
rs758922128 Chr.1: 220102267-220102267 on Build GRCh38
rs965681285 Chr.1: 220102108-220102111 on Build GRCh38
rs747645857 Chr.1: 220102420-220102420 on Build GRCh38
rs749790428 Chr.1: 220102298-220102299 on Build GRCh38
rs370222815 Chr.1: 220102438-220102438 on Build GRCh38
rs868785797 Chr.1: 220102375-220102375 on Build GRCh38
rs917287197 Chr.1: 220102190-220102190 on Build GRCh38
rs780062054 Chr.1: 220102403-220102403 on Build GRCh38
rs766712241 Chr.1: 220102126-220102126 on Build GRCh38
rs767135644 Chr.1: 220102188-220102188 on Build GRCh38
rs774960759 Chr.1: 220102463-220102463 on Build GRCh38
rs762954815 Chr.1: 220102177-220102177 on Build GRCh38
rs745688807 Chr.1: 220102313-220102313 on Build GRCh38
rs369155122 Chr.1: 220102364-220102364 on Build GRCh38
rs779751372 Chr.1: 220102132-220102132 on Build GRCh38
rs758434992 Chr.1: 220102145-220102145 on Build GRCh38
rs776470541 Chr.1: 220102105-220102105 on Build GRCh38
rs574253271 Chr.1: 220102119-220102119 on Build GRCh38
rs149353254 Chr.1: 220102216-220102216 on Build GRCh38
rs372618006 Chr.1: 220102429-220102429 on Build GRCh38
rs751587007 Chr.1: 220102271-220102271 on Build GRCh38
rs182226161 Chr.1: 220102352-220102352 on Build GRCh38
rs372276818 Chr.1: 220102317-220102317 on Build GRCh38
rs201539020 Chr.1: 220102436-220102436 on Build GRCh38
rs754766431 Chr.1: 220102498-220102498 on Build GRCh38
rs764539319 Chr.1: 220102449-220102449 on Build GRCh38
rs199601961 Chr.1: 220102404-220102404 on Build GRCh38
rs141996928 Chr.1: 220102214-220102214 on Build GRCh38
rs199575981 Chr.1: 220102412-220102412 on Build GRCh38
rs201063152 Chr.1: 220102091-220102091 on Build GRCh38
rs745600952 Chr.1: 220102158-220102158 on Build GRCh38
rs374990412 Chr.1: 220102358-220102358 on Build GRCh38
rs775841640 Chr.1: 220102431-220102431 on Build GRCh38
rs531152257 Chr.1: 220102095-220102095 on Build GRCh38
rs202128341 Chr.1: 220102341-220102341 on Build GRCh38
rs747221820 Chr.1: 220102093-220102093 on Build GRCh38
rs769873679 Chr.1: 220102165-220102165 on Build GRCh38
rs756410015 Chr.1: 220102490-220102490 on Build GRCh38
rs773899849 Chr.1: 220102180-220102180 on Build GRCh38
rs531862813 Chr.1: 220102095-220102096 on Build GRCh38
rs376610134 Chr.1: 220102318-220102318 on Build GRCh38
rs370306595 Chr.1: 220102452-220102452 on Build GRCh38
rs988091643 Chr.1: 220102232-220102232 on Build GRCh38
rs187251505 Chr.1: 220102355-220102355 on Build GRCh38
rs778544749 Chr.1: 220102507-220102507 on Build GRCh38
rs771648093 Chr.1: 220102348-220102348 on Build GRCh38
rs761439693 Chr.1: 220102187-220102187 on Build GRCh38
rs914016212 Chr.1: 220102242-220102242 on Build GRCh38
rs759609759 Chr.1: 220102107-220102107 on Build GRCh38
rs1054248277 Chr.1: 220102312-220102312 on Build GRCh38
rs781426318 Chr.1: 220102294-220102294 on Build GRCh38
rs373464940 Chr.1: 220102113-220102113 on Build GRCh38
rs772692964 Chr.1: 220102423-220102423 on Build GRCh38
rs374904870 Chr.1: 220102430-220102430 on Build GRCh38
rs951649229 Chr.1: 220102118-220102119 on Build GRCh38
rs753601921 Chr.1: 220102497-220102497 on Build GRCh38
rs750724636 Chr.1: 220102390-220102390 on Build GRCh38

View Full Product Details