Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220786491-220786998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARC1
Entrez Gene ID
64757
Gene Name
mitochondrial amidoxime reducing component 1
Gene Aliases
MOSC1
UniGene
Hs.497816
Gene Chromosome Location
Chr.1: 220786697-220814399 on Build GRCh38
Transcripts
NM_022746.3 XM_011509900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7412895 c.1A>G p.M1V Chr.1: 220786945-220786945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs887507954 Chr.1: 220786705-220786705 on Build GRCh38
rs1050126744 Chr.1: 220786698-220786698 on Build GRCh38
rs3738176 Chr.1: 220786898-220786898 on Build GRCh38
rs771123838 Chr.1: 220786582-220786582 on Build GRCh38
rs72470570 Chr.1: 220786756-220786756 on Build GRCh38
rs990386372 Chr.1: 220786540-220786540 on Build GRCh38
rs556720666 Chr.1: 220786967-220786967 on Build GRCh38
rs955490297 Chr.1: 220786968-220786968 on Build GRCh38
rs527862878 Chr.1: 220786549-220786549 on Build GRCh38
rs1013765992 Chr.1: 220786725-220786725 on Build GRCh38
rs975969262 Chr.1: 220786819-220786819 on Build GRCh38
rs905438821 Chr.1: 220786945-220786945 on Build GRCh38
rs1037015436 Chr.1: 220786641-220786641 on Build GRCh38
rs1016864486 Chr.1: 220786952-220786952 on Build GRCh38
rs1007025475 Chr.1: 220786699-220786699 on Build GRCh38
rs988250785 Chr.1: 220786791-220786791 on Build GRCh38
rs1040319602 Chr.1: 220786636-220786636 on Build GRCh38
rs774600257 Chr.1: 220786661-220786661 on Build GRCh38
rs905479504 Chr.1: 220786744-220786744 on Build GRCh38
rs1034899570 Chr.1: 220786747-220786747 on Build GRCh38
rs981773806 Chr.1: 220786743-220786743 on Build GRCh38
rs1003199900 Chr.1: 220786909-220786909 on Build GRCh38
rs1025192462 Chr.1: 220786742-220786742 on Build GRCh38
rs537057905 Chr.1: 220786946-220786946 on Build GRCh38
rs1011097266 Chr.1: 220786712-220786712 on Build GRCh38
rs1021064084 Chr.1: 220786971-220786971 on Build GRCh38
rs1032723356 Chr.1: 220786955-220786955 on Build GRCh38
rs941743284 Chr.1: 220786860-220786860 on Build GRCh38
rs991066700 Chr.1: 220786536-220786536 on Build GRCh38
rs72470569 Chr.1: 220786721-220786721 on Build GRCh38
rs534496491 Chr.1: 220786831-220786831 on Build GRCh38
rs897967786 Chr.1: 220786892-220786892 on Build GRCh38
rs1051263941 Chr.1: 220786681-220786682 on Build GRCh38
rs1020676150 Chr.1: 220786885-220786885 on Build GRCh38
rs3820357 Chr.1: 220786790-220786790 on Build GRCh38
rs564537351 Chr.1: 220786647-220786647 on Build GRCh38
rs931941970 Chr.1: 220786839-220786839 on Build GRCh38
rs941480611 Chr.1: 220786599-220786599 on Build GRCh38
rs897190262 Chr.1: 220786656-220786656 on Build GRCh38
rs893325091 Chr.1: 220786878-220786878 on Build GRCh38
rs962839279 Chr.1: 220786954-220786954 on Build GRCh38
rs893809635 Chr.1: 220786957-220786957 on Build GRCh38
rs890030763 Chr.1: 220786859-220786859 on Build GRCh38
rs3806326 Chr.1: 220786574-220786574 on Build GRCh38
rs1034722347 Chr.1: 220786918-220786918 on Build GRCh38
rs772086792 Chr.1: 220786711-220786711 on Build GRCh38
rs3806327 Chr.1: 220786518-220786518 on Build GRCh38
rs746571735 Chr.1: 220786926-220786926 on Build GRCh38
rs1036074777 Chr.1: 220786876-220786876 on Build GRCh38
rs1054612282 Chr.1: 220786871-220786871 on Build GRCh38
rs182558189 Chr.1: 220786745-220786745 on Build GRCh38
rs746124450 Chr.1: 220786693-220786693 on Build GRCh38
rs1032382882 Chr.1: 220786776-220786776 on Build GRCh38
rs941831054 Chr.1: 220786707-220786707 on Build GRCh38
rs3820356 Chr.1: 220786795-220786795 on Build GRCh38
rs908914536 Chr.1: 220786858-220786858 on Build GRCh38
rs918690670 Chr.1: 220786833-220786833 on Build GRCh38
rs533384581 Chr.1: 220786676-220786676 on Build GRCh38
rs973200135 Chr.1: 220786796-220786796 on Build GRCh38
rs967223948 Chr.1: 220786811-220786811 on Build GRCh38
rs72470571 Chr.1: 220786846-220786846 on Build GRCh38
rs1051054496 Chr.1: 220786855-220786855 on Build GRCh38
rs1010421324 Chr.1: 220786881-220786881 on Build GRCh38
rs937785608 Chr.1: 220786519-220786520 on Build GRCh38

View Full Product Details