Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224967345-224967855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
UniGene
Hs.133977
Gene Chromosome Location
Chr.1: 224928422-225399294 on Build GRCh38
Transcripts
NM_001145154.1 NM_001373.1 NM_144989.2 XM_006711735.3 XM_011544057.2 XM_011544058.2 XM_011544059.2 XM_011544061.2 XM_011544062.2 XM_011544063.2 XM_011544064.2 XM_011544065.2 XM_011544066.2 XM_011544067.2 XM_011544069.2 XM_011544070.2 XM_011544071.2 XM_011544072.2 XM_017000295.1 XM_017000296.1 XM_017000297.1 XM_017000298.1 XM_017000300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3484201 c.922C>A p.R308R Chr.1: 224967473-224967473 on Build GRCh38
1339183 c.980C>T p.T327I Chr.1: 224967531-224967531 on Build GRCh38
1339179 c.591G>A p.S197S Chr.1: 224967523-224967523 on Build GRCh38
1134816 c.625G>T p.V209F Chr.1: 224967557-224967557 on Build GRCh38
6646256 c.678T>C p.V226V Chr.1: 224967706-224967706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761784985 Chr.1: 224967406-224967406 on Build GRCh38
rs61732752 Chr.1: 224967523-224967523 on Build GRCh38
rs774206519 Chr.1: 224967755-224967755 on Build GRCh38
rs545267177 Chr.1: 224967750-224967750 on Build GRCh38
rs146938681 Chr.1: 224967584-224967584 on Build GRCh38
rs1006079804 Chr.1: 224967452-224967452 on Build GRCh38
rs201898712 Chr.1: 224967412-224967412 on Build GRCh38
rs779684244 Chr.1: 224967522-224967522 on Build GRCh38
rs767306173 Chr.1: 224967598-224967598 on Build GRCh38
rs773242725 Chr.1: 224967388-224967388 on Build GRCh38
rs759642532 Chr.1: 224967484-224967484 on Build GRCh38
rs760377919 Chr.1: 224967637-224967637 on Build GRCh38
rs751410566 Chr.1: 224967504-224967504 on Build GRCh38
rs748151884 Chr.1: 224967759-224967759 on Build GRCh38
rs772500314 Chr.1: 224967384-224967384 on Build GRCh38
rs767106082 Chr.1: 224967512-224967512 on Build GRCh38
rs755642319 Chr.1: 224967424-224967424 on Build GRCh38
rs183978159 Chr.1: 224967793-224967793 on Build GRCh38
rs375791690 Chr.1: 224967434-224967434 on Build GRCh38
rs771446718 Chr.1: 224967549-224967549 on Build GRCh38
rs765785545 Chr.1: 224967642-224967642 on Build GRCh38
rs761024332 Chr.1: 224967392-224967392 on Build GRCh38
rs201578812 Chr.1: 224967678-224967678 on Build GRCh38
rs757688753 Chr.1: 224967673-224967673 on Build GRCh38
rs898224263 Chr.1: 224967800-224967800 on Build GRCh38
rs2501155 Chr.1: 224967412-224967412 on Build GRCh38
rs370187831 Chr.1: 224967473-224967473 on Build GRCh38
rs769996137 Chr.1: 224967485-224967485 on Build GRCh38
rs372706704 Chr.1: 224967575-224967575 on Build GRCh38
rs763771594 Chr.1: 224967502-224967502 on Build GRCh38
rs766553927 Chr.1: 224967398-224967398 on Build GRCh38
rs761626755 Chr.1: 224967508-224967508 on Build GRCh38
rs199899173 Chr.1: 224967415-224967415 on Build GRCh38
rs780170097 Chr.1: 224967724-224967724 on Build GRCh38
rs909854662 Chr.1: 224967801-224967801 on Build GRCh38
rs758671845 Chr.1: 224967409-224967410 on Build GRCh38
rs759519679 Chr.1: 224967404-224967404 on Build GRCh38
rs982194166 Chr.1: 224967679-224967679 on Build GRCh38
rs367971582 Chr.1: 224967592-224967592 on Build GRCh38
rs748695334 Chr.1: 224967430-224967430 on Build GRCh38
rs190977826 Chr.1: 224967722-224967722 on Build GRCh38
rs199814205 Chr.1: 224967542-224967542 on Build GRCh38
rs180670639 Chr.1: 224967369-224967369 on Build GRCh38
rs745439395 Chr.1: 224967409-224967412 on Build GRCh38
rs777678622 Chr.1: 224967413-224967413 on Build GRCh38
rs370146344 Chr.1: 224967594-224967594 on Build GRCh38
rs749895478 Chr.1: 224967423-224967423 on Build GRCh38
rs776903488 Chr.1: 224967403-224967403 on Build GRCh38
rs370749461 Chr.1: 224967560-224967560 on Build GRCh38
rs201411461 Chr.1: 224967409-224967409 on Build GRCh38
rs753432129 Chr.1: 224967647-224967647 on Build GRCh38
rs752129317 Chr.1: 224967665-224967665 on Build GRCh38
rs779231996 Chr.1: 224967426-224967426 on Build GRCh38
rs781509822 Chr.1: 224967557-224967557 on Build GRCh38
rs573158578 Chr.1: 224967422-224967422 on Build GRCh38
rs938020370 Chr.1: 224967731-224967731 on Build GRCh38
rs763000820 Chr.1: 224967498-224967498 on Build GRCh38
rs756396083 Chr.1: 224967709-224967709 on Build GRCh38
rs553805845 Chr.1: 224967417-224967417 on Build GRCh38
rs374449785 Chr.1: 224967655-224967655 on Build GRCh38
rs778259191 Chr.1: 224967439-224967439 on Build GRCh38
rs939860888 Chr.1: 224967761-224967761 on Build GRCh38
rs764660908 Chr.1: 224967405-224967406 on Build GRCh38
rs749317368 Chr.1: 224967413-224967414 on Build GRCh38
rs559085893 Chr.1: 224967518-224967518 on Build GRCh38
rs764014458 Chr.1: 224967413-224967413 on Build GRCh38
rs764616265 Chr.1: 224967662-224967662 on Build GRCh38
rs747179804 Chr.1: 224967458-224967458 on Build GRCh38
rs769978167 Chr.1: 224967564-224967564 on Build GRCh38
rs747341722 Chr.1: 224967544-224967544 on Build GRCh38
rs574098608 Chr.1: 224967572-224967572 on Build GRCh38
rs752706439 Chr.1: 224967408-224967408 on Build GRCh38
rs777972126 Chr.1: 224967536-224967536 on Build GRCh38
rs750088260 Chr.1: 224967516-224967516 on Build GRCh38
rs749337818 Chr.1: 224967732-224967732 on Build GRCh38
rs765229066 Chr.1: 224967406-224967406 on Build GRCh38
rs201807754 Chr.1: 224967474-224967474 on Build GRCh38
rs780926616 Chr.1: 224967411-224967412 on Build GRCh38
rs750073901 Chr.1: 224967631-224967631 on Build GRCh38
rs371407749 Chr.1: 224967701-224967701 on Build GRCh38
rs755953306 Chr.1: 224967412-224967413 on Build GRCh38
rs145350269 Chr.1: 224967409-224967409 on Build GRCh38
rs781777256 Chr.1: 224967824-224967824 on Build GRCh38
rs775282606 Chr.1: 224967489-224967489 on Build GRCh38
rs758994377 Chr.1: 224967532-224967532 on Build GRCh38

View Full Product Details