Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226965538-226966046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ8A
Entrez Gene ID
56997
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9
UniGene
Hs.118241
Gene Chromosome Location
Chr.1: 226939339-226987545 on Build GRCh38
Transcripts
NM_020247.4 XM_005273201.1 XM_011544238.1 XM_011544239.2 XM_011544240.2 XM_011544241.2 XM_017001852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3785171 c.594C>T p.S198S Chr.1: 226965676-226965676 on Build GRCh38
146799 c.605G>A p.R202Q Chr.1: 226965687-226965687 on Build GRCh38
4659831 c.652G>A p.G218R Chr.1: 226965734-226965734 on Build GRCh38
4659830 c.651C>T p.F217F Chr.1: 226965733-226965733 on Build GRCh38
3977119 c.601G>T p.A201S Chr.1: 226965683-226965683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910962267 Chr.1: 226965621-226965621 on Build GRCh38
rs576469992 Chr.1: 226965641-226965641 on Build GRCh38
rs119468005 Chr.1: 226965719-226965719 on Build GRCh38
rs772788042 Chr.1: 226965779-226965779 on Build GRCh38
rs764640143 Chr.1: 226965647-226965647 on Build GRCh38
rs755950454 Chr.1: 226965727-226965727 on Build GRCh38
rs540671891 Chr.1: 226965838-226965838 on Build GRCh38
rs188599627 Chr.1: 226965683-226965683 on Build GRCh38
rs547676292 Chr.1: 226965967-226965967 on Build GRCh38
rs373872875 Chr.1: 226965644-226965644 on Build GRCh38
rs929594082 Chr.1: 226965944-226965944 on Build GRCh38
rs567939383 Chr.1: 226965951-226965951 on Build GRCh38
rs1024443371 Chr.1: 226965791-226965791 on Build GRCh38
rs548298271 Chr.1: 226965836-226965836 on Build GRCh38
rs912033509 Chr.1: 226965830-226965830 on Build GRCh38
rs866489971 Chr.1: 226965677-226965677 on Build GRCh38
rs1033076858 Chr.1: 226966001-226966001 on Build GRCh38
rs146526153 Chr.1: 226965746-226965746 on Build GRCh38
rs530459148 Chr.1: 226965813-226965813 on Build GRCh38
rs72760475 Chr.1: 226965635-226965635 on Build GRCh38
rs771765465 Chr.1: 226965773-226965773 on Build GRCh38
rs750421327 Chr.1: 226965722-226965722 on Build GRCh38
rs952408318 Chr.1: 226965599-226965599 on Build GRCh38
rs552129296 Chr.1: 226965856-226965856 on Build GRCh38
rs370519461 Chr.1: 226965637-226965637 on Build GRCh38
rs774353638 Chr.1: 226965649-226965649 on Build GRCh38
rs202175838 Chr.1: 226965658-226965658 on Build GRCh38
rs1014233735 Chr.1: 226965559-226965559 on Build GRCh38
rs752607267 Chr.1: 226966011-226966011 on Build GRCh38
rs746389612 Chr.1: 226965782-226965782 on Build GRCh38
rs745867840 Chr.1: 226965711-226965712 on Build GRCh38
rs138547552 Chr.1: 226965676-226965676 on Build GRCh38
rs918238399 Chr.1: 226965667-226965667 on Build GRCh38
rs760018980 Chr.1: 226965624-226965624 on Build GRCh38
rs540907139 Chr.1: 226965692-226965692 on Build GRCh38
rs764173526 Chr.1: 226965708-226965708 on Build GRCh38
rs774016800 Chr.1: 226965665-226965665 on Build GRCh38
rs1055661529 Chr.1: 226965958-226965958 on Build GRCh38
rs897986149 Chr.1: 226965803-226965803 on Build GRCh38
rs754776488 Chr.1: 226965749-226965749 on Build GRCh38
rs984303676 Chr.1: 226965603-226965603 on Build GRCh38
rs143981558 Chr.1: 226965653-226965653 on Build GRCh38
rs953561889 Chr.1: 226965796-226965796 on Build GRCh38
rs371870568 Chr.1: 226965723-226965723 on Build GRCh38
rs765094300 Chr.1: 226966010-226966010 on Build GRCh38
rs185346992 Chr.1: 226965560-226965560 on Build GRCh38
rs368074800 Chr.1: 226965671-226965671 on Build GRCh38
rs1040742697 Chr.1: 226965661-226965661 on Build GRCh38
rs75286417 Chr.1: 226965995-226965995 on Build GRCh38
rs75936420 Chr.1: 226965952-226965952 on Build GRCh38
rs767584322 Chr.1: 226965720-226965720 on Build GRCh38
rs747797043 Chr.1: 226965770-226965770 on Build GRCh38
rs781160424 Chr.1: 226965648-226965648 on Build GRCh38
rs918259030 Chr.1: 226965863-226965863 on Build GRCh38
rs903202416 Chr.1: 226965979-226965979 on Build GRCh38
rs963726674 Chr.1: 226965865-226965865 on Build GRCh38
rs200541514 Chr.1: 226965693-226965693 on Build GRCh38
rs765515195 Chr.1: 226965627-226965627 on Build GRCh38
rs768150745 Chr.1: 226965659-226965659 on Build GRCh38
rs1025763613 Chr.1: 226965594-226965594 on Build GRCh38
rs962247940 Chr.1: 226965561-226965561 on Build GRCh38
rs983883944 Chr.1: 226965945-226965945 on Build GRCh38
rs145350362 Chr.1: 226965687-226965687 on Build GRCh38
rs895631249 Chr.1: 226965960-226965960 on Build GRCh38
rs376099510 Chr.1: 226965629-226965629 on Build GRCh38
rs564074965 Chr.1: 226965936-226965936 on Build GRCh38
rs886042265 Chr.1: 226965720-226965727 on Build GRCh38
rs967349187 Chr.1: 226965754-226965754 on Build GRCh38
rs751760028 Chr.1: 226965709-226965709 on Build GRCh38
rs375627978 Chr.1: 226965733-226965733 on Build GRCh38
rs779703398 Chr.1: 226965657-226965657 on Build GRCh38
rs367598725 Chr.1: 226965630-226965630 on Build GRCh38
rs1014575164 Chr.1: 226965721-226965721 on Build GRCh38
rs115855910 Chr.1: 226965700-226965700 on Build GRCh38
rs925862929 Chr.1: 226965968-226965968 on Build GRCh38
rs967518496 Chr.1: 226965618-226965618 on Build GRCh38
rs950036545 Chr.1: 226965964-226965964 on Build GRCh38
rs117785541 Chr.1: 226965573-226965573 on Build GRCh38
rs556883875 Chr.1: 226966003-226966003 on Build GRCh38
rs1033024584 Chr.1: 226965790-226965790 on Build GRCh38
rs767597982 Chr.1: 226965839-226965839 on Build GRCh38
rs778744225 Chr.1: 226965755-226965755 on Build GRCh38

View Full Product Details