Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227657900-227658441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF678
Entrez Gene ID
339500
Gene Name
zinc finger protein 678
Gene Aliases
-
UniGene
Hs.30323
Gene Chromosome Location
Chr.1: 227563519-227677443 on Build GRCh38
Transcripts
NM_178549.3 NR_033184.1 NR_102302.1

SNP Detail

SNP ID Chromosome Location
rs773384319 Chr.1: 227658238-227658238 on Build GRCh38
rs565478456 Chr.1: 227658188-227658188 on Build GRCh38
rs1002839525 Chr.1: 227658134-227658134 on Build GRCh38
rs1007470711 Chr.1: 227658091-227658091 on Build GRCh38
rs564683378 Chr.1: 227658324-227658324 on Build GRCh38
rs532708694 Chr.1: 227658213-227658213 on Build GRCh38
rs959610537 Chr.1: 227657947-227657947 on Build GRCh38
rs573942452 Chr.1: 227658101-227658101 on Build GRCh38
rs1013546780 Chr.1: 227658203-227658204 on Build GRCh38
rs560954860 Chr.1: 227658199-227658199 on Build GRCh38
rs28373918 Chr.1: 227658415-227658415 on Build GRCh38
rs966999361 Chr.1: 227658349-227658349 on Build GRCh38
rs910480858 Chr.1: 227658396-227658396 on Build GRCh38
rs905469024 Chr.1: 227658303-227658303 on Build GRCh38
rs986414929 Chr.1: 227658380-227658380 on Build GRCh38
rs1042408271 Chr.1: 227658002-227658002 on Build GRCh38
rs1040115490 Chr.1: 227658008-227658008 on Build GRCh38
rs913685292 Chr.1: 227658041-227658042 on Build GRCh38
rs575884781 Chr.1: 227657941-227657941 on Build GRCh38
rs925278115 Chr.1: 227658376-227658376 on Build GRCh38
rs755542396 Chr.1: 227658076-227658076 on Build GRCh38
rs566009472 Chr.1: 227658261-227658261 on Build GRCh38
rs770057774 Chr.1: 227658393-227658393 on Build GRCh38
rs548268518 Chr.1: 227658286-227658286 on Build GRCh38
rs978465167 Chr.1: 227658358-227658358 on Build GRCh38
rs1040724794 Chr.1: 227658133-227658133 on Build GRCh38
rs530234503 Chr.1: 227658270-227658270 on Build GRCh38
rs993547879 Chr.1: 227658203-227658203 on Build GRCh38
rs925330598 Chr.1: 227658026-227658026 on Build GRCh38
rs904090015 Chr.1: 227658112-227658112 on Build GRCh38
rs992854043 Chr.1: 227657957-227657957 on Build GRCh38
rs945391836 Chr.1: 227657992-227657992 on Build GRCh38
rs917911763 Chr.1: 227657965-227657965 on Build GRCh38
rs534019200 Chr.1: 227658048-227658048 on Build GRCh38
rs959560042 Chr.1: 227658279-227658279 on Build GRCh38
rs1054250868 Chr.1: 227658051-227658051 on Build GRCh38
rs540324332 Chr.1: 227658312-227658312 on Build GRCh38
rs951757448 Chr.1: 227658242-227658242 on Build GRCh38
rs986080362 Chr.1: 227657924-227657924 on Build GRCh38
rs6699448 Chr.1: 227658251-227658251 on Build GRCh38
rs532025553 Chr.1: 227658132-227658132 on Build GRCh38
rs10916191 Chr.1: 227658083-227658083 on Build GRCh38
rs372853625 Chr.1: 227658135-227658135 on Build GRCh38

View Full Product Details