Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247606291-247606797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2G3
Entrez Gene ID
81469
Gene Name
olfactory receptor family 2 subfamily G member 3
Gene Aliases
OR1-33
UniGene
Hs.653227
Gene Chromosome Location
Chr.1: 247605586-247606515 on Build GRCh38
Transcripts
NM_001001914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
356266 c.852T>C p.T284T Chr.1: 247606437-247606437 on Build GRCh38
2232222 c.920G>A p.G307E Chr.1: 247606505-247606505 on Build GRCh38
6572531 c.859C>T p.P287S Chr.1: 247606444-247606444 on Build GRCh38
6062929 c.849C>A p.P283P Chr.1: 247606434-247606434 on Build GRCh38
3486744 c.864C>T p.I288I Chr.1: 247606449-247606449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183119197 Chr.1: 247606391-247606391 on Build GRCh38
rs780242070 Chr.1: 247606423-247606423 on Build GRCh38
rs745920770 Chr.1: 247606403-247606403 on Build GRCh38
rs754327639 Chr.1: 247606389-247606389 on Build GRCh38
rs765206162 Chr.1: 247606345-247606345 on Build GRCh38
rs759301157 Chr.1: 247606377-247606377 on Build GRCh38
rs749294919 Chr.1: 247606449-247606449 on Build GRCh38
rs897803116 Chr.1: 247606475-247606475 on Build GRCh38
rs530857838 Chr.1: 247606352-247606352 on Build GRCh38
rs766155773 Chr.1: 247606430-247606430 on Build GRCh38
rs747216053 Chr.1: 247606524-247606524 on Build GRCh38
rs773974525 Chr.1: 247606375-247606375 on Build GRCh38
rs758416394 Chr.1: 247606515-247606515 on Build GRCh38
rs12072304 Chr.1: 247606357-247606357 on Build GRCh38
rs201693543 Chr.1: 247606525-247606525 on Build GRCh38
rs750394134 Chr.1: 247606402-247606402 on Build GRCh38
rs927773146 Chr.1: 247606559-247606559 on Build GRCh38
rs757826660 Chr.1: 247606503-247606503 on Build GRCh38
rs751087492 Chr.1: 247606564-247606564 on Build GRCh38
rs778068648 Chr.1: 247606447-247606447 on Build GRCh38
rs141464415 Chr.1: 247606466-247606466 on Build GRCh38
rs1000492414 Chr.1: 247606615-247606615 on Build GRCh38
rs904497493 Chr.1: 247606754-247606754 on Build GRCh38
rs1021603239 Chr.1: 247606441-247606441 on Build GRCh38
rs61737569 Chr.1: 247606484-247606484 on Build GRCh38
rs909615108 Chr.1: 247606325-247606325 on Build GRCh38
rs753165616 Chr.1: 247606380-247606380 on Build GRCh38
rs368638155 Chr.1: 247606543-247606543 on Build GRCh38
rs751599975 Chr.1: 247606348-247606348 on Build GRCh38
rs747951872 Chr.1: 247606432-247606432 on Build GRCh38
rs147001668 Chr.1: 247606674-247606674 on Build GRCh38
rs558907945 Chr.1: 247606641-247606641 on Build GRCh38
rs369337684 Chr.1: 247606528-247606528 on Build GRCh38
rs774368936 Chr.1: 247606564-247606564 on Build GRCh38
rs138441168 Chr.1: 247606390-247606390 on Build GRCh38
rs939731048 Chr.1: 247606332-247606332 on Build GRCh38
rs534661893 Chr.1: 247606383-247606383 on Build GRCh38
rs201707034 Chr.1: 247606557-247606557 on Build GRCh38
rs199624559 Chr.1: 247606434-247606434 on Build GRCh38
rs570832403 Chr.1: 247606367-247606367 on Build GRCh38
rs764510563 Chr.1: 247606385-247606385 on Build GRCh38
rs745372725 Chr.1: 247606565-247606565 on Build GRCh38
rs946612426 Chr.1: 247606719-247606719 on Build GRCh38
rs867097876 Chr.1: 247606728-247606728 on Build GRCh38
rs772264488 Chr.1: 247606322-247606322 on Build GRCh38
rs1031605901 Chr.1: 247606655-247606655 on Build GRCh38
rs200370417 Chr.1: 247606324-247606324 on Build GRCh38
rs1028980569 Chr.1: 247606471-247606471 on Build GRCh38
rs764473365 Chr.1: 247606497-247606497 on Build GRCh38
rs1039258550 Chr.1: 247606560-247606560 on Build GRCh38
rs758687288 Chr.1: 247606405-247606405 on Build GRCh38
rs139607108 Chr.1: 247606473-247606473 on Build GRCh38
rs780260355 Chr.1: 247606522-247606522 on Build GRCh38
rs1054994089 Chr.1: 247606581-247606581 on Build GRCh38
rs965927825 Chr.1: 247606461-247606461 on Build GRCh38
rs779468160 Chr.1: 247606363-247606363 on Build GRCh38
rs748665104 Chr.1: 247606379-247606379 on Build GRCh38
rs371711041 Chr.1: 247606474-247606474 on Build GRCh38
rs759099777 Chr.1: 247606470-247606470 on Build GRCh38
rs267598476 Chr.1: 247606505-247606505 on Build GRCh38
rs765812919 Chr.1: 247606497-247606498 on Build GRCh38
rs904551280 Chr.1: 247606444-247606444 on Build GRCh38
rs895034882 Chr.1: 247606582-247606582 on Build GRCh38
rs939172125 Chr.1: 247606575-247606575 on Build GRCh38
rs61730407 Chr.1: 247606450-247606450 on Build GRCh38
rs754133704 Chr.1: 247606502-247606502 on Build GRCh38
rs769678031 Chr.1: 247606356-247606356 on Build GRCh38
rs750559921 Chr.1: 247606514-247606514 on Build GRCh38
rs567924164 Chr.1: 247606534-247606534 on Build GRCh38
rs773044341 Chr.1: 247606435-247606435 on Build GRCh38
rs1007680285 Chr.1: 247606553-247606553 on Build GRCh38

View Full Product Details