Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5723546-5724047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208B
Entrez Gene ID
54906
Gene Name
family with sequence similarity 208 member B
Gene Aliases
C10orf18, bA318E3.2
UniGene
Hs.610717
Gene Chromosome Location
Chr.10: 5684759-5763740 on Build GRCh38
Transcripts
NM_001321783.1 NM_001321784.1 NM_001321785.1 NM_017782.4 XM_005252474.3 XM_005252475.3 XM_005252476.4 XM_005252480.4 XM_005252481.4 XM_005252482.2 XM_011519515.2 XM_011519516.2 XM_011519517.2 XM_011519518.2 XM_011519519.2 XM_011519520.2 XM_011519521.2 XM_011519522.2 XM_011519525.2 XM_017016361.1 XM_017016362.1 XM_017016363.1 XM_017016364.1 XM_017016365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2147851 c.208G>C p.E70Q Chr.10: 5723738-5723738 on Build GRCh38
4682886 c.149C>T p.P50L Chr.10: 5723679-5723679 on Build GRCh38
5475448 c.150G>A p.P50P Chr.10: 5723680-5723680 on Build GRCh38
6238011 c.192G>A p.L64L Chr.10: 5723722-5723722 on Build GRCh38
7426238 c.152A>T p.Q51L Chr.10: 5723682-5723682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031753854 Chr.10: 5723694-5723694 on Build GRCh38
rs568117164 Chr.10: 5723818-5723818 on Build GRCh38
rs779590002 Chr.10: 5723708-5723708 on Build GRCh38
rs780439015 Chr.10: 5723822-5723822 on Build GRCh38
rs778715512 Chr.10: 5723725-5723725 on Build GRCh38
rs201741308 Chr.10: 5723705-5723705 on Build GRCh38
rs762944149 Chr.10: 5723762-5723762 on Build GRCh38
rs756042299 Chr.10: 5723707-5723707 on Build GRCh38
rs369161573 Chr.10: 5723734-5723734 on Build GRCh38
rs767265278 Chr.10: 5723793-5723793 on Build GRCh38
rs775635450 Chr.10: 5723755-5723755 on Build GRCh38
rs907377906 Chr.10: 5723982-5723982 on Build GRCh38
rs371521388 Chr.10: 5723788-5723788 on Build GRCh38
rs765095923 Chr.10: 5723864-5723864 on Build GRCh38
rs762489504 Chr.10: 5723599-5723599 on Build GRCh38
rs775474495 Chr.10: 5723635-5723635 on Build GRCh38
rs754678014 Chr.10: 5723719-5723719 on Build GRCh38
rs770261799 Chr.10: 5723745-5723745 on Build GRCh38
rs3793708 Chr.10: 5723674-5723674 on Build GRCh38
rs912388336 Chr.10: 5723803-5723803 on Build GRCh38
rs543812996 Chr.10: 5723913-5723913 on Build GRCh38
rs979272899 Chr.10: 5723701-5723701 on Build GRCh38
rs781751729 Chr.10: 5723737-5723737 on Build GRCh38
rs749316226 Chr.10: 5723828-5723828 on Build GRCh38
rs768807570 Chr.10: 5723765-5723765 on Build GRCh38
rs528482966 Chr.10: 5723584-5723584 on Build GRCh38
rs1037860624 Chr.10: 5723872-5723872 on Build GRCh38
rs756575834 Chr.10: 5723820-5723820 on Build GRCh38
rs750977562 Chr.10: 5723896-5723896 on Build GRCh38
rs940421803 Chr.10: 5723853-5723853 on Build GRCh38
rs767390308 Chr.10: 5723684-5723684 on Build GRCh38
rs374967651 Chr.10: 5723800-5723800 on Build GRCh38
rs931085331 Chr.10: 5723890-5723890 on Build GRCh38
rs746284188 Chr.10: 5723819-5723819 on Build GRCh38
rs907554100 Chr.10: 5723846-5723846 on Build GRCh38
rs566881443 Chr.10: 5723992-5723992 on Build GRCh38
rs750230967 Chr.10: 5723797-5723797 on Build GRCh38
rs971566793 Chr.10: 5723638-5723638 on Build GRCh38
rs540133947 Chr.10: 5723957-5723957 on Build GRCh38
rs569490520 Chr.10: 5723953-5723953 on Build GRCh38
rs760650370 Chr.10: 5723799-5723799 on Build GRCh38
rs764366254 Chr.10: 5723666-5723666 on Build GRCh38
rs375654540 Chr.10: 5723680-5723680 on Build GRCh38
rs774575780 Chr.10: 5723768-5723768 on Build GRCh38
rs998938337 Chr.10: 5723984-5723984 on Build GRCh38
rs967790691 Chr.10: 5723999-5723999 on Build GRCh38
rs1045996522 Chr.10: 5723955-5723955 on Build GRCh38
rs1036780428 Chr.10: 5723881-5723881 on Build GRCh38
rs1033551648 Chr.10: 5724016-5724016 on Build GRCh38
rs138841985 Chr.10: 5723964-5723964 on Build GRCh38
rs747323205 Chr.10: 5723728-5723728 on Build GRCh38
rs528034281 Chr.10: 5723716-5723716 on Build GRCh38
rs916536451 Chr.10: 5723805-5723805 on Build GRCh38
rs1000246419 Chr.10: 5724002-5724002 on Build GRCh38
rs758167039 Chr.10: 5723802-5723802 on Build GRCh38
rs939871676 Chr.10: 5723838-5723838 on Build GRCh38
rs529490538 Chr.10: 5723861-5723861 on Build GRCh38
rs778320998 Chr.10: 5723718-5723718 on Build GRCh38
rs1050627895 Chr.10: 5723903-5723903 on Build GRCh38
rs1031845018 Chr.10: 5724001-5724001 on Build GRCh38
rs1020260061 Chr.10: 5723988-5723988 on Build GRCh38
rs763223390 Chr.10: 5723663-5723663 on Build GRCh38
rs368840515 Chr.10: 5723640-5723640 on Build GRCh38
rs752429574 Chr.10: 5723801-5723801 on Build GRCh38
rs530701927 Chr.10: 5723633-5723633 on Build GRCh38
rs1055172003 Chr.10: 5723907-5723907 on Build GRCh38
rs557460873 Chr.10: 5723938-5723938 on Build GRCh38
rs919631012 Chr.10: 5723882-5723882 on Build GRCh38
rs746355058 Chr.10: 5723742-5723742 on Build GRCh38
rs267602530 Chr.10: 5723743-5723743 on Build GRCh38

View Full Product Details