Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13229529-13230025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCMA
Entrez Gene ID
221044
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
C10orf49, GRP, GRP/UCMA
UniGene
Hs.112998
Gene Chromosome Location
Chr.10: 13221766-13234328 on Build GRCh38
Transcripts
NM_001303118.1 NM_001303119.1 NM_145314.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1346830 c.316G>A p.D106N Chr.10: 13229614-13229614 on Build GRCh38
916234 c.231G>A p.R77R Chr.10: 13229699-13229699 on Build GRCh38
916233 c.262G>T p.E88* Chr.10: 13229668-13229668 on Build GRCh38
130056 c.292G>A p.E98K Chr.10: 13229638-13229638 on Build GRCh38
286269 c.300C>T p.F100F Chr.10: 13229630-13229630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026107095 Chr.10: 13229931-13229931 on Build GRCh38
rs756597511 Chr.10: 13229578-13229578 on Build GRCh38
rs753915346 Chr.10: 13229600-13229600 on Build GRCh38
rs981630300 Chr.10: 13229620-13229620 on Build GRCh38
rs779030660 Chr.10: 13229578-13229578 on Build GRCh38
rs4750320 Chr.10: 13229834-13229834 on Build GRCh38
rs1046824385 Chr.10: 13229865-13229865 on Build GRCh38
rs533463595 Chr.10: 13229887-13229887 on Build GRCh38
rs768203649 Chr.10: 13229660-13229660 on Build GRCh38
rs570369602 Chr.10: 13229755-13229755 on Build GRCh38
rs372587250 Chr.10: 13229705-13229705 on Build GRCh38
rs200520685 Chr.10: 13229733-13229733 on Build GRCh38
rs765199412 Chr.10: 13229670-13229670 on Build GRCh38
rs763688616 Chr.10: 13229988-13229989 on Build GRCh38
rs745773066 Chr.10: 13229707-13229707 on Build GRCh38
rs974727103 Chr.10: 13229615-13229615 on Build GRCh38
rs550765394 Chr.10: 13229767-13229767 on Build GRCh38
rs1030296623 Chr.10: 13230000-13230000 on Build GRCh38
rs767473230 Chr.10: 13229775-13229775 on Build GRCh38
rs568165579 Chr.10: 13229650-13229650 on Build GRCh38
rs190112667 Chr.10: 13229986-13229986 on Build GRCh38
rs554487224 Chr.10: 13229781-13229781 on Build GRCh38
rs963070982 Chr.10: 13229909-13229909 on Build GRCh38
rs765426204 Chr.10: 13229761-13229761 on Build GRCh38
rs914811557 Chr.10: 13230002-13230002 on Build GRCh38
rs566442943 Chr.10: 13229987-13229987 on Build GRCh38
rs1035876191 Chr.10: 13229951-13229951 on Build GRCh38
rs1056755294 Chr.10: 13229555-13229555 on Build GRCh38
rs551694134 Chr.10: 13229942-13229942 on Build GRCh38
rs184841299 Chr.10: 13229627-13229627 on Build GRCh38
rs148425191 Chr.10: 13229673-13229673 on Build GRCh38
rs761048350 Chr.10: 13229742-13229742 on Build GRCh38
rs748913414 Chr.10: 13229640-13229640 on Build GRCh38
rs764998260 Chr.10: 13229570-13229570 on Build GRCh38
rs369577986 Chr.10: 13229606-13229606 on Build GRCh38
rs546560592 Chr.10: 13229588-13229588 on Build GRCh38
rs758546425 Chr.10: 13229674-13229674 on Build GRCh38
rs774961083 Chr.10: 13229571-13229571 on Build GRCh38
rs776653974 Chr.10: 13229626-13229626 on Build GRCh38
rs376615420 Chr.10: 13229836-13229836 on Build GRCh38
rs777985262 Chr.10: 13229677-13229677 on Build GRCh38
rs751413925 Chr.10: 13229688-13229688 on Build GRCh38
rs964328595 Chr.10: 13229612-13229612 on Build GRCh38
rs778363402 Chr.10: 13229753-13229766 on Build GRCh38
rs760958464 Chr.10: 13229645-13229645 on Build GRCh38
rs760151703 Chr.10: 13229614-13229614 on Build GRCh38
rs573989280 Chr.10: 13229809-13229809 on Build GRCh38
rs1005852154 Chr.10: 13229585-13229585 on Build GRCh38
rs1016170554 Chr.10: 13229902-13229902 on Build GRCh38
rs80070283 Chr.10: 13229628-13229628 on Build GRCh38
rs576230237 Chr.10: 13229887-13229887 on Build GRCh38
rs762494319 Chr.10: 13229630-13229630 on Build GRCh38
rs773840629 Chr.10: 13229738-13229738 on Build GRCh38
rs752094093 Chr.10: 13229559-13229559 on Build GRCh38
rs750984703 Chr.10: 13229574-13229574 on Build GRCh38
rs776276988 Chr.10: 13229562-13229562 on Build GRCh38
rs1051615345 Chr.10: 13229829-13229829 on Build GRCh38
rs759050653 Chr.10: 13229567-13229567 on Build GRCh38
rs759937696 Chr.10: 13229760-13229760 on Build GRCh38
rs758349686 Chr.10: 13229679-13229679 on Build GRCh38
rs748229662 Chr.10: 13229621-13229621 on Build GRCh38
rs74123515 Chr.10: 13229629-13229629 on Build GRCh38
rs758252025 Chr.10: 13229610-13229610 on Build GRCh38
rs535673436 Chr.10: 13229981-13229981 on Build GRCh38
rs769398936 Chr.10: 13229717-13229717 on Build GRCh38
rs757073452 Chr.10: 13229689-13229689 on Build GRCh38
rs200425647 Chr.10: 13229572-13229572 on Build GRCh38
rs895563026 Chr.10: 13229558-13229558 on Build GRCh38
rs749032485 Chr.10: 13229728-13229728 on Build GRCh38
rs770624904 Chr.10: 13229560-13229560 on Build GRCh38
rs754294170 Chr.10: 13229659-13229659 on Build GRCh38
rs766509091 Chr.10: 13229598-13229598 on Build GRCh38
rs777418655 Chr.10: 13229613-13229613 on Build GRCh38
rs754992807 Chr.10: 13229602-13229602 on Build GRCh38
rs767269906 Chr.10: 13229584-13229584 on Build GRCh38
rs760430685 Chr.10: 13229830-13229830 on Build GRCh38
rs113913020 Chr.10: 13229937-13229937 on Build GRCh38
rs375579693 Chr.10: 13229724-13229724 on Build GRCh38
rs888253199 Chr.10: 13229914-13229914 on Build GRCh38
rs767980356 Chr.10: 13229633-13229633 on Build GRCh38

View Full Product Details