Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13336152-13336665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPHS1
Entrez Gene ID
22929
Gene Name
selenophosphate synthetase 1
Gene Aliases
SELD, SPS, SPS1
UniGene
Hs.124027
Gene Chromosome Location
Chr.10: 13317428-13348298 on Build GRCh38
Transcripts
NM_001195602.1 NM_001195604.1 NM_012247.4 XM_017015942.1 XM_017015943.1 XM_017015944.1 XM_017015945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4633135 c.356G>A p.C119Y Chr.10: 13336292-13336292 on Build GRCh38
4945069 c.340A>T p.M114L Chr.10: 13336308-13336308 on Build GRCh38
6128631 c.400G>A p.D134N Chr.10: 13336248-13336248 on Build GRCh38
916298 c.397A>G p.T133A Chr.10: 13336251-13336251 on Build GRCh38
4641343 c.399C>T p.T133T Chr.10: 13336249-13336249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919008763 Chr.10: 13336436-13336436 on Build GRCh38
rs937507145 Chr.10: 13336182-13336182 on Build GRCh38
rs1056347271 Chr.10: 13336522-13336523 on Build GRCh38
rs145348335 Chr.10: 13336327-13336327 on Build GRCh38
rs937621443 Chr.10: 13336474-13336474 on Build GRCh38
rs752102842 Chr.10: 13336340-13336340 on Build GRCh38
rs750506188 Chr.10: 13336444-13336444 on Build GRCh38
rs372858494 Chr.10: 13336308-13336308 on Build GRCh38
rs547315070 Chr.10: 13336451-13336451 on Build GRCh38
rs542605530 Chr.10: 13336334-13336334 on Build GRCh38
rs764552609 Chr.10: 13336419-13336419 on Build GRCh38
rs180691350 Chr.10: 13336226-13336226 on Build GRCh38
rs772642840 Chr.10: 13336441-13336441 on Build GRCh38
rs771939588 Chr.10: 13336391-13336391 on Build GRCh38
rs535579223 Chr.10: 13336589-13336589 on Build GRCh38
rs375133467 Chr.10: 13336297-13336297 on Build GRCh38
rs887842291 Chr.10: 13336598-13336598 on Build GRCh38
rs1039194995 Chr.10: 13336623-13336623 on Build GRCh38
rs1041766887 Chr.10: 13336592-13336592 on Build GRCh38
rs755438016 Chr.10: 13336217-13336217 on Build GRCh38
rs200835015 Chr.10: 13336298-13336298 on Build GRCh38
rs566951044 Chr.10: 13336533-13336533 on Build GRCh38
rs754055126 Chr.10: 13336466-13336466 on Build GRCh38
rs185739439 Chr.10: 13336403-13336403 on Build GRCh38
rs750463579 Chr.10: 13336380-13336380 on Build GRCh38
rs767206232 Chr.10: 13336202-13336202 on Build GRCh38
rs769996817 Chr.10: 13336259-13336259 on Build GRCh38
rs551718302 Chr.10: 13336409-13336409 on Build GRCh38
rs758852573 Chr.10: 13336225-13336225 on Build GRCh38
rs756318787 Chr.10: 13336381-13336381 on Build GRCh38
rs928882458 Chr.10: 13336468-13336468 on Build GRCh38
rs372937968 Chr.10: 13336384-13336384 on Build GRCh38
rs754539389 Chr.10: 13336180-13336180 on Build GRCh38
rs767676668 Chr.10: 13336200-13336200 on Build GRCh38
rs62641683 Chr.10: 13336273-13336273 on Build GRCh38
rs947553509 Chr.10: 13336586-13336586 on Build GRCh38
rs370560910 Chr.10: 13336209-13336209 on Build GRCh38
rs766566806 Chr.10: 13336456-13336456 on Build GRCh38
rs748082771 Chr.10: 13336390-13336390 on Build GRCh38
rs772621833 Chr.10: 13336400-13336400 on Build GRCh38
rs903309427 Chr.10: 13336628-13336628 on Build GRCh38
rs374556965 Chr.10: 13336224-13336224 on Build GRCh38
rs565047976 Chr.10: 13336420-13336420 on Build GRCh38
rs768619457 Chr.10: 13336285-13336285 on Build GRCh38
rs1007676009 Chr.10: 13336616-13336616 on Build GRCh38
rs902049121 Chr.10: 13336635-13336635 on Build GRCh38
rs377241729 Chr.10: 13336309-13336309 on Build GRCh38
rs751017583 Chr.10: 13336408-13336411 on Build GRCh38
rs372138605 Chr.10: 13336315-13336315 on Build GRCh38
rs763473341 Chr.10: 13336410-13336410 on Build GRCh38
rs372297316 Chr.10: 13336249-13336249 on Build GRCh38
rs2275128 Chr.10: 13336445-13336445 on Build GRCh38
rs780290435 Chr.10: 13336383-13336383 on Build GRCh38
rs3215961 Chr.10: 13336543-13336544 on Build GRCh38
rs369311909 Chr.10: 13336367-13336367 on Build GRCh38
rs762427620 Chr.10: 13336440-13336440 on Build GRCh38
rs62641684 Chr.10: 13336300-13336300 on Build GRCh38
rs763967212 Chr.10: 13336199-13336199 on Build GRCh38
rs751731346 Chr.10: 13336229-13336229 on Build GRCh38
rs761279323 Chr.10: 13336201-13336201 on Build GRCh38
rs746078177 Chr.10: 13336255-13336255 on Build GRCh38
rs2275129 Chr.10: 13336181-13336181 on Build GRCh38
rs770532029 Chr.10: 13336404-13336404 on Build GRCh38
rs774247442 Chr.10: 13336288-13336288 on Build GRCh38
rs201414633 Chr.10: 13336339-13336339 on Build GRCh38
rs377429516 Chr.10: 13336385-13336385 on Build GRCh38
rs757482094 Chr.10: 13336358-13336358 on Build GRCh38
rs773881714 Chr.10: 13336200-13336200 on Build GRCh38
rs773953901 Chr.10: 13336271-13336271 on Build GRCh38
rs970326493 Chr.10: 13336437-13336437 on Build GRCh38
rs1049109070 Chr.10: 13336543-13336543 on Build GRCh38
rs776938591 Chr.10: 13336470-13336470 on Build GRCh38

View Full Product Details