Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13646766-13647270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
UniGene
Hs.330463
Gene Chromosome Location
Chr.10: 13643706-14330952 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

SNP Detail

SNP ID Chromosome Location
rs940421910 Chr.10: 13646819-13646819 on Build GRCh38
rs369304389 Chr.10: 13646927-13646927 on Build GRCh38
rs1023507830 Chr.10: 13646875-13646875 on Build GRCh38
rs956936759 Chr.10: 13646924-13646924 on Build GRCh38
rs770288461 Chr.10: 13647213-13647213 on Build GRCh38
rs565476689 Chr.10: 13646852-13646852 on Build GRCh38
rs903909332 Chr.10: 13646844-13646844 on Build GRCh38
rs1053082215 Chr.10: 13646792-13646792 on Build GRCh38
rs548094990 Chr.10: 13646966-13646966 on Build GRCh38
rs1035368585 Chr.10: 13647114-13647114 on Build GRCh38
rs1036538651 Chr.10: 13646821-13646821 on Build GRCh38
rs900718378 Chr.10: 13647171-13647171 on Build GRCh38
rs954509469 Chr.10: 13647236-13647236 on Build GRCh38
rs954437174 Chr.10: 13646981-13646981 on Build GRCh38
rs961685421 Chr.10: 13647235-13647235 on Build GRCh38
rs921528925 Chr.10: 13646801-13646801 on Build GRCh38
rs746561513 Chr.10: 13647150-13647150 on Build GRCh38
rs930367878 Chr.10: 13647238-13647238 on Build GRCh38
rs1028418174 Chr.10: 13647185-13647185 on Build GRCh38
rs968084575 Chr.10: 13647164-13647164 on Build GRCh38
rs745764083 Chr.10: 13647072-13647074 on Build GRCh38
rs573750498 Chr.10: 13646972-13646972 on Build GRCh38
rs1016402349 Chr.10: 13646957-13646957 on Build GRCh38
rs987470723 Chr.10: 13647012-13647012 on Build GRCh38
rs543381891 Chr.10: 13647021-13647021 on Build GRCh38
rs1016349895 Chr.10: 13647157-13647157 on Build GRCh38
rs898959090 Chr.10: 13646845-13646845 on Build GRCh38
rs182847039 Chr.10: 13646802-13646802 on Build GRCh38
rs1055091451 Chr.10: 13646902-13646902 on Build GRCh38
rs1020377414 Chr.10: 13647240-13647240 on Build GRCh38
rs938073256 Chr.10: 13647049-13647049 on Build GRCh38
rs955745223 Chr.10: 13647212-13647212 on Build GRCh38
rs761756368 Chr.10: 13646926-13646926 on Build GRCh38
rs890144983 Chr.10: 13646814-13646817 on Build GRCh38
rs975600261 Chr.10: 13646799-13646799 on Build GRCh38
rs896523040 Chr.10: 13646903-13646903 on Build GRCh38
rs768076546 Chr.10: 13646939-13646939 on Build GRCh38
rs56161216 Chr.10: 13646798-13646798 on Build GRCh38
rs72767564 Chr.10: 13647081-13647081 on Build GRCh38
rs531317063 Chr.10: 13647024-13647024 on Build GRCh38
rs532781466 Chr.10: 13646938-13646938 on Build GRCh38
rs117362358 Chr.10: 13647011-13647011 on Build GRCh38
rs1026595579 Chr.10: 13646925-13646925 on Build GRCh38
rs1042928368 Chr.10: 13646829-13646829 on Build GRCh38
rs975548034 Chr.10: 13646973-13646973 on Build GRCh38
rs1035015154 Chr.10: 13646912-13646912 on Build GRCh38
rs978557169 Chr.10: 13647018-13647018 on Build GRCh38
rs1029950911 Chr.10: 13646969-13646969 on Build GRCh38
rs369154530 Chr.10: 13647172-13647172 on Build GRCh38
rs74121341 Chr.10: 13647117-13647117 on Build GRCh38
rs140032412 Chr.10: 13647066-13647069 on Build GRCh38
rs540783886 Chr.10: 13646810-13646810 on Build GRCh38
rs566637901 Chr.10: 13647237-13647237 on Build GRCh38
rs921453819 Chr.10: 13646975-13646975 on Build GRCh38
rs751680808 Chr.10: 13646911-13646911 on Build GRCh38
rs115657776 Chr.10: 13646968-13646968 on Build GRCh38
rs947304151 Chr.10: 13647014-13647014 on Build GRCh38
rs989121993 Chr.10: 13646989-13646989 on Build GRCh38
rs75206552 Chr.10: 13647073-13647076 on Build GRCh38
rs1048212797 Chr.10: 13647110-13647110 on Build GRCh38
rs59208412 Chr.10: 13647084-13647084 on Build GRCh38
rs146945206 Chr.10: 13647100-13647100 on Build GRCh38
rs1006438311 Chr.10: 13647149-13647149 on Build GRCh38
rs908794653 Chr.10: 13647233-13647233 on Build GRCh38
rs898906901 Chr.10: 13647052-13647052 on Build GRCh38
rs1030067477 Chr.10: 13647215-13647215 on Build GRCh38
rs1014244028 Chr.10: 13646910-13646910 on Build GRCh38

View Full Product Details