Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17951721-17952230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A12
Entrez Gene ID
221074
Gene Name
solute carrier family 39 member 12
Gene Aliases
LZT-Hs8, ZIP-12, bA570F3.1
UniGene
Hs.350895
Gene Chromosome Location
Chr.10: 17951839-18043292 on Build GRCh38
Transcripts
NM_001145195.1 NM_001282733.1 NM_001282734.1 NM_152725.3

SNP Detail

SNP ID Chromosome Location
rs935053719 Chr.10: 17951768-17951768 on Build GRCh38
rs1048228155 Chr.10: 17951885-17951885 on Build GRCh38
rs782521300 Chr.10: 17952172-17952172 on Build GRCh38
rs895802465 Chr.10: 17951927-17951927 on Build GRCh38
rs979003607 Chr.10: 17952051-17952051 on Build GRCh38
rs1048344977 Chr.10: 17952187-17952187 on Build GRCh38
rs553203836 Chr.10: 17952138-17952138 on Build GRCh38
rs943749743 Chr.10: 17951832-17951832 on Build GRCh38
rs568878809 Chr.10: 17951893-17951893 on Build GRCh38
rs41277350 Chr.10: 17952083-17952083 on Build GRCh38
rs74642760 Chr.10: 17952196-17952196 on Build GRCh38
rs181338759 Chr.10: 17952053-17952053 on Build GRCh38
rs557016053 Chr.10: 17952197-17952197 on Build GRCh38
rs1023161041 Chr.10: 17951955-17951955 on Build GRCh38
rs375445471 Chr.10: 17951968-17951968 on Build GRCh38
rs924797568 Chr.10: 17951750-17951750 on Build GRCh38
rs912182116 Chr.10: 17951803-17951803 on Build GRCh38
rs561363525 Chr.10: 17951756-17951756 on Build GRCh38
rs899771908 Chr.10: 17951841-17951841 on Build GRCh38
rs1040022037 Chr.10: 17951833-17951833 on Build GRCh38
rs1035786743 Chr.10: 17951909-17951909 on Build GRCh38
rs575461372 Chr.10: 17952179-17952179 on Build GRCh38
rs535785128 Chr.10: 17951897-17951897 on Build GRCh38
rs201867364 Chr.10: 17951769-17951770 on Build GRCh38
rs987912737 Chr.10: 17951787-17951787 on Build GRCh38
rs931119298 Chr.10: 17951861-17951861 on Build GRCh38
rs781844615 Chr.10: 17952159-17952159 on Build GRCh38
rs988285650 Chr.10: 17952108-17952108 on Build GRCh38
rs943682470 Chr.10: 17952131-17952131 on Build GRCh38
rs886908008 Chr.10: 17952188-17952188 on Build GRCh38
rs78199097 Chr.10: 17951950-17951950 on Build GRCh38
rs782750795 Chr.10: 17952130-17952130 on Build GRCh38

View Full Product Details