Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17981030-17981530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A12
Entrez Gene ID
221074
Gene Name
solute carrier family 39 member 12
Gene Aliases
LZT-Hs8, ZIP-12, bA570F3.1
UniGene
Hs.350895
Gene Chromosome Location
Chr.10: 17951839-18043292 on Build GRCh38
Transcripts
NM_001145195.1 NM_001282733.1 NM_001282734.1 NM_152725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4013164 c.942G>T p.R314S Chr.10: 17981329-17981329 on Build GRCh38
7221806 c.925-6A>G p.? Chr.10: 17981306-17981306 on Build GRCh38
7227712 c.969G>C p.K323N Chr.10: 17981356-17981356 on Build GRCh38
7390539 c.941G>T p.R314M Chr.10: 17981328-17981328 on Build GRCh38
683695 c.946C>G p.L316V Chr.10: 17981333-17981333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773987169 Chr.10: 17981373-17981373 on Build GRCh38
rs1030624891 Chr.10: 17981309-17981309 on Build GRCh38
rs554483342 Chr.10: 17981186-17981186 on Build GRCh38
rs72778328 Chr.10: 17981412-17981412 on Build GRCh38
rs911484927 Chr.10: 17981057-17981057 on Build GRCh38
rs372763135 Chr.10: 17981288-17981288 on Build GRCh38
rs745889465 Chr.10: 17981324-17981324 on Build GRCh38
rs937201705 Chr.10: 17981143-17981143 on Build GRCh38
rs1056002617 Chr.10: 17981154-17981154 on Build GRCh38
rs146367461 Chr.10: 17981360-17981360 on Build GRCh38
rs7086141 Chr.10: 17981117-17981117 on Build GRCh38
rs772721594 Chr.10: 17981393-17981393 on Build GRCh38
rs572724088 Chr.10: 17981194-17981194 on Build GRCh38
rs772807530 Chr.10: 17981482-17981482 on Build GRCh38
rs1032446692 Chr.10: 17981199-17981199 on Build GRCh38
rs267602432 Chr.10: 17981447-17981447 on Build GRCh38
rs918289091 Chr.10: 17981104-17981104 on Build GRCh38
rs894616856 Chr.10: 17981242-17981242 on Build GRCh38
rs761592286 Chr.10: 17981378-17981378 on Build GRCh38
rs986705350 Chr.10: 17981342-17981342 on Build GRCh38
rs959130447 Chr.10: 17981061-17981061 on Build GRCh38
rs552305565 Chr.10: 17981468-17981468 on Build GRCh38
rs143932694 Chr.10: 17981166-17981166 on Build GRCh38
rs762746559 Chr.10: 17981263-17981263 on Build GRCh38
rs531399248 Chr.10: 17981477-17981477 on Build GRCh38
rs188148584 Chr.10: 17981178-17981178 on Build GRCh38
rs139060671 Chr.10: 17981078-17981078 on Build GRCh38
rs763576102 Chr.10: 17981494-17981494 on Build GRCh38
rs781708942 Chr.10: 17981298-17981298 on Build GRCh38
rs557980170 Chr.10: 17981222-17981222 on Build GRCh38
rs768546774 Chr.10: 17981371-17981371 on Build GRCh38
rs540758607 Chr.10: 17981069-17981069 on Build GRCh38
rs781482975 Chr.10: 17981313-17981313 on Build GRCh38
rs942516961 Chr.10: 17981384-17981384 on Build GRCh38
rs774006961 Chr.10: 17981264-17981264 on Build GRCh38
rs62618669 Chr.10: 17981381-17981381 on Build GRCh38
rs769863215 Chr.10: 17981335-17981335 on Build GRCh38
rs965528268 Chr.10: 17981248-17981248 on Build GRCh38
rs11011848 Chr.10: 17981168-17981168 on Build GRCh38
rs757568527 Chr.10: 17981433-17981433 on Build GRCh38
rs746276419 Chr.10: 17981484-17981485 on Build GRCh38
rs139456412 Chr.10: 17981474-17981474 on Build GRCh38
rs985647687 Chr.10: 17981220-17981220 on Build GRCh38
rs771785439 Chr.10: 17981198-17981199 on Build GRCh38
rs2437261 Chr.10: 17981211-17981211 on Build GRCh38
rs749928511 Chr.10: 17981282-17981282 on Build GRCh38
rs760110052 Chr.10: 17981403-17981403 on Build GRCh38
rs775528646 Chr.10: 17981338-17981338 on Build GRCh38
rs911034061 Chr.10: 17981140-17981140 on Build GRCh38
rs779975343 Chr.10: 17981467-17981467 on Build GRCh38
rs763343718 Chr.10: 17981413-17981413 on Build GRCh38
rs202200743 Chr.10: 17981287-17981287 on Build GRCh38
rs781501644 Chr.10: 17981448-17981448 on Build GRCh38
rs987122376 Chr.10: 17981138-17981138 on Build GRCh38
rs751962888 Chr.10: 17981501-17981501 on Build GRCh38
rs760195895 Chr.10: 17981489-17981489 on Build GRCh38
rs917117319 Chr.10: 17981386-17981386 on Build GRCh38
rs568089017 Chr.10: 17981498-17981498 on Build GRCh38
rs368238099 Chr.10: 17981303-17981303 on Build GRCh38
rs372124071 Chr.10: 17981473-17981473 on Build GRCh38
rs777877665 Chr.10: 17981295-17981295 on Build GRCh38
rs1047086428 Chr.10: 17981192-17981192 on Build GRCh38
rs967694601 Chr.10: 17981095-17981095 on Build GRCh38
rs144056997 Chr.10: 17981405-17981405 on Build GRCh38
rs753004716 Chr.10: 17981290-17981290 on Build GRCh38
rs79375232 Chr.10: 17981224-17981224 on Build GRCh38
rs143757655 Chr.10: 17981475-17981475 on Build GRCh38
rs909642711 Chr.10: 17981291-17981291 on Build GRCh38
rs747301784 Chr.10: 17981302-17981302 on Build GRCh38
rs768260965 Chr.10: 17981212-17981212 on Build GRCh38
rs182567016 Chr.10: 17981107-17981107 on Build GRCh38
rs1926739 Chr.10: 17981493-17981493 on Build GRCh38
rs750566813 Chr.10: 17981453-17981453 on Build GRCh38
rs764453387 Chr.10: 17981496-17981496 on Build GRCh38
rs200334409 Chr.10: 17981292-17981292 on Build GRCh38
rs977029859 Chr.10: 17981251-17981251 on Build GRCh38
rs559004101 Chr.10: 17981123-17981123 on Build GRCh38
rs200804085 Chr.10: 17981422-17981422 on Build GRCh38
rs145853200 Chr.10: 17981280-17981280 on Build GRCh38
rs942540899 Chr.10: 17981153-17981153 on Build GRCh38
rs11011850 Chr.10: 17981279-17981279 on Build GRCh38
rs79322533 Chr.10: 17981429-17981429 on Build GRCh38
rs756215469 Chr.10: 17981454-17981454 on Build GRCh38
rs950001967 Chr.10: 17981409-17981409 on Build GRCh38

View Full Product Details