Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 25021789-25022286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
UniGene
Hs.534486
Gene Chromosome Location
Chr.10: 24981979-25062279 on Build GRCh38
Transcripts
NM_001270383.1 NM_024838.4 XM_005252597.2 XM_017016665.1

Gene Symbol
THNSL1
Entrez Gene ID
79896
Gene Name
threonine synthase like 1
Gene Aliases
TSH1
UniGene
Hs.645274
Gene Chromosome Location
Chr.10: 24952319-25026664 on Build GRCh38
Transcripts
NM_001270383.1 NM_024838.4 XM_005252597.2 XM_017016665.1

SNP Detail

SNP ID Chromosome Location
rs975504126 Chr.10: 25022202-25022202 on Build GRCh38
rs560382829 Chr.10: 25021860-25021860 on Build GRCh38
rs907068363 Chr.10: 25022088-25022088 on Build GRCh38
rs775844758 Chr.10: 25021809-25021809 on Build GRCh38
rs1036066341 Chr.10: 25022204-25022204 on Build GRCh38
rs769154966 Chr.10: 25022074-25022074 on Build GRCh38
rs915370471 Chr.10: 25022096-25022096 on Build GRCh38
rs7089295 Chr.10: 25021821-25021821 on Build GRCh38
rs1002787302 Chr.10: 25022095-25022095 on Build GRCh38
rs761236037 Chr.10: 25022026-25022026 on Build GRCh38
rs1051328185 Chr.10: 25021883-25021883 on Build GRCh38
rs963162462 Chr.10: 25021867-25021867 on Build GRCh38
rs939618094 Chr.10: 25022151-25022151 on Build GRCh38
rs776922714 Chr.10: 25022097-25022097 on Build GRCh38
rs546719165 Chr.10: 25021847-25021847 on Build GRCh38
rs898222669 Chr.10: 25022055-25022055 on Build GRCh38
rs895546781 Chr.10: 25021835-25021835 on Build GRCh38
rs532782995 Chr.10: 25022044-25022044 on Build GRCh38
rs955562713 Chr.10: 25022224-25022224 on Build GRCh38
rs760687733 Chr.10: 25022139-25022139 on Build GRCh38
rs895473228 Chr.10: 25022180-25022180 on Build GRCh38
rs953424751 Chr.10: 25021980-25021980 on Build GRCh38
rs753843764 Chr.10: 25022144-25022144 on Build GRCh38
rs538557404 Chr.10: 25022126-25022126 on Build GRCh38
rs908185922 Chr.10: 25021892-25021892 on Build GRCh38
rs150162748 Chr.10: 25022101-25022101 on Build GRCh38
rs1038460323 Chr.10: 25022046-25022046 on Build GRCh38
rs137903766 Chr.10: 25021842-25021842 on Build GRCh38
rs1014655705 Chr.10: 25021844-25021844 on Build GRCh38
rs1004528468 Chr.10: 25022183-25022183 on Build GRCh38
rs1045649416 Chr.10: 25022135-25022135 on Build GRCh38
rs762144166 Chr.10: 25022121-25022121 on Build GRCh38
rs1028901293 Chr.10: 25021910-25021910 on Build GRCh38
rs968126440 Chr.10: 25022133-25022133 on Build GRCh38
rs1009541750 Chr.10: 25022102-25022102 on Build GRCh38
rs763955005 Chr.10: 25022142-25022142 on Build GRCh38
rs774015110 Chr.10: 25022259-25022259 on Build GRCh38
rs1013080801 Chr.10: 25022062-25022062 on Build GRCh38
rs1056771694 Chr.10: 25022161-25022161 on Build GRCh38
rs1028446185 Chr.10: 25022223-25022223 on Build GRCh38
rs1001175766 Chr.10: 25021813-25021813 on Build GRCh38
rs765987596 Chr.10: 25022141-25022141 on Build GRCh38
rs1044215446 Chr.10: 25022071-25022071 on Build GRCh38
rs552508979 Chr.10: 25022085-25022085 on Build GRCh38

View Full Product Details