Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 25175257-25175743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR158
Entrez Gene ID
57512
Gene Name
G protein-coupled receptor 158
Gene Aliases
-
UniGene
Hs.499108
Gene Chromosome Location
Chr.10: 25174722-25602229 on Build GRCh38
Transcripts
NM_020752.2 NR_027333.1

Gene Symbol
GPR158-AS1
Entrez Gene ID
100128811
Gene Name
GPR158 antisense RNA 1
Gene Aliases
-
UniGene
Hs.700407
Gene Chromosome Location
Chr.10: 25158072-25176276 on Build GRCh38
Transcripts
NM_020752.2 NR_027333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050116 c.91C>T p.R31W Chr.10: 25175511-25175511 on Build GRCh38
4510851 c.84C>T p.P28P Chr.10: 25175504-25175504 on Build GRCh38
5554844 c.53G>A p.G18E Chr.10: 25175473-25175473 on Build GRCh38
5836706 c.296G>A p.C99Y Chr.10: 25175716-25175716 on Build GRCh38
3978399 c.290C>G p.A97G Chr.10: 25175710-25175710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760492345 Chr.10: 25175663-25175663 on Build GRCh38
rs143696360 Chr.10: 25175618-25175618 on Build GRCh38
rs374227433 Chr.10: 25175434-25175434 on Build GRCh38
rs781083815 Chr.10: 25175383-25175383 on Build GRCh38
rs542701721 Chr.10: 25175279-25175279 on Build GRCh38
rs200242847 Chr.10: 25175724-25175724 on Build GRCh38
rs771952291 Chr.10: 25175465-25175465 on Build GRCh38
rs200216703 Chr.10: 25175615-25175615 on Build GRCh38
rs780402048 Chr.10: 25175666-25175666 on Build GRCh38
rs764541621 Chr.10: 25175539-25175539 on Build GRCh38
rs183511748 Chr.10: 25175386-25175386 on Build GRCh38
rs764746465 Chr.10: 25175397-25175397 on Build GRCh38
rs751208093 Chr.10: 25175547-25175547 on Build GRCh38
rs781623149 Chr.10: 25175676-25175676 on Build GRCh38
rs781030971 Chr.10: 25175433-25175433 on Build GRCh38
rs530580826 Chr.10: 25175459-25175459 on Build GRCh38
rs777538063 Chr.10: 25175575-25175575 on Build GRCh38
rs566727690 Chr.10: 25175569-25175569 on Build GRCh38
rs149308611 Chr.10: 25175722-25175722 on Build GRCh38
rs771077755 Chr.10: 25175387-25175387 on Build GRCh38
rs754374604 Chr.10: 25175378-25175379 on Build GRCh38
rs536224494 Chr.10: 25175527-25175527 on Build GRCh38
rs960038101 Chr.10: 25175370-25175370 on Build GRCh38
rs1057413193 Chr.10: 25175590-25175590 on Build GRCh38
rs772501752 Chr.10: 25175521-25175521 on Build GRCh38
rs913336834 Chr.10: 25175490-25175490 on Build GRCh38
rs761801989 Chr.10: 25175720-25175720 on Build GRCh38
rs745977042 Chr.10: 25175662-25175662 on Build GRCh38
rs769628744 Chr.10: 25175446-25175446 on Build GRCh38
rs769112602 Chr.10: 25175608-25175608 on Build GRCh38
rs749924978 Chr.10: 25175399-25175399 on Build GRCh38
rs758593687 Chr.10: 25175664-25175664 on Build GRCh38
rs367581451 Chr.10: 25175607-25175607 on Build GRCh38
rs558281200 Chr.10: 25175641-25175641 on Build GRCh38
rs889649274 Chr.10: 25175721-25175721 on Build GRCh38
rs371327078 Chr.10: 25175388-25175388 on Build GRCh38
rs758031096 Chr.10: 25175400-25175400 on Build GRCh38
rs147366412 Chr.10: 25175667-25175667 on Build GRCh38
rs775003956 Chr.10: 25175706-25175706 on Build GRCh38
rs754754633 Chr.10: 25175416-25175416 on Build GRCh38
rs540015058 Chr.10: 25175380-25175381 on Build GRCh38
rs371970423 Chr.10: 25175536-25175536 on Build GRCh38
rs1036672664 Chr.10: 25175655-25175655 on Build GRCh38
rs759135219 Chr.10: 25175623-25175623 on Build GRCh38
rs1035660425 Chr.10: 25175365-25175365 on Build GRCh38
rs139582068 Chr.10: 25175680-25175680 on Build GRCh38
rs779087604 Chr.10: 25175659-25175659 on Build GRCh38
rs751259202 Chr.10: 25175409-25175409 on Build GRCh38
rs755851703 Chr.10: 25175646-25175646 on Build GRCh38
rs368107656 Chr.10: 25175526-25175526 on Build GRCh38
rs757283006 Chr.10: 25175586-25175586 on Build GRCh38
rs1051983417 Chr.10: 25175440-25175440 on Build GRCh38
rs776012582 Chr.10: 25175531-25175531 on Build GRCh38
rs752430885 Chr.10: 25175642-25175642 on Build GRCh38
rs567297305 Chr.10: 25175505-25175505 on Build GRCh38
rs374922131 Chr.10: 25175585-25175585 on Build GRCh38
rs765929787 Chr.10: 25175546-25175546 on Build GRCh38
rs796948411 Chr.10: 25175388-25175388 on Build GRCh38
rs752506942 Chr.10: 25175567-25175567 on Build GRCh38
rs1015375374 Chr.10: 25175378-25175378 on Build GRCh38
rs745351671 Chr.10: 25175350-25175350 on Build GRCh38
rs749355440 Chr.10: 25175491-25175491 on Build GRCh38
rs981954943 Chr.10: 25175359-25175359 on Build GRCh38
rs78985567 Chr.10: 25175598-25175598 on Build GRCh38
rs146045468 Chr.10: 25175339-25175342 on Build GRCh38
rs574076490 Chr.10: 25175707-25175707 on Build GRCh38
rs757277921 Chr.10: 25175658-25175658 on Build GRCh38
rs775959503 Chr.10: 25175605-25175605 on Build GRCh38
rs779142792 Chr.10: 25175591-25175591 on Build GRCh38
rs768478165 Chr.10: 25175382-25175382 on Build GRCh38
rs773806437 Chr.10: 25175684-25175684 on Build GRCh38
rs748545032 Chr.10: 25175678-25175678 on Build GRCh38
rs779541090 Chr.10: 25175403-25175403 on Build GRCh38
rs773861324 Chr.10: 25175620-25175620 on Build GRCh38
rs146938051 Chr.10: 25175593-25175593 on Build GRCh38
rs762420887 Chr.10: 25175316-25175316 on Build GRCh38
rs374397908 Chr.10: 25175342-25175345 on Build GRCh38
rs559973565 Chr.10: 25175330-25175330 on Build GRCh38
rs762347005 Chr.10: 25175541-25175541 on Build GRCh38
rs771616703 Chr.10: 25175689-25175689 on Build GRCh38
rs746163727 Chr.10: 25175516-25175516 on Build GRCh38
rs753693801 Chr.10: 25175714-25175714 on Build GRCh38
rs764018775 Chr.10: 25175651-25175651 on Build GRCh38
rs777237072 Chr.10: 25175540-25175540 on Build GRCh38
rs761092136 Chr.10: 25175396-25175396 on Build GRCh38
rs748141703 Chr.10: 25175385-25175385 on Build GRCh38
rs369134778 Chr.10: 25175388-25175389 on Build GRCh38
rs868534288 Chr.10: 25175507-25175507 on Build GRCh38
rs937554187 Chr.10: 25175327-25175327 on Build GRCh38
rs746839641 Chr.10: 25175381-25175381 on Build GRCh38
rs55717617 Chr.10: 25175381-25175381 on Build GRCh38
rs557423753 Chr.10: 25175687-25175687 on Build GRCh38
rs754629829 Chr.10: 25175566-25175566 on Build GRCh38
rs550687410 Chr.10: 25175499-25175499 on Build GRCh38

View Full Product Details