Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27403076-27403612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTCHD3
Entrez Gene ID
374308
Gene Name
patched domain containing 3
Gene Aliases
PTR
UniGene
Hs.631832
Gene Chromosome Location
Chr.10: 27398188-27414368 on Build GRCh38
Transcripts
NM_001034842.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702198 c.1216G>A p.D406N Chr.10: 27403353-27403353 on Build GRCh38
2134573 c.1223T>C p.I408T Chr.10: 27403346-27403346 on Build GRCh38
917442 c.1226G>A p.R409Q Chr.10: 27403343-27403343 on Build GRCh38
7386021 c.1253G>T p.G418V Chr.10: 27403316-27403316 on Build GRCh38
4767834 c.1293C>T p.G431G Chr.10: 27403276-27403276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902987161 Chr.10: 27403125-27403125 on Build GRCh38
rs113139577 Chr.10: 27403517-27403518 on Build GRCh38
rs934503118 Chr.10: 27403351-27403351 on Build GRCh38
rs1053682130 Chr.10: 27403157-27403157 on Build GRCh38
rs183634324 Chr.10: 27403431-27403431 on Build GRCh38
rs1030718742 Chr.10: 27403499-27403499 on Build GRCh38
rs1026057642 Chr.10: 27403534-27403534 on Build GRCh38
rs887094127 Chr.10: 27403182-27403182 on Build GRCh38
rs11015742 Chr.10: 27403276-27403276 on Build GRCh38
rs970489273 Chr.10: 27403278-27403278 on Build GRCh38
rs1002549800 Chr.10: 27403502-27403502 on Build GRCh38
rs761359143 Chr.10: 27403197-27403197 on Build GRCh38
rs759874751 Chr.10: 27403324-27403324 on Build GRCh38
rs369696037 Chr.10: 27403313-27403313 on Build GRCh38
rs779924934 Chr.10: 27403238-27403238 on Build GRCh38
rs751391359 Chr.10: 27403242-27403242 on Build GRCh38
rs748222703 Chr.10: 27403165-27403165 on Build GRCh38
rs910134524 Chr.10: 27403241-27403241 on Build GRCh38
rs991637515 Chr.10: 27403262-27403262 on Build GRCh38
rs757200215 Chr.10: 27403335-27403335 on Build GRCh38
rs75114068 Chr.10: 27403517-27403517 on Build GRCh38
rs760103669 Chr.10: 27403191-27403191 on Build GRCh38
rs2505307 Chr.10: 27403474-27403474 on Build GRCh38
rs546332239 Chr.10: 27403153-27403153 on Build GRCh38
rs543806018 Chr.10: 27403503-27403503 on Build GRCh38
rs767999633 Chr.10: 27403195-27403195 on Build GRCh38
rs1003314090 Chr.10: 27403495-27403495 on Build GRCh38
rs2484180 Chr.10: 27403350-27403350 on Build GRCh38
rs769876247 Chr.10: 27403285-27403285 on Build GRCh38
rs901301990 Chr.10: 27403178-27403178 on Build GRCh38
rs756161488 Chr.10: 27403386-27403386 on Build GRCh38
rs574520591 Chr.10: 27403530-27403530 on Build GRCh38
rs769962110 Chr.10: 27403176-27403176 on Build GRCh38
rs534508762 Chr.10: 27403576-27403576 on Build GRCh38
rs2484181 Chr.10: 27403442-27403442 on Build GRCh38
rs11599425 Chr.10: 27403378-27403378 on Build GRCh38
rs755046985 Chr.10: 27403544-27403544 on Build GRCh38
rs747571467 Chr.10: 27403325-27403325 on Build GRCh38
rs373340793 Chr.10: 27403196-27403196 on Build GRCh38
rs149028689 Chr.10: 27403181-27403181 on Build GRCh38
rs777841338 Chr.10: 27403389-27403389 on Build GRCh38
rs761253736 Chr.10: 27403339-27403339 on Build GRCh38
rs374082153 Chr.10: 27403321-27403321 on Build GRCh38
rs143009394 Chr.10: 27403464-27403464 on Build GRCh38
rs762730271 Chr.10: 27403344-27403344 on Build GRCh38
rs934103737 Chr.10: 27403338-27403338 on Build GRCh38
rs942366933 Chr.10: 27403341-27403341 on Build GRCh38
rs916292399 Chr.10: 27403127-27403127 on Build GRCh38
rs113945146 Chr.10: 27403180-27403180 on Build GRCh38
rs949040445 Chr.10: 27403130-27403130 on Build GRCh38
rs764831245 Chr.10: 27403201-27403201 on Build GRCh38
rs950563021 Chr.10: 27403549-27403549 on Build GRCh38
rs989197465 Chr.10: 27403580-27403580 on Build GRCh38
rs148140519 Chr.10: 27403327-27403327 on Build GRCh38
rs771191726 Chr.10: 27403409-27403409 on Build GRCh38
rs201314603 Chr.10: 27403378-27403379 on Build GRCh38
rs923584082 Chr.10: 27403146-27403146 on Build GRCh38
rs34233861 Chr.10: 27403518-27403519 on Build GRCh38
rs749315662 Chr.10: 27403390-27403390 on Build GRCh38
rs766716738 Chr.10: 27403184-27403184 on Build GRCh38
rs989808934 Chr.10: 27403304-27403304 on Build GRCh38
rs749976887 Chr.10: 27403159-27403159 on Build GRCh38
rs750048210 Chr.10: 27403206-27403206 on Build GRCh38
rs563514987 Chr.10: 27403578-27403578 on Build GRCh38
rs188661198 Chr.10: 27403103-27403103 on Build GRCh38
rs377105186 Chr.10: 27403284-27403284 on Build GRCh38
rs868369984 Chr.10: 27403381-27403381 on Build GRCh38
rs542585753 Chr.10: 27403143-27403143 on Build GRCh38
rs201736385 Chr.10: 27403360-27403360 on Build GRCh38
rs934960693 Chr.10: 27403292-27403292 on Build GRCh38
rs2429491 Chr.10: 27403133-27403133 on Build GRCh38
rs941641087 Chr.10: 27403250-27403250 on Build GRCh38
rs758145163 Chr.10: 27403231-27403231 on Build GRCh38
rs764750380 Chr.10: 27403343-27403343 on Build GRCh38
rs141883836 Chr.10: 27403257-27403257 on Build GRCh38
rs1054734818 Chr.10: 27403371-27403371 on Build GRCh38
rs775281362 Chr.10: 27403530-27403531 on Build GRCh38
rs566496335 Chr.10: 27403160-27403160 on Build GRCh38
rs78873159 Chr.10: 27403518-27403518 on Build GRCh38
rs2484179 Chr.10: 27403170-27403170 on Build GRCh38
rs771138476 Chr.10: 27403318-27403318 on Build GRCh38
rs1012581969 Chr.10: 27403154-27403154 on Build GRCh38
rs200010341 Chr.10: 27403380-27403380 on Build GRCh38

View Full Product Details