Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35607785-35608279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD4
Entrez Gene ID
219770
Gene Name
gap junction protein delta 4
Gene Aliases
CX40.1
UniGene
Hs.638922
Gene Chromosome Location
Chr.10: 35605410-35608935 on Build GRCh38
Transcripts
NM_153368.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1347697 c.510G>A p.P170P Chr.10: 35608023-35608023 on Build GRCh38
4144661 c.516G>A p.K172K Chr.10: 35608029-35608029 on Build GRCh38
4752742 c.200A>G p.E67G Chr.10: 35607953-35607953 on Build GRCh38
6671294 c.249G>A p.A83A Chr.10: 35607904-35607904 on Build GRCh38
6671295 c.662G>A p.R221Q Chr.10: 35608175-35608175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200373511 Chr.10: 35608198-35608198 on Build GRCh38
rs544122382 Chr.10: 35607963-35607963 on Build GRCh38
rs991560017 Chr.10: 35608201-35608201 on Build GRCh38
rs761174554 Chr.10: 35608184-35608184 on Build GRCh38
rs751391222 Chr.10: 35608032-35608032 on Build GRCh38
rs201813002 Chr.10: 35607871-35607871 on Build GRCh38
rs762037125 Chr.10: 35607906-35607906 on Build GRCh38
rs371962831 Chr.10: 35607996-35607996 on Build GRCh38
rs867452720 Chr.10: 35608227-35608227 on Build GRCh38
rs770953239 Chr.10: 35607933-35607933 on Build GRCh38
rs752575669 Chr.10: 35608099-35608099 on Build GRCh38
rs772440122 Chr.10: 35608065-35608065 on Build GRCh38
rs371862112 Chr.10: 35608040-35608040 on Build GRCh38
rs748547746 Chr.10: 35608212-35608212 on Build GRCh38
rs775642510 Chr.10: 35607849-35607849 on Build GRCh38
rs535238831 Chr.10: 35607887-35607887 on Build GRCh38
rs529933444 Chr.10: 35608190-35608190 on Build GRCh38
rs772326760 Chr.10: 35607848-35607848 on Build GRCh38
rs868391411 Chr.10: 35607942-35607942 on Build GRCh38
rs992228596 Chr.10: 35608150-35608150 on Build GRCh38
rs199590260 Chr.10: 35608206-35608206 on Build GRCh38
rs771414305 Chr.10: 35608055-35608055 on Build GRCh38
rs141850840 Chr.10: 35607852-35607852 on Build GRCh38
rs775689336 Chr.10: 35608143-35608143 on Build GRCh38
rs111304561 Chr.10: 35608232-35608232 on Build GRCh38
rs770618498 Chr.10: 35608154-35608154 on Build GRCh38
rs921800386 Chr.10: 35607886-35607886 on Build GRCh38
rs780621371 Chr.10: 35608181-35608181 on Build GRCh38
rs376029347 Chr.10: 35607951-35607951 on Build GRCh38
rs772578703 Chr.10: 35608086-35608086 on Build GRCh38
rs775266410 Chr.10: 35608239-35608239 on Build GRCh38
rs595652 Chr.10: 35608029-35608029 on Build GRCh38
rs757334208 Chr.10: 35608126-35608126 on Build GRCh38
rs776358447 Chr.10: 35608247-35608247 on Build GRCh38
rs767586928 Chr.10: 35608159-35608159 on Build GRCh38
rs756177143 Chr.10: 35608044-35608044 on Build GRCh38
rs761330756 Chr.10: 35608018-35608018 on Build GRCh38
rs778713646 Chr.10: 35608225-35608225 on Build GRCh38
rs760523540 Chr.10: 35608171-35608171 on Build GRCh38
rs368902279 Chr.10: 35608075-35608075 on Build GRCh38
rs956833628 Chr.10: 35608237-35608237 on Build GRCh38
rs202076131 Chr.10: 35607922-35607922 on Build GRCh38
rs774403435 Chr.10: 35608155-35608155 on Build GRCh38
rs772989493 Chr.10: 35608146-35608146 on Build GRCh38
rs867282957 Chr.10: 35608248-35608248 on Build GRCh38
rs138948612 Chr.10: 35607975-35607975 on Build GRCh38
rs777232499 Chr.10: 35607855-35607855 on Build GRCh38
rs769774388 Chr.10: 35607986-35607986 on Build GRCh38
rs555177601 Chr.10: 35608234-35608234 on Build GRCh38
rs747267315 Chr.10: 35608184-35608184 on Build GRCh38
rs750782675 Chr.10: 35607858-35607858 on Build GRCh38
rs748991317 Chr.10: 35608189-35608189 on Build GRCh38
rs747005535 Chr.10: 35608243-35608243 on Build GRCh38
rs749323803 Chr.10: 35608124-35608124 on Build GRCh38
rs200962125 Chr.10: 35607908-35607908 on Build GRCh38
rs577209079 Chr.10: 35607971-35607971 on Build GRCh38
rs766949119 Chr.10: 35608201-35608201 on Build GRCh38
rs780762161 Chr.10: 35607820-35607820 on Build GRCh38
rs751137150 Chr.10: 35607811-35607811 on Build GRCh38
rs771507371 Chr.10: 35608002-35608002 on Build GRCh38
rs558730235 Chr.10: 35607834-35607834 on Build GRCh38
rs776825225 Chr.10: 35607905-35607905 on Build GRCh38
rs747649153 Chr.10: 35607825-35607825 on Build GRCh38
rs764677868 Chr.10: 35608019-35608019 on Build GRCh38
rs747259722 Chr.10: 35607898-35607898 on Build GRCh38
rs896249537 Chr.10: 35607919-35607919 on Build GRCh38
rs778114104 Chr.10: 35608121-35608121 on Build GRCh38
rs754402244 Chr.10: 35607807-35607807 on Build GRCh38
rs764218355 Chr.10: 35608174-35608174 on Build GRCh38
rs535200408 Chr.10: 35608223-35608223 on Build GRCh38
rs762445530 Chr.10: 35607856-35607856 on Build GRCh38
rs146377807 Chr.10: 35608013-35608013 on Build GRCh38
rs538629092 Chr.10: 35608199-35608199 on Build GRCh38
rs773951943 Chr.10: 35608194-35608194 on Build GRCh38
rs748959114 Chr.10: 35608132-35608132 on Build GRCh38
rs776271764 Chr.10: 35608017-35608017 on Build GRCh38
rs867654897 Chr.10: 35608153-35608153 on Build GRCh38
rs35613011 Chr.10: 35607964-35607964 on Build GRCh38
rs746165291 Chr.10: 35608057-35608057 on Build GRCh38
rs777681433 Chr.10: 35608046-35608046 on Build GRCh38
rs759952364 Chr.10: 35608009-35608009 on Build GRCh38
rs763316477 Chr.10: 35607914-35607914 on Build GRCh38
rs563952204 Chr.10: 35607880-35607880 on Build GRCh38
rs751624521 Chr.10: 35608261-35608261 on Build GRCh38
rs778773861 Chr.10: 35607885-35607885 on Build GRCh38
rs749432779 Chr.10: 35608054-35608054 on Build GRCh38
rs148077650 Chr.10: 35607835-35607835 on Build GRCh38
rs759132395 Chr.10: 35608196-35608196 on Build GRCh38
rs368672506 Chr.10: 35607961-35607961 on Build GRCh38
rs372321157 Chr.10: 35608120-35608120 on Build GRCh38
rs753333160 Chr.10: 35607953-35607953 on Build GRCh38
rs759509399 Chr.10: 35608156-35608156 on Build GRCh38
rs143962024 Chr.10: 35608077-35608077 on Build GRCh38
rs752304477 Chr.10: 35607924-35607924 on Build GRCh38
rs983527286 Chr.10: 35607868-35607868 on Build GRCh38
rs746398260 Chr.10: 35608136-35608136 on Build GRCh38
rs779002375 Chr.10: 35608177-35608177 on Build GRCh38
rs147032111 Chr.10: 35607829-35607829 on Build GRCh38
rs768982773 Chr.10: 35607980-35607980 on Build GRCh38
rs374732161 Chr.10: 35607890-35607890 on Build GRCh38
rs200332566 Chr.10: 35607912-35607912 on Build GRCh38
rs752161045 Chr.10: 35608209-35608209 on Build GRCh38
rs112658635 Chr.10: 35608217-35608217 on Build GRCh38
rs757304072 Chr.10: 35608175-35608175 on Build GRCh38
rs145646764 Chr.10: 35608089-35608089 on Build GRCh38
rs756784586 Chr.10: 35607954-35607954 on Build GRCh38
rs749244709 Chr.10: 35607830-35607830 on Build GRCh38
rs931668103 Chr.10: 35608140-35608150 on Build GRCh38
rs750338418 Chr.10: 35608026-35608026 on Build GRCh38
rs754768023 Chr.10: 35608095-35608095 on Build GRCh38
rs779335829 Chr.10: 35608127-35608127 on Build GRCh38
rs35793334 Chr.10: 35607931-35607931 on Build GRCh38
rs377737707 Chr.10: 35607904-35607904 on Build GRCh38
rs755622022 Chr.10: 35607828-35607828 on Build GRCh38
rs760773078 Chr.10: 35607851-35607851 on Build GRCh38
rs766524465 Chr.10: 35608260-35608260 on Build GRCh38
rs974565197 Chr.10: 35607874-35607874 on Build GRCh38
rs995090249 Chr.10: 35607817-35607817 on Build GRCh38
rs75803523 Chr.10: 35608197-35608197 on Build GRCh38
rs773485636 Chr.10: 35608250-35608250 on Build GRCh38
rs765570077 Chr.10: 35608152-35608152 on Build GRCh38
rs547212582 Chr.10: 35608163-35608163 on Build GRCh38
rs761980497 Chr.10: 35608202-35608202 on Build GRCh38
rs760289637 Chr.10: 35607937-35607937 on Build GRCh38
rs765372882 Chr.10: 35608249-35608249 on Build GRCh38
rs768263111 Chr.10: 35608133-35608134 on Build GRCh38
rs906012374 Chr.10: 35608161-35608161 on Build GRCh38
rs370405105 Chr.10: 35607862-35607862 on Build GRCh38
rs879364739 Chr.10: 35608023-35608023 on Build GRCh38
rs773599249 Chr.10: 35608144-35608158 on Build GRCh38
rs763398861 Chr.10: 35607989-35607989 on Build GRCh38
rs770741306 Chr.10: 35608191-35608191 on Build GRCh38
rs186885885 Chr.10: 35608256-35608256 on Build GRCh38
rs756265974 Chr.10: 35608113-35608113 on Build GRCh38
rs765464798 Chr.10: 35608204-35608204 on Build GRCh38
rs766348272 Chr.10: 35608092-35608092 on Build GRCh38
rs934981384 Chr.10: 35608145-35608145 on Build GRCh38
rs768505889 Chr.10: 35608246-35608246 on Build GRCh38
rs374801107 Chr.10: 35608003-35608003 on Build GRCh38
rs767449758 Chr.10: 35608097-35608097 on Build GRCh38
rs765707958 Chr.10: 35607857-35607857 on Build GRCh38
rs754631664 Chr.10: 35607812-35607812 on Build GRCh38
rs767127652 Chr.10: 35607870-35607870 on Build GRCh38
rs766673350 Chr.10: 35607917-35607917 on Build GRCh38
rs201736651 Chr.10: 35608178-35608178 on Build GRCh38
rs897994887 Chr.10: 35608164-35608164 on Build GRCh38
rs774905110 Chr.10: 35608056-35608056 on Build GRCh38
rs866307881 Chr.10: 35608138-35608138 on Build GRCh38
rs755271116 Chr.10: 35608211-35608211 on Build GRCh38
rs1044061563 Chr.10: 35607897-35607897 on Build GRCh38
rs773717750 Chr.10: 35607987-35607987 on Build GRCh38
rs757641724 Chr.10: 35607808-35607808 on Build GRCh38
rs778845656 Chr.10: 35607959-35607959 on Build GRCh38
rs142132151 Chr.10: 35607984-35607984 on Build GRCh38
rs974846535 Chr.10: 35608081-35608081 on Build GRCh38
rs770672341 Chr.10: 35607833-35607833 on Build GRCh38
rs201912261 Chr.10: 35608042-35608042 on Build GRCh38
rs747458710 Chr.10: 35608144-35608144 on Build GRCh38
rs751552783 Chr.10: 35608093-35608093 on Build GRCh38
rs769127011 Chr.10: 35608187-35608187 on Build GRCh38
rs151211621 Chr.10: 35608180-35608180 on Build GRCh38
rs141232781 Chr.10: 35607990-35607990 on Build GRCh38
rs148604965 Chr.10: 35608082-35608082 on Build GRCh38

View Full Product Details