Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45412099-45412589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4495191 c.448C>T p.P150S Chr.10: 45412207-45412207 on Build GRCh38
7355264 c.499G>C p.D167H Chr.10: 45412258-45412258 on Build GRCh38
5915896 c.487G>A p.D163N Chr.10: 45412246-45412246 on Build GRCh38
5912995 c.458C>T p.P153L Chr.10: 45412217-45412217 on Build GRCh38
278791 c.469G>A p.D157N Chr.10: 45412228-45412228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777353460 Chr.10: 45412189-45412192 on Build GRCh38
rs757189124 Chr.10: 45412158-45412158 on Build GRCh38
rs142524463 Chr.10: 45412227-45412227 on Build GRCh38
rs938068152 Chr.10: 45412133-45412133 on Build GRCh38
rs980105192 Chr.10: 45412436-45412436 on Build GRCh38
rs567732167 Chr.10: 45412474-45412474 on Build GRCh38
rs929891143 Chr.10: 45412360-45412360 on Build GRCh38
rs551997771 Chr.10: 45412346-45412346 on Build GRCh38
rs995193493 Chr.10: 45412299-45412299 on Build GRCh38
rs988322914 Chr.10: 45412381-45412381 on Build GRCh38
rs746778522 Chr.10: 45412325-45412325 on Build GRCh38
rs372914677 Chr.10: 45412323-45412323 on Build GRCh38
rs953041046 Chr.10: 45412332-45412332 on Build GRCh38
rs377513193 Chr.10: 45412324-45412324 on Build GRCh38
rs892163902 Chr.10: 45412486-45412486 on Build GRCh38
rs778896824 Chr.10: 45412174-45412174 on Build GRCh38
rs780134493 Chr.10: 45412398-45412398 on Build GRCh38
rs956510614 Chr.10: 45412173-45412173 on Build GRCh38
rs769847356 Chr.10: 45412361-45412361 on Build GRCh38
rs1035920152 Chr.10: 45412166-45412166 on Build GRCh38
rs1036187544 Chr.10: 45412531-45412531 on Build GRCh38
rs866759905 Chr.10: 45412399-45412399 on Build GRCh38
rs112595663 Chr.10: 45412455-45412455 on Build GRCh38
rs373850707 Chr.10: 45412555-45412555 on Build GRCh38
rs897668359 Chr.10: 45412541-45412541 on Build GRCh38
rs549498772 Chr.10: 45412473-45412473 on Build GRCh38
rs918000055 Chr.10: 45412515-45412515 on Build GRCh38
rs774553398 Chr.10: 45412520-45412520 on Build GRCh38
rs778720389 Chr.10: 45412313-45412313 on Build GRCh38
rs531087124 Chr.10: 45412408-45412408 on Build GRCh38
rs151005540 Chr.10: 45412290-45412290 on Build GRCh38
rs766792732 Chr.10: 45412263-45412263 on Build GRCh38
rs953374283 Chr.10: 45412363-45412363 on Build GRCh38
rs553385434 Chr.10: 45412501-45412501 on Build GRCh38
rs746531930 Chr.10: 45412331-45412333 on Build GRCh38
rs745728536 Chr.10: 45412183-45412183 on Build GRCh38
rs1011401180 Chr.10: 45412372-45412372 on Build GRCh38
rs376698025 Chr.10: 45412147-45412147 on Build GRCh38
rs369562709 Chr.10: 45412188-45412188 on Build GRCh38
rs756787338 Chr.10: 45412305-45412305 on Build GRCh38
rs566013274 Chr.10: 45412526-45412526 on Build GRCh38
rs753356554 Chr.10: 45412149-45412149 on Build GRCh38
rs779023294 Chr.10: 45412385-45412385 on Build GRCh38
rs770523968 Chr.10: 45412337-45412340 on Build GRCh38
rs753345637 Chr.10: 45412304-45412304 on Build GRCh38
rs773768932 Chr.10: 45412246-45412246 on Build GRCh38
rs761933659 Chr.10: 45412236-45412236 on Build GRCh38
rs968896012 Chr.10: 45412231-45412231 on Build GRCh38
rs200822559 Chr.10: 45412122-45412122 on Build GRCh38
rs763416007 Chr.10: 45412256-45412256 on Build GRCh38
rs199499510 Chr.10: 45412314-45412314 on Build GRCh38
rs528880325 Chr.10: 45412121-45412121 on Build GRCh38
rs869025237 Chr.10: 45412249-45412249 on Build GRCh38
rs760364642 Chr.10: 45412271-45412271 on Build GRCh38
rs201476005 Chr.10: 45412223-45412223 on Build GRCh38
rs200774770 Chr.10: 45412342-45412342 on Build GRCh38
rs182992958 Chr.10: 45412553-45412553 on Build GRCh38
rs371261845 Chr.10: 45412483-45412483 on Build GRCh38
rs747113279 Chr.10: 45412289-45412289 on Build GRCh38
rs560031203 Chr.10: 45412228-45412228 on Build GRCh38
rs770266202 Chr.10: 45412242-45412242 on Build GRCh38
rs111350502 Chr.10: 45412125-45412125 on Build GRCh38
rs748246739 Chr.10: 45412357-45412357 on Build GRCh38
rs749310232 Chr.10: 45412444-45412444 on Build GRCh38
rs780264294 Chr.10: 45412191-45412191 on Build GRCh38
rs771784295 Chr.10: 45412186-45412186 on Build GRCh38
rs1007025211 Chr.10: 45412535-45412535 on Build GRCh38
rs749106048 Chr.10: 45412221-45412221 on Build GRCh38
rs149097803 Chr.10: 45412492-45412492 on Build GRCh38
rs565619560 Chr.10: 45412376-45412376 on Build GRCh38
rs909715013 Chr.10: 45412411-45412411 on Build GRCh38
rs1036645395 Chr.10: 45412278-45412278 on Build GRCh38
rs1044826799 Chr.10: 45412448-45412448 on Build GRCh38
rs747182768 Chr.10: 45412200-45412200 on Build GRCh38

View Full Product Details