Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70772433-70772926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBATA
Entrez Gene ID
219793
Gene Name
thymus, brain and testes associated
Gene Aliases
C10orf27, SPATIAL
UniGene
Hs.386698
Gene Chromosome Location
Chr.10: 70771237-70785419 on Build GRCh38
Transcripts
NM_001318241.1 NM_001318242.1 NM_001318243.1 NM_152710.3 NR_134531.1 NR_134532.1 NR_134533.1 NR_134534.1 XM_017015845.1 XM_017015846.1 XM_017015847.1 XM_017015848.1 XM_017015849.1 XM_017015852.1 XM_017015853.1 XM_017015854.1 XM_017015855.1 XM_017015856.1 XM_017015857.1 XM_017015858.1 XM_017015859.1 XM_017015860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
920052 c.927G>A p.P309P Chr.10: 70772557-70772557 on Build GRCh38
295640 c.960C>T p.S320S Chr.10: 70772524-70772524 on Build GRCh38
1745865 c.961G>A p.E321K Chr.10: 70772523-70772523 on Build GRCh38
7234055 c.970+9C>T p.? Chr.10: 70772505-70772505 on Build GRCh38
920050 c.941T>C p.I314T Chr.10: 70772543-70772543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560810843 Chr.10: 70772888-70772888 on Build GRCh38
rs771015912 Chr.10: 70772473-70772473 on Build GRCh38
rs202218556 Chr.10: 70772557-70772557 on Build GRCh38
rs1022505944 Chr.10: 70772787-70772787 on Build GRCh38
rs749464433 Chr.10: 70772780-70772780 on Build GRCh38
rs574735003 Chr.10: 70772871-70772871 on Build GRCh38
rs969776534 Chr.10: 70772631-70772631 on Build GRCh38
rs779596620 Chr.10: 70772529-70772529 on Build GRCh38
rs554404951 Chr.10: 70772700-70772700 on Build GRCh38
rs764351493 Chr.10: 70772593-70772593 on Build GRCh38
rs749992519 Chr.10: 70772524-70772524 on Build GRCh38
rs757959045 Chr.10: 70772489-70772489 on Build GRCh38
rs1005525957 Chr.10: 70772706-70772706 on Build GRCh38
rs1018705146 Chr.10: 70772708-70772708 on Build GRCh38
rs556284850 Chr.10: 70772851-70772851 on Build GRCh38
rs775630791 Chr.10: 70772481-70772482 on Build GRCh38
rs564742690 Chr.10: 70772880-70772880 on Build GRCh38
rs796276485 Chr.10: 70772802-70772802 on Build GRCh38
rs746779801 Chr.10: 70772531-70772531 on Build GRCh38
rs532217090 Chr.10: 70772612-70772612 on Build GRCh38
rs762957357 Chr.10: 70772471-70772471 on Build GRCh38
rs929100470 Chr.10: 70772875-70772875 on Build GRCh38
rs754410048 Chr.10: 70772507-70772507 on Build GRCh38
rs1052568296 Chr.10: 70772748-70772748 on Build GRCh38
rs770240036 Chr.10: 70772792-70772792 on Build GRCh38
rs757897270 Chr.10: 70772517-70772517 on Build GRCh38
rs535577351 Chr.10: 70772898-70772898 on Build GRCh38
rs899602716 Chr.10: 70772814-70772814 on Build GRCh38
rs369110349 Chr.10: 70772465-70772465 on Build GRCh38
rs867638152 Chr.10: 70772896-70772896 on Build GRCh38
rs939045624 Chr.10: 70772811-70772811 on Build GRCh38
rs988723340 Chr.10: 70772763-70772763 on Build GRCh38
rs2253801 Chr.10: 70772482-70772482 on Build GRCh38
rs761038602 Chr.10: 70772589-70772589 on Build GRCh38
rs777149828 Chr.10: 70772716-70772716 on Build GRCh38
rs753054240 Chr.10: 70772484-70772484 on Build GRCh38
rs891018026 Chr.10: 70772840-70772840 on Build GRCh38
rs10823618 Chr.10: 70772687-70772687 on Build GRCh38
rs998251962 Chr.10: 70772746-70772746 on Build GRCh38
rs757879579 Chr.10: 70772606-70772606 on Build GRCh38
rs764591910 Chr.10: 70772499-70772499 on Build GRCh38
rs939490136 Chr.10: 70772469-70772469 on Build GRCh38
rs765892566 Chr.10: 70772603-70772603 on Build GRCh38
rs558309942 Chr.10: 70772799-70772799 on Build GRCh38
rs566420935 Chr.10: 70772703-70772703 on Build GRCh38
rs376080763 Chr.10: 70772573-70772573 on Build GRCh38
rs775575090 Chr.10: 70772588-70772588 on Build GRCh38
rs777202128 Chr.10: 70772598-70772598 on Build GRCh38
rs919013226 Chr.10: 70772857-70772857 on Build GRCh38
rs897645969 Chr.10: 70772622-70772622 on Build GRCh38
rs370227499 Chr.10: 70772548-70772548 on Build GRCh38
rs929173998 Chr.10: 70772649-70772649 on Build GRCh38
rs577302565 Chr.10: 70772828-70772828 on Build GRCh38
rs148997480 Chr.10: 70772836-70772836 on Build GRCh38
rs550318566 Chr.10: 70772618-70772618 on Build GRCh38
rs372420502 Chr.10: 70772532-70772532 on Build GRCh38
rs751188408 Chr.10: 70772613-70772613 on Build GRCh38
rs980851147 Chr.10: 70772549-70772549 on Build GRCh38
rs143899859 Chr.10: 70772544-70772544 on Build GRCh38
rs568243678 Chr.10: 70772878-70772878 on Build GRCh38
rs568809704 Chr.10: 70772644-70772644 on Build GRCh38
rs899861756 Chr.10: 70772730-70772730 on Build GRCh38
rs376929862 Chr.10: 70772477-70772477 on Build GRCh38
rs920730155 Chr.10: 70772625-70772625 on Build GRCh38
rs553914355 Chr.10: 70772899-70772899 on Build GRCh38
rs148121646 Chr.10: 70772525-70772525 on Build GRCh38
rs12415264 Chr.10: 70772558-70772558 on Build GRCh38
rs770596624 Chr.10: 70772659-70772659 on Build GRCh38
rs201710832 Chr.10: 70772804-70772804 on Build GRCh38
rs996904228 Chr.10: 70772815-70772815 on Build GRCh38
rs113540558 Chr.10: 70772750-70772750 on Build GRCh38
rs956851175 Chr.10: 70772759-70772759 on Build GRCh38
rs202040509 Chr.10: 70772523-70772523 on Build GRCh38
rs754721551 Chr.10: 70772617-70772617 on Build GRCh38
rs762376436 Chr.10: 70772599-70772599 on Build GRCh38
rs760974947 Chr.10: 70772487-70772487 on Build GRCh38
rs749904959 Chr.10: 70772605-70772605 on Build GRCh38
rs943267123 Chr.10: 70772897-70772897 on Build GRCh38
rs915514400 Chr.10: 70772545-70772545 on Build GRCh38
rs980827677 Chr.10: 70772643-70772643 on Build GRCh38
rs530348290 Chr.10: 70772470-70772470 on Build GRCh38

View Full Product Details