Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73647728-73648216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNPO2L
Entrez Gene ID
79933
Gene Name
synaptopodin 2 like
Gene Aliases
-
UniGene
Hs.645273
Gene Chromosome Location
Chr.10: 73644881-73656105 on Build GRCh38
Transcripts
NM_001114133.2 NM_024875.4 XM_005270158.2 XM_005270159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7392763 c.1711C>A p.P571T Chr.10: 73647941-73647941 on Build GRCh38
3751968 c.1089T>G p.P363P Chr.10: 73647891-73647891 on Build GRCh38
6795323 c.1898G>A p.R633Q Chr.10: 73647754-73647754 on Build GRCh38
4015779 c.1861A>G p.T621A Chr.10: 73647791-73647791 on Build GRCh38
1964027 c.1107C>T p.R369R Chr.10: 73647873-73647873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs57006992 Chr.10: 73648129-73648129 on Build GRCh38
rs776237409 Chr.10: 73647987-73647987 on Build GRCh38
rs573227336 Chr.10: 73647814-73647814 on Build GRCh38
rs779391504 Chr.10: 73648106-73648106 on Build GRCh38
rs752937425 Chr.10: 73648036-73648036 on Build GRCh38
rs779548300 Chr.10: 73648081-73648081 on Build GRCh38
rs759446036 Chr.10: 73647782-73647782 on Build GRCh38
rs759581542 Chr.10: 73648015-73648016 on Build GRCh38
rs770534757 Chr.10: 73647876-73647876 on Build GRCh38
rs978824880 Chr.10: 73647991-73647991 on Build GRCh38
rs745364754 Chr.10: 73647972-73647972 on Build GRCh38
rs560442481 Chr.10: 73648189-73648189 on Build GRCh38
rs773960032 Chr.10: 73647881-73647881 on Build GRCh38
rs753063899 Chr.10: 73647931-73647931 on Build GRCh38
rs764157215 Chr.10: 73648148-73648148 on Build GRCh38
rs767671228 Chr.10: 73648138-73648138 on Build GRCh38
rs957946381 Chr.10: 73648109-73648109 on Build GRCh38
rs758778667 Chr.10: 73648180-73648180 on Build GRCh38
rs771908237 Chr.10: 73647974-73647974 on Build GRCh38
rs754310220 Chr.10: 73648157-73648157 on Build GRCh38
rs531403580 Chr.10: 73648174-73648174 on Build GRCh38
rs771579030 Chr.10: 73648095-73648095 on Build GRCh38
rs866003442 Chr.10: 73648172-73648172 on Build GRCh38
rs990699257 Chr.10: 73648111-73648111 on Build GRCh38
rs760426522 Chr.10: 73647982-73647982 on Build GRCh38
rs1020409825 Chr.10: 73647954-73647954 on Build GRCh38
rs770274328 Chr.10: 73647857-73647857 on Build GRCh38
rs200938828 Chr.10: 73648169-73648169 on Build GRCh38
rs766197020 Chr.10: 73647771-73647771 on Build GRCh38
rs939098955 Chr.10: 73648026-73648026 on Build GRCh38
rs773114857 Chr.10: 73648132-73648132 on Build GRCh38
rs769783352 Chr.10: 73648131-73648131 on Build GRCh38
rs977720659 Chr.10: 73647947-73647947 on Build GRCh38
rs369315169 Chr.10: 73647903-73647903 on Build GRCh38
rs779626407 Chr.10: 73648183-73648183 on Build GRCh38
rs150403805 Chr.10: 73647783-73647783 on Build GRCh38
rs201121778 Chr.10: 73647953-73647953 on Build GRCh38
rs759382538 Chr.10: 73647866-73647866 on Build GRCh38
rs752297941 Chr.10: 73648149-73648150 on Build GRCh38
rs368785926 Chr.10: 73647859-73647859 on Build GRCh38
rs760509432 Chr.10: 73647794-73647795 on Build GRCh38
rs940395814 Chr.10: 73647834-73647834 on Build GRCh38
rs781547598 Chr.10: 73647839-73647839 on Build GRCh38
rs766465831 Chr.10: 73647914-73647914 on Build GRCh38
rs967765956 Chr.10: 73647975-73647975 on Build GRCh38
rs775212790 Chr.10: 73647894-73647894 on Build GRCh38
rs765884312 Chr.10: 73648171-73648171 on Build GRCh38
rs775457373 Chr.10: 73647929-73647929 on Build GRCh38
rs780290902 Chr.10: 73647832-73647832 on Build GRCh38
rs372058702 Chr.10: 73647773-73647773 on Build GRCh38
rs375335895 Chr.10: 73647781-73647781 on Build GRCh38
rs761749365 Chr.10: 73647996-73647996 on Build GRCh38
rs756007816 Chr.10: 73647805-73647805 on Build GRCh38
rs185248935 Chr.10: 73648124-73648124 on Build GRCh38
rs757846217 Chr.10: 73648080-73648080 on Build GRCh38
rs760504758 Chr.10: 73647900-73647900 on Build GRCh38
rs766285135 Chr.10: 73647821-73647822 on Build GRCh38
rs778187269 Chr.10: 73647942-73647942 on Build GRCh38
rs754199606 Chr.10: 73648072-73648072 on Build GRCh38
rs760299945 Chr.10: 73648057-73648057 on Build GRCh38
rs781475255 Chr.10: 73647925-73647925 on Build GRCh38
rs755141853 Chr.10: 73647837-73647837 on Build GRCh38
rs544176673 Chr.10: 73648029-73648029 on Build GRCh38
rs576968134 Chr.10: 73647983-73647983 on Build GRCh38
rs778582448 Chr.10: 73647971-73647971 on Build GRCh38
rs143723429 Chr.10: 73647763-73647763 on Build GRCh38
rs775185487 Chr.10: 73647979-73647979 on Build GRCh38
rs774125238 Chr.10: 73647779-73647779 on Build GRCh38
rs753777568 Chr.10: 73647907-73647907 on Build GRCh38
rs771517903 Chr.10: 73647969-73647969 on Build GRCh38
rs767685931 Chr.10: 73648035-73648035 on Build GRCh38
rs765090590 Chr.10: 73648007-73648007 on Build GRCh38
rs756249642 Chr.10: 73648047-73648047 on Build GRCh38
rs761694207 Chr.10: 73647908-73647908 on Build GRCh38
rs758451069 Chr.10: 73647823-73647823 on Build GRCh38
rs540423694 Chr.10: 73647836-73647836 on Build GRCh38
rs4746139 Chr.10: 73647891-73647891 on Build GRCh38
rs757672597 Chr.10: 73648163-73648163 on Build GRCh38
rs767453923 Chr.10: 73647789-73647789 on Build GRCh38
rs148105482 Chr.10: 73647790-73647790 on Build GRCh38
rs756482053 Chr.10: 73647936-73647936 on Build GRCh38
rs770274901 Chr.10: 73647762-73647762 on Build GRCh38
rs750526493 Chr.10: 73647822-73647822 on Build GRCh38
rs916422977 Chr.10: 73647764-73647764 on Build GRCh38
rs573354456 Chr.10: 73648146-73648146 on Build GRCh38
rs925249140 Chr.10: 73647756-73647756 on Build GRCh38
rs775383008 Chr.10: 73648140-73648140 on Build GRCh38
rs776377829 Chr.10: 73647830-73647831 on Build GRCh38
rs760966081 Chr.10: 73648142-73648142 on Build GRCh38
rs748480832 Chr.10: 73647845-73647845 on Build GRCh38
rs544284671 Chr.10: 73648149-73648151 on Build GRCh38
rs1038338455 Chr.10: 73647862-73647862 on Build GRCh38
rs544564741 Chr.10: 73647915-73647915 on Build GRCh38
rs745419556 Chr.10: 73648087-73648087 on Build GRCh38
rs755195176 Chr.10: 73647921-73647921 on Build GRCh38
rs746692338 Chr.10: 73648116-73648116 on Build GRCh38
rs558424319 Chr.10: 73647858-73647858 on Build GRCh38
rs180752653 Chr.10: 73647882-73647882 on Build GRCh38

View Full Product Details