Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73765715-73766216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC24C
Entrez Gene ID
9632
Gene Name
SEC24 homolog C, COPII coat complex component
Gene Aliases
-
UniGene
Hs.81964
Gene Chromosome Location
Chr.10: 73744164-73772175 on Build GRCh38
Transcripts
NM_004922.3 NM_198597.2 XM_011540380.2 XM_011540381.2 XM_017016967.1 XM_017016968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130554 c.1084C>T p.L362L Chr.10: 73765874-73765874 on Build GRCh38
5826791 c.1011_1012insC p.Q340fs*17 Chr.10: 73765801-73765802 on Build GRCh38
4015788 c.1575G>T p.E525D Chr.10: 73766178-73766178 on Build GRCh38
1964212 c.1368_1369insCC p.Q459fs*33 Chr.10: 73765801-73765802 on Build GRCh38
3738718 c.1450T>C p.Y484H Chr.10: 73765883-73765883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371408137 Chr.10: 73766049-73766049 on Build GRCh38
rs144494394 Chr.10: 73765977-73765978 on Build GRCh38
rs747450230 Chr.10: 73765955-73765955 on Build GRCh38
rs772852585 Chr.10: 73765861-73765861 on Build GRCh38
rs768167472 Chr.10: 73765959-73765959 on Build GRCh38
rs758739472 Chr.10: 73765941-73765941 on Build GRCh38
rs201681162 Chr.10: 73766040-73766040 on Build GRCh38
rs374607897 Chr.10: 73766074-73766074 on Build GRCh38
rs762996842 Chr.10: 73765872-73765872 on Build GRCh38
rs759536234 Chr.10: 73766057-73766057 on Build GRCh38
rs768112378 Chr.10: 73765835-73765835 on Build GRCh38
rs764332026 Chr.10: 73766065-73766065 on Build GRCh38
rs544229394 Chr.10: 73766036-73766036 on Build GRCh38
rs781081066 Chr.10: 73765859-73765859 on Build GRCh38
rs760357262 Chr.10: 73765765-73765765 on Build GRCh38
rs764652839 Chr.10: 73765801-73765802 on Build GRCh38
rs998989793 Chr.10: 73765758-73765758 on Build GRCh38
rs780732293 Chr.10: 73766153-73766153 on Build GRCh38
rs752803726 Chr.10: 73765795-73765795 on Build GRCh38
rs987303809 Chr.10: 73766001-73766001 on Build GRCh38
rs141567992 Chr.10: 73766111-73766111 on Build GRCh38
rs1039097714 Chr.10: 73766062-73766062 on Build GRCh38
rs759899094 Chr.10: 73765751-73765751 on Build GRCh38
rs749457980 Chr.10: 73765805-73765805 on Build GRCh38
rs937648272 Chr.10: 73766029-73766029 on Build GRCh38
rs565250700 Chr.10: 73766025-73766025 on Build GRCh38
rs900475281 Chr.10: 73765750-73765750 on Build GRCh38
rs747747931 Chr.10: 73766089-73766089 on Build GRCh38
rs768013427 Chr.10: 73766070-73766074 on Build GRCh38
rs746543668 Chr.10: 73765834-73765834 on Build GRCh38
rs573242472 Chr.10: 73765864-73765864 on Build GRCh38
rs755284674 Chr.10: 73765745-73765745 on Build GRCh38
rs779130794 Chr.10: 73765808-73765808 on Build GRCh38
rs1030327632 Chr.10: 73766140-73766140 on Build GRCh38
rs758265928 Chr.10: 73765781-73765781 on Build GRCh38
rs771668560 Chr.10: 73765752-73765752 on Build GRCh38
rs752128575 Chr.10: 73766105-73766105 on Build GRCh38
rs377324151 Chr.10: 73766044-73766044 on Build GRCh38
rs756269013 Chr.10: 73765891-73765891 on Build GRCh38
rs764215279 Chr.10: 73765895-73765895 on Build GRCh38
rs754167310 Chr.10: 73765896-73765896 on Build GRCh38
rs779370342 Chr.10: 73765912-73765912 on Build GRCh38
rs778176043 Chr.10: 73765800-73765800 on Build GRCh38
rs375272522 Chr.10: 73766174-73766174 on Build GRCh38
rs775029519 Chr.10: 73765811-73765811 on Build GRCh38
rs200672501 Chr.10: 73766097-73766097 on Build GRCh38
rs780568747 Chr.10: 73765952-73765952 on Build GRCh38
rs751505979 Chr.10: 73765881-73765881 on Build GRCh38
rs200985881 Chr.10: 73766166-73766166 on Build GRCh38
rs550791385 Chr.10: 73766094-73766094 on Build GRCh38
rs577155814 Chr.10: 73766031-73766031 on Build GRCh38
rs535635820 Chr.10: 73765771-73765771 on Build GRCh38
rs776268029 Chr.10: 73765839-73765839 on Build GRCh38
rs368096920 Chr.10: 73766161-73766161 on Build GRCh38
rs200016285 Chr.10: 73766103-73766103 on Build GRCh38
rs374796759 Chr.10: 73766053-73766053 on Build GRCh38
rs1048018129 Chr.10: 73766038-73766038 on Build GRCh38
rs911717321 Chr.10: 73766015-73766015 on Build GRCh38
rs750617493 Chr.10: 73766090-73766090 on Build GRCh38
rs199937551 Chr.10: 73765772-73765772 on Build GRCh38
rs746192574 Chr.10: 73765930-73765930 on Build GRCh38
rs781373238 Chr.10: 73765783-73765783 on Build GRCh38
rs991860156 Chr.10: 73766030-73766030 on Build GRCh38
rs757546572 Chr.10: 73766088-73766088 on Build GRCh38
rs375692050 Chr.10: 73765807-73765807 on Build GRCh38
rs934725153 Chr.10: 73766162-73766162 on Build GRCh38
rs761442412 Chr.10: 73765860-73765860 on Build GRCh38
rs553695287 Chr.10: 73765774-73765774 on Build GRCh38
rs756217742 Chr.10: 73765796-73765796 on Build GRCh38
rs146878586 Chr.10: 73765762-73765762 on Build GRCh38
rs772423989 Chr.10: 73765809-73765809 on Build GRCh38
rs377689283 Chr.10: 73765902-73765902 on Build GRCh38
rs540393385 Chr.10: 73765879-73765879 on Build GRCh38
rs776115491 Chr.10: 73765964-73765964 on Build GRCh38

View Full Product Details