Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86958452-86958959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
UniGene
Hs.524479
Gene Chromosome Location
Chr.10: 86935540-86971311 on Build GRCh38
Transcripts
XM_006717970.3 NM_003087.2 XM_011540105.2

Gene Symbol
SNCG
Entrez Gene ID
6623
Gene Name
synuclein gamma
Gene Aliases
BCSG1, SR
UniGene
Hs.349470
Gene Chromosome Location
Chr.10: 86958531-86963260 on Build GRCh38
Transcripts
XM_006717970.3 NM_003087.2 XM_011540105.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5539483 c.4G>A p.D2N Chr.10: 86958701-86958701 on Build GRCh38
5539484 c.4G>A p.D2N Chr.10: 86958701-86958701 on Build GRCh38
1684221 c.109delG p.V37fs*16 Chr.10: 86958806-86958806 on Build GRCh38
1603819 c.121+9G>T p.? Chr.10: 86958827-86958827 on Build GRCh38
139848 c.40G>A p.G14S Chr.10: 86958737-86958737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370211729 Chr.10: 86958517-86958517 on Build GRCh38
rs757193684 Chr.10: 86958648-86958648 on Build GRCh38
rs770142696 Chr.10: 86958764-86958764 on Build GRCh38
rs1800714 Chr.10: 86958649-86958649 on Build GRCh38
rs945013447 Chr.10: 86958580-86958580 on Build GRCh38
rs141968769 Chr.10: 86958704-86958704 on Build GRCh38
rs534041905 Chr.10: 86958595-86958595 on Build GRCh38
rs984959278 Chr.10: 86958881-86958881 on Build GRCh38
rs953098873 Chr.10: 86958530-86958530 on Build GRCh38
rs760075709 Chr.10: 86958808-86958808 on Build GRCh38
rs763058285 Chr.10: 86958676-86958676 on Build GRCh38
rs926699915 Chr.10: 86958624-86958624 on Build GRCh38
rs539422782 Chr.10: 86958495-86958495 on Build GRCh38
rs545915910 Chr.10: 86958637-86958637 on Build GRCh38
rs778081965 Chr.10: 86958766-86958766 on Build GRCh38
rs948444151 Chr.10: 86958500-86958500 on Build GRCh38
rs972713746 Chr.10: 86958596-86958596 on Build GRCh38
rs371943257 Chr.10: 86958747-86958747 on Build GRCh38
rs778767282 Chr.10: 86958650-86958650 on Build GRCh38
rs778801167 Chr.10: 86958778-86958778 on Build GRCh38
rs749400883 Chr.10: 86958794-86958794 on Build GRCh38
rs748383493 Chr.10: 86958758-86958758 on Build GRCh38
rs760893201 Chr.10: 86958815-86958815 on Build GRCh38
rs187691882 Chr.10: 86958928-86958928 on Build GRCh38
rs369666481 Chr.10: 86958830-86958830 on Build GRCh38
rs531523381 Chr.10: 86958785-86958785 on Build GRCh38
rs182363578 Chr.10: 86958611-86958611 on Build GRCh38
rs758348310 Chr.10: 86958861-86958861 on Build GRCh38
rs545602900 Chr.10: 86958636-86958636 on Build GRCh38
rs375296971 Chr.10: 86958751-86958751 on Build GRCh38
rs945385511 Chr.10: 86958570-86958570 on Build GRCh38
rs773418525 Chr.10: 86958673-86958673 on Build GRCh38
rs747921799 Chr.10: 86958812-86958812 on Build GRCh38
rs748376325 Chr.10: 86958662-86958662 on Build GRCh38
rs971731049 Chr.10: 86958810-86958810 on Build GRCh38
rs749513753 Chr.10: 86958848-86958850 on Build GRCh38
rs201625410 Chr.10: 86958681-86958681 on Build GRCh38
rs903072023 Chr.10: 86958690-86958690 on Build GRCh38
rs926775196 Chr.10: 86958882-86958882 on Build GRCh38
rs555640385 Chr.10: 86958605-86958605 on Build GRCh38
rs372862174 Chr.10: 86958661-86958661 on Build GRCh38
rs1045217381 Chr.10: 86958514-86958514 on Build GRCh38
rs753361891 Chr.10: 86958745-86958745 on Build GRCh38
rs750382106 Chr.10: 86958850-86958850 on Build GRCh38
rs771094834 Chr.10: 86958805-86958805 on Build GRCh38
rs771864911 Chr.10: 86958654-86958654 on Build GRCh38
rs926770869 Chr.10: 86958519-86958519 on Build GRCh38
rs374817748 Chr.10: 86958667-86958667 on Build GRCh38
rs774949338 Chr.10: 86958698-86958698 on Build GRCh38
rs758595946 Chr.10: 86958606-86958606 on Build GRCh38
rs764700777 Chr.10: 86958848-86958848 on Build GRCh38
rs1056548418 Chr.10: 86958927-86958927 on Build GRCh38
rs953173688 Chr.10: 86958728-86958728 on Build GRCh38
rs761356862 Chr.10: 86958840-86958840 on Build GRCh38
rs938110877 Chr.10: 86958926-86958926 on Build GRCh38
rs1800373 Chr.10: 86958679-86958679 on Build GRCh38
rs945584627 Chr.10: 86958856-86958856 on Build GRCh38
rs113805468 Chr.10: 86958610-86958610 on Build GRCh38
rs890559509 Chr.10: 86958586-86958586 on Build GRCh38
rs753451764 Chr.10: 86958838-86958838 on Build GRCh38
rs371264406 Chr.10: 86958653-86958653 on Build GRCh38
rs551186729 Chr.10: 86958828-86958828 on Build GRCh38
rs1042045978 Chr.10: 86958684-86958684 on Build GRCh38
rs148591902 Chr.10: 86958811-86958811 on Build GRCh38
rs758173640 Chr.10: 86958754-86958754 on Build GRCh38
rs557161416 Chr.10: 86958621-86958621 on Build GRCh38
rs74150417 Chr.10: 86958727-86958727 on Build GRCh38
rs540522786 Chr.10: 86958739-86958739 on Build GRCh38
rs778856990 Chr.10: 86958750-86958750 on Build GRCh38
rs1007616896 Chr.10: 86958591-86958591 on Build GRCh38
rs985764567 Chr.10: 86958560-86958560 on Build GRCh38
rs1054077966 Chr.10: 86958543-86958543 on Build GRCh38
rs529452849 Chr.10: 86958756-86958756 on Build GRCh38
rs763672443 Chr.10: 86958740-86958740 on Build GRCh38
rs576668884 Chr.10: 86958524-86958524 on Build GRCh38
rs377375661 Chr.10: 86958563-86958563 on Build GRCh38
rs775047510 Chr.10: 86958806-86958806 on Build GRCh38

View Full Product Details