Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89306675-89307184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT2
Entrez Gene ID
3433
Gene Name
interferon induced protein with tetratricopeptide repeats 2
Gene Aliases
G10P2, GARG-39, IFI-54, IFI-54K, IFI54, IFIT-2, ISG-54 K, ISG-54K, ISG54, P54, cig42
UniGene
Hs.437609
Gene Chromosome Location
Chr.10: 89301949-89309276 on Build GRCh38
Transcripts
NM_001547.4

Gene Symbol
LOC105378419
Entrez Gene ID
105378419
Gene Name
uncharacterized LOC105378419
Gene Aliases
-
UniGene
Hs.702467
Gene Chromosome Location
Chr.10: 89293473-89316145 on Build GRCh38
Transcripts
NM_001547.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466059 c.955G>T p.A319S Chr.10: 89306911-89306911 on Build GRCh38
6683837 c.758C>T p.A253V Chr.10: 89306714-89306714 on Build GRCh38
4758734 c.886T>A p.F296I Chr.10: 89306842-89306842 on Build GRCh38
3397292 c.1208A>T p.Q403L Chr.10: 89307164-89307164 on Build GRCh38
6683838 c.1225G>T p.E409* Chr.10: 89307181-89307181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926080408 Chr.10: 89306934-89306934 on Build GRCh38
rs371414597 Chr.10: 89307018-89307018 on Build GRCh38
rs558689246 Chr.10: 89307024-89307024 on Build GRCh38
rs776936260 Chr.10: 89306794-89306794 on Build GRCh38
rs918527205 Chr.10: 89307041-89307041 on Build GRCh38
rs751422356 Chr.10: 89307104-89307104 on Build GRCh38
rs748367775 Chr.10: 89306857-89306857 on Build GRCh38
rs745389813 Chr.10: 89306960-89306960 on Build GRCh38
rs1051622441 Chr.10: 89306969-89306969 on Build GRCh38
rs754754578 Chr.10: 89307115-89307115 on Build GRCh38
rs988857913 Chr.10: 89306940-89306940 on Build GRCh38
rs61729485 Chr.10: 89306849-89306849 on Build GRCh38
rs779315161 Chr.10: 89306717-89306717 on Build GRCh38
rs761596604 Chr.10: 89307073-89307073 on Build GRCh38
rs773199281 Chr.10: 89306758-89306758 on Build GRCh38
rs373734660 Chr.10: 89306712-89306712 on Build GRCh38
rs200688103 Chr.10: 89306745-89306745 on Build GRCh38
rs758825312 Chr.10: 89306831-89306831 on Build GRCh38
rs768098655 Chr.10: 89307087-89307087 on Build GRCh38
rs368019671 Chr.10: 89306959-89306959 on Build GRCh38
rs1026002095 Chr.10: 89307084-89307084 on Build GRCh38
rs768227773 Chr.10: 89307062-89307062 on Build GRCh38
rs775434792 Chr.10: 89307057-89307057 on Build GRCh38
rs150267128 Chr.10: 89306910-89306910 on Build GRCh38
rs760287568 Chr.10: 89306927-89306927 on Build GRCh38
rs758215504 Chr.10: 89307144-89307144 on Build GRCh38
rs1054617659 Chr.10: 89307111-89307111 on Build GRCh38
rs114752135 Chr.10: 89306967-89306967 on Build GRCh38
rs750791162 Chr.10: 89306820-89306820 on Build GRCh38
rs759583775 Chr.10: 89306903-89306903 on Build GRCh38
rs780379595 Chr.10: 89306847-89306847 on Build GRCh38
rs865927859 Chr.10: 89307124-89307124 on Build GRCh38
rs202222227 Chr.10: 89306744-89306744 on Build GRCh38
rs758912657 Chr.10: 89306789-89306789 on Build GRCh38
rs775027680 Chr.10: 89306911-89306911 on Build GRCh38
rs201873020 Chr.10: 89307033-89307033 on Build GRCh38
rs376793909 Chr.10: 89306854-89306854 on Build GRCh38
rs756283221 Chr.10: 89307007-89307007 on Build GRCh38
rs752866846 Chr.10: 89307006-89307006 on Build GRCh38
rs551876697 Chr.10: 89307060-89307060 on Build GRCh38
rs537050168 Chr.10: 89306707-89306707 on Build GRCh38
rs199839737 Chr.10: 89307083-89307083 on Build GRCh38
rs140944939 Chr.10: 89306756-89306756 on Build GRCh38
rs770732258 Chr.10: 89306774-89306774 on Build GRCh38
rs770191578 Chr.10: 89306864-89306864 on Build GRCh38
rs1727 Chr.10: 89307012-89307012 on Build GRCh38
rs756997117 Chr.10: 89307014-89307014 on Build GRCh38
rs990612354 Chr.10: 89306946-89306946 on Build GRCh38
rs748387974 Chr.10: 89306754-89306754 on Build GRCh38
rs752339055 Chr.10: 89306800-89306800 on Build GRCh38
rs777594055 Chr.10: 89306874-89306874 on Build GRCh38
rs763699307 Chr.10: 89306935-89306935 on Build GRCh38
rs577049617 Chr.10: 89306793-89306793 on Build GRCh38
rs750923936 Chr.10: 89306728-89306728 on Build GRCh38
rs371767716 Chr.10: 89307086-89307086 on Build GRCh38
rs767685090 Chr.10: 89306988-89306988 on Build GRCh38
rs768881048 Chr.10: 89306751-89306751 on Build GRCh38
rs774089331 Chr.10: 89306893-89306893 on Build GRCh38
rs758874731 Chr.10: 89306742-89306742 on Build GRCh38
rs765561408 Chr.10: 89306812-89306812 on Build GRCh38
rs770771403 Chr.10: 89306877-89306877 on Build GRCh38
rs770640915 Chr.10: 89306767-89306767 on Build GRCh38
rs780155307 Chr.10: 89306748-89306748 on Build GRCh38
rs750351586 Chr.10: 89306966-89306966 on Build GRCh38
rs369628682 Chr.10: 89307118-89307118 on Build GRCh38
rs954439 Chr.10: 89306703-89306703 on Build GRCh38
rs755600592 Chr.10: 89306970-89306970 on Build GRCh38
rs778538745 Chr.10: 89307128-89307128 on Build GRCh38
rs3898075 Chr.10: 89306876-89306876 on Build GRCh38
rs765322482 Chr.10: 89306942-89306942 on Build GRCh38
rs1020601979 Chr.10: 89306819-89306819 on Build GRCh38
rs766194472 Chr.10: 89307102-89307102 on Build GRCh38
rs367905315 Chr.10: 89307082-89307082 on Build GRCh38
rs370750247 Chr.10: 89306753-89306753 on Build GRCh38
rs745779294 Chr.10: 89307054-89307054 on Build GRCh38
rs893318694 Chr.10: 89307142-89307142 on Build GRCh38
rs182750115 Chr.10: 89306945-89306945 on Build GRCh38
rs760373769 Chr.10: 89306808-89306808 on Build GRCh38
rs755550339 Chr.10: 89306733-89306733 on Build GRCh38
rs374921431 Chr.10: 89307063-89307063 on Build GRCh38
rs377588926 Chr.10: 89306749-89306749 on Build GRCh38
rs756195054 Chr.10: 89307099-89307099 on Build GRCh38
rs745619883 Chr.10: 89307138-89307138 on Build GRCh38
rs755045060 Chr.10: 89306856-89306856 on Build GRCh38
rs772035501 Chr.10: 89307056-89307056 on Build GRCh38
rs762927189 Chr.10: 89306773-89306773 on Build GRCh38
rs376313240 Chr.10: 89307093-89307093 on Build GRCh38
rs774366317 Chr.10: 89306785-89306785 on Build GRCh38

View Full Product Details