Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91485069-91485547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD2
Entrez Gene ID
143279
Gene Name
HECT domain E3 ubiquitin protein ligase 2
Gene Aliases
-
UniGene
Hs.596096
Gene Chromosome Location
Chr.10: 91409280-91514829 on Build GRCh38
Transcripts
NM_001284274.1 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 XM_017015752.1 NR_024467.1

Gene Symbol
HECTD2-AS1
Entrez Gene ID
100188947
Gene Name
HECTD2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.535293
Gene Chromosome Location
Chr.10: 91306962-91611460 on Build GRCh38
Transcripts
NM_001284274.1 NM_182765.4 NR_104291.1 XM_017015750.1 XM_017015751.1 XM_017015752.1 NR_024467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265435 c.1070G>T p.W357L Chr.10: 91485279-91485279 on Build GRCh38
4573151 c.989T>C p.V330A Chr.10: 91485198-91485198 on Build GRCh38
6907910 c.1079T>G p.F360C Chr.10: 91485288-91485288 on Build GRCh38
5937431 c.1006C>T p.P336S Chr.10: 91485215-91485215 on Build GRCh38
4016578 c.977C>T p.A326V Chr.10: 91485186-91485186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749759731 Chr.10: 91485310-91485310 on Build GRCh38
rs948063309 Chr.10: 91485502-91485502 on Build GRCh38
rs577204113 Chr.10: 91485383-91485383 on Build GRCh38
rs374775084 Chr.10: 91485346-91485346 on Build GRCh38
rs764060856 Chr.10: 91485140-91485140 on Build GRCh38
rs778428769 Chr.10: 91485170-91485170 on Build GRCh38
rs1021336682 Chr.10: 91485361-91485361 on Build GRCh38
rs554155639 Chr.10: 91485211-91485211 on Build GRCh38
rs760494442 Chr.10: 91485348-91485348 on Build GRCh38
rs373344630 Chr.10: 91485223-91485223 on Build GRCh38
rs931143591 Chr.10: 91485480-91485480 on Build GRCh38
rs772387348 Chr.10: 91485332-91485332 on Build GRCh38
rs959193818 Chr.10: 91485415-91485415 on Build GRCh38
rs958215643 Chr.10: 91485133-91485133 on Build GRCh38
rs367603140 Chr.10: 91485339-91485339 on Build GRCh38
rs1048501455 Chr.10: 91485491-91485491 on Build GRCh38
rs895262442 Chr.10: 91485492-91485492 on Build GRCh38
rs759523040 Chr.10: 91485325-91485325 on Build GRCh38
rs961796856 Chr.10: 91485443-91485443 on Build GRCh38
rs187058490 Chr.10: 91485431-91485431 on Build GRCh38
rs1003283552 Chr.10: 91485367-91485367 on Build GRCh38
rs778451899 Chr.10: 91485296-91485296 on Build GRCh38
rs954433920 Chr.10: 91485507-91485507 on Build GRCh38
rs756782847 Chr.10: 91485159-91485159 on Build GRCh38
rs191481603 Chr.10: 91485502-91485502 on Build GRCh38
rs1034641641 Chr.10: 91485402-91485402 on Build GRCh38
rs770248919 Chr.10: 91485281-91485281 on Build GRCh38
rs1030082340 Chr.10: 91485453-91485453 on Build GRCh38
rs774560079 Chr.10: 91485316-91485316 on Build GRCh38
rs763827980 Chr.10: 91485349-91485349 on Build GRCh38
rs1016411669 Chr.10: 91485438-91485438 on Build GRCh38
rs11186582 Chr.10: 91485519-91485519 on Build GRCh38
rs779557677 Chr.10: 91485219-91485219 on Build GRCh38
rs369939363 Chr.10: 91485157-91485157 on Build GRCh38
rs563523433 Chr.10: 91485452-91485452 on Build GRCh38
rs184177361 Chr.10: 91485364-91485364 on Build GRCh38
rs750047460 Chr.10: 91485191-91485191 on Build GRCh38
rs770924154 Chr.10: 91485313-91485313 on Build GRCh38
rs985569188 Chr.10: 91485395-91485395 on Build GRCh38

View Full Product Details