Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99695869-99696379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX15
Entrez Gene ID
1355
Gene Name
COX15, cytochrome c oxidase assembly homolog
Gene Aliases
CEMCOX2
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99694793-99732667 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1126904 c.954C>T p.N318N Chr.10: 99696066-99696066 on Build GRCh38
913777 c.972C>T p.Y324Y Chr.10: 99696084-99696084 on Build GRCh38
2057416 c.899A>G p.H300R Chr.10: 99696011-99696011 on Build GRCh38
426908 c.883G>C p.G295R Chr.10: 99695995-99695995 on Build GRCh38
4010937 c.962G>A p.R321Q Chr.10: 99696074-99696074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750316046 Chr.10: 99695920-99695920 on Build GRCh38
rs749791682 Chr.10: 99695949-99695949 on Build GRCh38
rs939996966 Chr.10: 99696165-99696165 on Build GRCh38
rs756395067 Chr.10: 99696054-99696054 on Build GRCh38
rs767630171 Chr.10: 99696127-99696127 on Build GRCh38
rs777697527 Chr.10: 99696250-99696250 on Build GRCh38
rs771207556 Chr.10: 99696295-99696295 on Build GRCh38
rs1011275865 Chr.10: 99696109-99696109 on Build GRCh38
rs762868906 Chr.10: 99696100-99696100 on Build GRCh38
rs202230145 Chr.10: 99696073-99696073 on Build GRCh38
rs144074709 Chr.10: 99696084-99696084 on Build GRCh38
rs765096479 Chr.10: 99695988-99695988 on Build GRCh38
rs916679387 Chr.10: 99696188-99696191 on Build GRCh38
rs889951517 Chr.10: 99696252-99696252 on Build GRCh38
rs533453559 Chr.10: 99696307-99696307 on Build GRCh38
rs948046504 Chr.10: 99696199-99696199 on Build GRCh38
rs760237478 Chr.10: 99695963-99695963 on Build GRCh38
rs1018738513 Chr.10: 99696007-99696007 on Build GRCh38
rs770456537 Chr.10: 99696079-99696079 on Build GRCh38
rs992182370 Chr.10: 99696183-99696184 on Build GRCh38
rs901618466 Chr.10: 99696284-99696284 on Build GRCh38
rs984779387 Chr.10: 99696201-99696201 on Build GRCh38
rs1049845441 Chr.10: 99696242-99696242 on Build GRCh38
rs770452082 Chr.10: 99695968-99695968 on Build GRCh38
rs201208234 Chr.10: 99696136-99696136 on Build GRCh38
rs543727175 Chr.10: 99695946-99695946 on Build GRCh38
rs1036978236 Chr.10: 99695976-99695976 on Build GRCh38
rs528317677 Chr.10: 99696155-99696155 on Build GRCh38
rs146820785 Chr.10: 99696121-99696121 on Build GRCh38
rs746457576 Chr.10: 99696074-99696074 on Build GRCh38
rs898715656 Chr.10: 99696209-99696209 on Build GRCh38
rs561996054 Chr.10: 99696021-99696021 on Build GRCh38
rs764094290 Chr.10: 99696014-99696014 on Build GRCh38
rs139305536 Chr.10: 99696058-99696058 on Build GRCh38
rs61744356 Chr.10: 99696057-99696057 on Build GRCh38
rs541215340 Chr.10: 99696148-99696148 on Build GRCh38
rs987855333 Chr.10: 99696022-99696022 on Build GRCh38
rs888891964 Chr.10: 99696339-99696339 on Build GRCh38
rs1035895337 Chr.10: 99696157-99696157 on Build GRCh38
rs767359204 Chr.10: 99695911-99695914 on Build GRCh38
rs768978614 Chr.10: 99696066-99696066 on Build GRCh38
rs761695011 Chr.10: 99696113-99696113 on Build GRCh38
rs759313528 Chr.10: 99695986-99695986 on Build GRCh38
rs1005034756 Chr.10: 99695950-99695950 on Build GRCh38
rs916543766 Chr.10: 99695896-99695896 on Build GRCh38
rs545985868 Chr.10: 99695931-99695931 on Build GRCh38
rs369563076 Chr.10: 99696011-99696011 on Build GRCh38
rs188123130 Chr.10: 99696262-99696262 on Build GRCh38
rs1008386738 Chr.10: 99696258-99696258 on Build GRCh38
rs758988340 Chr.10: 99696237-99696237 on Build GRCh38
rs370070544 Chr.10: 99696166-99696166 on Build GRCh38
rs747121848 Chr.10: 99695906-99695906 on Build GRCh38
rs769580214 Chr.10: 99696085-99696085 on Build GRCh38
rs112158716 Chr.10: 99696098-99696098 on Build GRCh38
rs752609712 Chr.10: 99696002-99696002 on Build GRCh38
rs7919268 Chr.10: 99695920-99695920 on Build GRCh38
rs201693312 Chr.10: 99696140-99696140 on Build GRCh38
rs750612877 Chr.10: 99696042-99696042 on Build GRCh38
rs770120495 Chr.10: 99695941-99695941 on Build GRCh38
rs373504480 Chr.10: 99696039-99696039 on Build GRCh38
rs775354653 Chr.10: 99696095-99696095 on Build GRCh38
rs765942072 Chr.10: 99695977-99695977 on Build GRCh38
rs772540029 Chr.10: 99695962-99695962 on Build GRCh38
rs769210199 Chr.10: 99695961-99695961 on Build GRCh38
rs933488162 Chr.10: 99696292-99696292 on Build GRCh38
rs901983747 Chr.10: 99696283-99696283 on Build GRCh38
rs776370975 Chr.10: 99695978-99695978 on Build GRCh38
rs1050158777 Chr.10: 99696313-99696313 on Build GRCh38
rs1021265840 Chr.10: 99696044-99696044 on Build GRCh38
rs763886692 Chr.10: 99696103-99696103 on Build GRCh38
rs753316658 Chr.10: 99696037-99696038 on Build GRCh38
rs34801570 Chr.10: 99696244-99696245 on Build GRCh38
rs746168281 Chr.10: 99695938-99695938 on Build GRCh38
rs931574668 Chr.10: 99696239-99696239 on Build GRCh38
rs751490406 Chr.10: 99696108-99696108 on Build GRCh38
rs200492698 Chr.10: 99696080-99696080 on Build GRCh38
rs757378688 Chr.10: 99696031-99696031 on Build GRCh38
rs781505876 Chr.10: 99695921-99695921 on Build GRCh38

View Full Product Details