Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102835165-102835661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP17A1
Entrez Gene ID
1586
Gene Name
cytochrome P450 family 17 subfamily A member 1
Gene Aliases
CPT7, CYP17, P450C17, S17AH
UniGene
Hs.438016
Gene Chromosome Location
Chr.10: 102830531-102837533 on Build GRCh38
Transcripts
NM_000102.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6577700 c.395T>A p.F132Y Chr.10: 102835295-102835295 on Build GRCh38
465208 c.340T>C p.F114L Chr.10: 102835350-102835350 on Build GRCh38
6905862 c.350_351delCT p.S117fs*38 Chr.10: 102835339-102835340 on Build GRCh38
4675868 c.387G>T p.M129I Chr.10: 102835303-102835303 on Build GRCh38
6323109 c.326G>A p.R109H Chr.10: 102835364-102835364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188885755 Chr.10: 102835403-102835403 on Build GRCh38
rs1034339180 Chr.10: 102835452-102835452 on Build GRCh38
rs552673286 Chr.10: 102835500-102835500 on Build GRCh38
rs779930218 Chr.10: 102835238-102835238 on Build GRCh38
rs1051558297 Chr.10: 102835189-102835189 on Build GRCh38
rs756371704 Chr.10: 102835235-102835235 on Build GRCh38
rs551992399 Chr.10: 102835568-102835568 on Build GRCh38
rs201386446 Chr.10: 102835640-102835640 on Build GRCh38
rs774228870 Chr.10: 102835348-102835348 on Build GRCh38
rs139936404 Chr.10: 102835376-102835376 on Build GRCh38
rs767158887 Chr.10: 102835357-102835357 on Build GRCh38
rs745456299 Chr.10: 102835309-102835309 on Build GRCh38
rs755526392 Chr.10: 102835275-102835275 on Build GRCh38
rs750557590 Chr.10: 102835227-102835227 on Build GRCh38
rs104894135 Chr.10: 102835374-102835374 on Build GRCh38
rs768102031 Chr.10: 102835437-102835437 on Build GRCh38
rs976046546 Chr.10: 102835615-102835615 on Build GRCh38
rs767372790 Chr.10: 102835208-102835208 on Build GRCh38
rs369461376 Chr.10: 102835307-102835307 on Build GRCh38
rs780614918 Chr.10: 102835236-102835244 on Build GRCh38
rs1001521465 Chr.10: 102835414-102835414 on Build GRCh38
rs372791822 Chr.10: 102835401-102835401 on Build GRCh38
rs786205061 Chr.10: 102835249-102835249 on Build GRCh38
rs112470177 Chr.10: 102835430-102835430 on Build GRCh38
rs104894148 Chr.10: 102835343-102835343 on Build GRCh38
rs766679884 Chr.10: 102835372-102835372 on Build GRCh38
rs745351748 Chr.10: 102835257-102835259 on Build GRCh38
rs1039360325 Chr.10: 102835599-102835599 on Build GRCh38
rs750236382 Chr.10: 102835365-102835365 on Build GRCh38
rs3020717 Chr.10: 102835489-102835489 on Build GRCh38
rs775111286 Chr.10: 102835407-102835407 on Build GRCh38
rs747477069 Chr.10: 102835383-102835383 on Build GRCh38
rs4919687 Chr.10: 102835491-102835491 on Build GRCh38
rs966427411 Chr.10: 102835549-102835549 on Build GRCh38
rs943205349 Chr.10: 102835593-102835593 on Build GRCh38
rs61754271 Chr.10: 102835204-102835204 on Build GRCh38
rs769319989 Chr.10: 102835385-102835385 on Build GRCh38
rs770495392 Chr.10: 102835229-102835230 on Build GRCh38
rs951562762 Chr.10: 102835525-102835525 on Build GRCh38
rs768546290 Chr.10: 102835337-102835337 on Build GRCh38
rs777625933 Chr.10: 102835486-102835486 on Build GRCh38
rs534015171 Chr.10: 102835454-102835454 on Build GRCh38
rs779613920 Chr.10: 102835276-102835276 on Build GRCh38
rs774980374 Chr.10: 102835335-102835335 on Build GRCh38
rs964270616 Chr.10: 102835559-102835559 on Build GRCh38
rs777937209 Chr.10: 102835378-102835378 on Build GRCh38
rs987232876 Chr.10: 102835565-102835565 on Build GRCh38
rs748494285 Chr.10: 102835277-102835277 on Build GRCh38
rs760801519 Chr.10: 102835367-102835367 on Build GRCh38
rs35115800 Chr.10: 102835429-102835430 on Build GRCh38
rs769502745 Chr.10: 102835289-102835290 on Build GRCh38
rs104894147 Chr.10: 102835350-102835350 on Build GRCh38
rs942376359 Chr.10: 102835328-102835328 on Build GRCh38
rs989126543 Chr.10: 102835501-102835501 on Build GRCh38
rs529909202 Chr.10: 102835211-102835211 on Build GRCh38
rs755271787 Chr.10: 102835191-102835191 on Build GRCh38
rs1038978276 Chr.10: 102835224-102835224 on Build GRCh38
rs104894154 Chr.10: 102835316-102835316 on Build GRCh38
rs772285507 Chr.10: 102835301-102835301 on Build GRCh38
rs758961949 Chr.10: 102835377-102835377 on Build GRCh38
rs754036409 Chr.10: 102835261-102835261 on Build GRCh38
rs550594217 Chr.10: 102835353-102835353 on Build GRCh38
rs913561550 Chr.10: 102835508-102835508 on Build GRCh38
rs912846063 Chr.10: 102835554-102835554 on Build GRCh38
rs548446966 Chr.10: 102835354-102835354 on Build GRCh38
rs911713161 Chr.10: 102835584-102835584 on Build GRCh38
rs570171570 Chr.10: 102835616-102835616 on Build GRCh38
rs1001724526 Chr.10: 102835446-102835446 on Build GRCh38
rs905111432 Chr.10: 102835408-102835408 on Build GRCh38
rs150698224 Chr.10: 102835375-102835375 on Build GRCh38
rs963013282 Chr.10: 102835477-102835477 on Build GRCh38
rs773859640 Chr.10: 102835438-102835438 on Build GRCh38
rs754000413 Chr.10: 102835591-102835591 on Build GRCh38
rs761632949 Chr.10: 102835206-102835206 on Build GRCh38

View Full Product Details